Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 22451153-22451678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRTA1
Entrez Gene ID
63951
Gene Name
DMRT like family A1
Gene Aliases
DMO, DMRT4
UniGene
Hs.371976
Gene Chromosome Location
Chr.9: 22446841-22452473 on Build GRCh38
Transcripts
NM_022160.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3092369 c.793T>C p.S265P Chr.9: 22451189-22451189 on Build GRCh38
5803712 c.794C>T p.S265L Chr.9: 22451190-22451190 on Build GRCh38
6645706 c.1049G>A p.R350Q Chr.9: 22451445-22451445 on Build GRCh38
4425581 c.794C>G p.S265* Chr.9: 22451190-22451190 on Build GRCh38
3382503 c.905C>T p.S302L Chr.9: 22451301-22451301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745339156 Chr.9: 22451294-22451294 on Build GRCh38
rs202197256 Chr.9: 22451350-22451350 on Build GRCh38
rs767960409 Chr.9: 22451600-22451600 on Build GRCh38
rs777522680 Chr.9: 22451191-22451191 on Build GRCh38
rs752706554 Chr.9: 22451237-22451237 on Build GRCh38
rs891099294 Chr.9: 22451421-22451421 on Build GRCh38
rs756763306 Chr.9: 22451195-22451195 on Build GRCh38
rs368798749 Chr.9: 22451328-22451328 on Build GRCh38
rs201374002 Chr.9: 22451232-22451232 on Build GRCh38
rs899760825 Chr.9: 22451387-22451387 on Build GRCh38
rs781618672 Chr.9: 22451513-22451513 on Build GRCh38
rs763988468 Chr.9: 22451256-22451256 on Build GRCh38
rs112667674 Chr.9: 22451372-22451372 on Build GRCh38
rs144441630 Chr.9: 22451552-22451552 on Build GRCh38
rs773060818 Chr.9: 22451652-22451652 on Build GRCh38
rs137914267 Chr.9: 22451356-22451356 on Build GRCh38
rs372597372 Chr.9: 22451414-22451414 on Build GRCh38
rs962865767 Chr.9: 22451233-22451233 on Build GRCh38
rs565930227 Chr.9: 22451465-22451465 on Build GRCh38
rs372873186 Chr.9: 22451420-22451420 on Build GRCh38
rs61729546 Chr.9: 22451340-22451340 on Build GRCh38
rs1015081364 Chr.9: 22451267-22451267 on Build GRCh38
rs74769337 Chr.9: 22451228-22451228 on Build GRCh38
rs140491258 Chr.9: 22451515-22451515 on Build GRCh38
rs746675446 Chr.9: 22451193-22451193 on Build GRCh38
rs551088682 Chr.9: 22451363-22451363 on Build GRCh38
rs769979537 Chr.9: 22451426-22451426 on Build GRCh38
rs181512603 Chr.9: 22451185-22451185 on Build GRCh38
rs772447910 Chr.9: 22451222-22451222 on Build GRCh38
rs755229177 Chr.9: 22451542-22451542 on Build GRCh38
rs751669342 Chr.9: 22451412-22451412 on Build GRCh38
rs760432570 Chr.9: 22451558-22451558 on Build GRCh38
rs761466154 Chr.9: 22451446-22451446 on Build GRCh38
rs772020536 Chr.9: 22451635-22451635 on Build GRCh38
rs750111980 Chr.9: 22451380-22451380 on Build GRCh38
rs201465667 Chr.9: 22451419-22451419 on Build GRCh38
rs776947180 Chr.9: 22451349-22451349 on Build GRCh38
rs771032631 Chr.9: 22451348-22451348 on Build GRCh38
rs756339037 Chr.9: 22451525-22451525 on Build GRCh38
rs757500819 Chr.9: 22451503-22451503 on Build GRCh38
rs767356686 Chr.9: 22451457-22451457 on Build GRCh38
rs541856132 Chr.9: 22451459-22451459 on Build GRCh38
rs369778210 Chr.9: 22451358-22451358 on Build GRCh38
rs779129735 Chr.9: 22451627-22451627 on Build GRCh38
rs777892111 Chr.9: 22451281-22451283 on Build GRCh38
rs375053235 Chr.9: 22451550-22451550 on Build GRCh38
rs34324424 Chr.9: 22451230-22451230 on Build GRCh38
rs758459142 Chr.9: 22451178-22451179 on Build GRCh38
rs780465193 Chr.9: 22451530-22451530 on Build GRCh38
rs368374702 Chr.9: 22451376-22451376 on Build GRCh38
rs1041549610 Chr.9: 22451182-22451182 on Build GRCh38
rs745441990 Chr.9: 22451204-22451204 on Build GRCh38
rs780375295 Chr.9: 22451618-22451618 on Build GRCh38
rs768684480 Chr.9: 22451444-22451444 on Build GRCh38
rs372803240 Chr.9: 22451611-22451611 on Build GRCh38
rs148928674 Chr.9: 22451593-22451593 on Build GRCh38
rs773362908 Chr.9: 22451223-22451223 on Build GRCh38
rs147178627 Chr.9: 22451502-22451502 on Build GRCh38
rs748120845 Chr.9: 22451543-22451543 on Build GRCh38
rs898054099 Chr.9: 22451509-22451509 on Build GRCh38
rs143713234 Chr.9: 22451203-22451203 on Build GRCh38
rs376989039 Chr.9: 22451619-22451619 on Build GRCh38
rs751379079 Chr.9: 22451369-22451369 on Build GRCh38
rs762982635 Chr.9: 22451362-22451362 on Build GRCh38
rs753740634 Chr.9: 22451181-22451181 on Build GRCh38
rs368202520 Chr.9: 22451445-22451445 on Build GRCh38
rs371496399 Chr.9: 22451384-22451384 on Build GRCh38
rs370009723 Chr.9: 22451598-22451598 on Build GRCh38
rs936560925 Chr.9: 22451215-22451215 on Build GRCh38
rs758212329 Chr.9: 22451239-22451239 on Build GRCh38
rs866640006 Chr.9: 22451474-22451474 on Build GRCh38
rs780948456 Chr.9: 22451287-22451287 on Build GRCh38
rs150162716 Chr.9: 22451585-22451585 on Build GRCh38
rs142995523 Chr.9: 22451270-22451270 on Build GRCh38
rs777036865 Chr.9: 22451211-22451211 on Build GRCh38
rs1029635429 Chr.9: 22451212-22451212 on Build GRCh38
rs775700148 Chr.9: 22451427-22451427 on Build GRCh38
rs371171820 Chr.9: 22451344-22451344 on Build GRCh38
rs754863697 Chr.9: 22451183-22451183 on Build GRCh38
rs371769774 Chr.9: 22451496-22451496 on Build GRCh38
rs748232506 Chr.9: 22451633-22451633 on Build GRCh38
rs768880615 Chr.9: 22451539-22451539 on Build GRCh38
rs267602201 Chr.9: 22451304-22451304 on Build GRCh38
rs1006426815 Chr.9: 22451266-22451266 on Build GRCh38
rs771947601 Chr.9: 22451268-22451268 on Build GRCh38
rs186108346 Chr.9: 22451639-22451639 on Build GRCh38
rs766076687 Chr.9: 22451472-22451472 on Build GRCh38
rs952326505 Chr.9: 22451259-22451259 on Build GRCh38
rs757771658 Chr.9: 22451599-22451599 on Build GRCh38
rs757632938 Chr.9: 22451533-22451534 on Build GRCh38
rs145152745 Chr.9: 22451278-22451278 on Build GRCh38
rs748471195 Chr.9: 22451220-22451220 on Build GRCh38
rs949805564 Chr.9: 22451354-22451354 on Build GRCh38
rs771353848 Chr.9: 22451417-22451417 on Build GRCh38
rs138782225 Chr.9: 22451402-22451402 on Build GRCh38
rs913826524 Chr.9: 22451647-22451647 on Build GRCh38
rs574289690 Chr.9: 22451533-22451533 on Build GRCh38
rs1001508505 Chr.9: 22451244-22451244 on Build GRCh38
rs758992139 Chr.9: 22451589-22451589 on Build GRCh38
rs142702676 Chr.9: 22451606-22451606 on Build GRCh38
rs760888914 Chr.9: 22451227-22451227 on Build GRCh38
rs536790652 Chr.9: 22451553-22451553 on Build GRCh38
rs139030144 Chr.9: 22451338-22451338 on Build GRCh38
rs754519600 Chr.9: 22451493-22451493 on Build GRCh38
rs368428828 Chr.9: 22451217-22451217 on Build GRCh38
rs1033333947 Chr.9: 22451188-22451188 on Build GRCh38
rs1034283321 Chr.9: 22451248-22451248 on Build GRCh38
rs150709609 Chr.9: 22451343-22451343 on Build GRCh38
rs779618649 Chr.9: 22451392-22451392 on Build GRCh38
rs758885818 Chr.9: 22451409-22451409 on Build GRCh38
rs756162445 Chr.9: 22451584-22451584 on Build GRCh38
rs753320911 Chr.9: 22451475-22451475 on Build GRCh38
rs200749823 Chr.9: 22451385-22451385 on Build GRCh38
rs779238351 Chr.9: 22451324-22451324 on Build GRCh38
rs148944998 Chr.9: 22451186-22451186 on Build GRCh38
rs755681561 Chr.9: 22451301-22451301 on Build GRCh38
rs932569296 Chr.9: 22451604-22451604 on Build GRCh38
rs747188563 Chr.9: 22451456-22451456 on Build GRCh38
rs199917145 Chr.9: 22451511-22451511 on Build GRCh38
rs35310697 Chr.9: 22451582-22451582 on Build GRCh38
rs763175158 Chr.9: 22451428-22451428 on Build GRCh38
rs996752450 Chr.9: 22451400-22451400 on Build GRCh38

View Full Product Details