Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 197510095-197510677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND1B
Entrez Gene ID
163486
Gene Name
DENN domain containing 1B
Gene Aliases
C1ORF18, C1orf218, FAM31B
UniGene
Hs.125056
Gene Chromosome Location
Chr.1: 197504748-197782138 on Build GRCh38
Transcripts
NM_001195215.1 NR_125340.1 XM_005244931.2 XM_006711192.2 XM_006711193.3 XM_006711194.3 XM_011509246.2 XM_011509247.1 XM_011509248.2 XM_011509249.2 XM_017000470.1 XM_017000471.1 XM_017000472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
678300 c.932A>G p.H311R Chr.1: 197510548-197510548 on Build GRCh38
678299 c.824C>T p.S275L Chr.1: 197510656-197510656 on Build GRCh38
1668335 c.977G>T p.S326I Chr.1: 197510503-197510503 on Build GRCh38
5529672 c.874C>T p.P292S Chr.1: 197510606-197510606 on Build GRCh38
2125652 c.872T>C p.V291A Chr.1: 197510608-197510608 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779523726 Chr.1: 197510637-197510637 on Build GRCh38
rs772355116 Chr.1: 197510578-197510578 on Build GRCh38
rs1055731564 Chr.1: 197510216-197510216 on Build GRCh38
rs759652031 Chr.1: 197510613-197510613 on Build GRCh38
rs773379564 Chr.1: 197510606-197510606 on Build GRCh38
rs373439877 Chr.1: 197510586-197510586 on Build GRCh38
rs1421389 Chr.1: 197510248-197510248 on Build GRCh38
rs963911994 Chr.1: 197510231-197510231 on Build GRCh38
rs774855452 Chr.1: 197510453-197510453 on Build GRCh38
rs1008643699 Chr.1: 197510374-197510374 on Build GRCh38
rs551124265 Chr.1: 197510459-197510459 on Build GRCh38
rs7554658 Chr.1: 197510240-197510240 on Build GRCh38
rs1033215170 Chr.1: 197510122-197510122 on Build GRCh38
rs762220808 Chr.1: 197510567-197510567 on Build GRCh38
rs569364308 Chr.1: 197510482-197510482 on Build GRCh38
rs772425181 Chr.1: 197510639-197510639 on Build GRCh38
rs551836552 Chr.1: 197510616-197510616 on Build GRCh38
rs1034586041 Chr.1: 197510651-197510651 on Build GRCh38
rs868016263 Chr.1: 197510549-197510549 on Build GRCh38
rs201905153 Chr.1: 197510352-197510353 on Build GRCh38
rs188581022 Chr.1: 197510232-197510232 on Build GRCh38
rs767757248 Chr.1: 197510460-197510460 on Build GRCh38
rs1047512383 Chr.1: 197510416-197510416 on Build GRCh38
rs930521496 Chr.1: 197510214-197510214 on Build GRCh38
rs769258926 Chr.1: 197510559-197510559 on Build GRCh38
rs908604842 Chr.1: 197510186-197510186 on Build GRCh38
rs777754291 Chr.1: 197510510-197510510 on Build GRCh38
rs573867563 Chr.1: 197510196-197510196 on Build GRCh38
rs571421121 Chr.1: 197510224-197510224 on Build GRCh38
rs561880998 Chr.1: 197510325-197510325 on Build GRCh38
rs376231862 Chr.1: 197510422-197510422 on Build GRCh38
rs1028519835 Chr.1: 197510349-197510349 on Build GRCh38
rs748368090 Chr.1: 197510143-197510143 on Build GRCh38
rs994149593 Chr.1: 197510195-197510195 on Build GRCh38
rs1008110837 Chr.1: 197510218-197510218 on Build GRCh38
rs555542969 Chr.1: 197510183-197510183 on Build GRCh38
rs768304710 Chr.1: 197510636-197510636 on Build GRCh38
rs748788909 Chr.1: 197510632-197510632 on Build GRCh38
rs1045440016 Chr.1: 197510237-197510237 on Build GRCh38
rs746999013 Chr.1: 197510577-197510578 on Build GRCh38
rs528512561 Chr.1: 197510319-197510319 on Build GRCh38
rs768426692 Chr.1: 197510610-197510610 on Build GRCh38
rs374881007 Chr.1: 197510476-197510476 on Build GRCh38
rs991342886 Chr.1: 197510544-197510544 on Build GRCh38
rs920026106 Chr.1: 197510361-197510361 on Build GRCh38
rs930031363 Chr.1: 197510367-197510367 on Build GRCh38
rs780851787 Chr.1: 197510623-197510623 on Build GRCh38
rs200343795 Chr.1: 197510524-197510524 on Build GRCh38
rs917558681 Chr.1: 197510271-197510271 on Build GRCh38
rs540329984 Chr.1: 197510149-197510149 on Build GRCh38
rs920436274 Chr.1: 197510205-197510205 on Build GRCh38
rs143462572 Chr.1: 197510627-197510627 on Build GRCh38
rs934382589 Chr.1: 197510620-197510620 on Build GRCh38
rs769278385 Chr.1: 197510527-197510527 on Build GRCh38
rs753909022 Chr.1: 197510612-197510612 on Build GRCh38
rs538571059 Chr.1: 197510594-197510594 on Build GRCh38
rs975639693 Chr.1: 197510158-197510158 on Build GRCh38
rs937541451 Chr.1: 197510323-197510323 on Build GRCh38
rs749719405 Chr.1: 197510177-197510177 on Build GRCh38
rs748876848 Chr.1: 197510550-197510550 on Build GRCh38
rs745534795 Chr.1: 197510525-197510525 on Build GRCh38
rs572882880 Chr.1: 197510127-197510127 on Build GRCh38
rs779800670 Chr.1: 197510630-197510630 on Build GRCh38
rs951844156 Chr.1: 197510528-197510528 on Build GRCh38
rs955645906 Chr.1: 197510362-197510362 on Build GRCh38
rs564092378 Chr.1: 197510287-197510287 on Build GRCh38
rs1028256504 Chr.1: 197510160-197510160 on Build GRCh38
rs761601269 Chr.1: 197510638-197510638 on Build GRCh38
rs1033556395 Chr.1: 197510308-197510308 on Build GRCh38
rs779418746 Chr.1: 197510546-197510546 on Build GRCh38
rs766679906 Chr.1: 197510597-197510597 on Build GRCh38
rs757003485 Chr.1: 197510621-197510621 on Build GRCh38
rs750633201 Chr.1: 197510466-197510466 on Build GRCh38
rs371428108 Chr.1: 197510516-197510516 on Build GRCh38
rs972060314 Chr.1: 197510273-197510273 on Build GRCh38
rs751324450 Chr.1: 197510515-197510515 on Build GRCh38
rs760780267 Chr.1: 197510424-197510424 on Build GRCh38
rs758317813 Chr.1: 197510503-197510503 on Build GRCh38
rs577884174 Chr.1: 197510560-197510560 on Build GRCh38
rs1032123013 Chr.1: 197510239-197510239 on Build GRCh38
rs949731986 Chr.1: 197510228-197510228 on Build GRCh38
rs972302754 Chr.1: 197510203-197510203 on Build GRCh38
rs371100267 Chr.1: 197510619-197510619 on Build GRCh38
rs879521599 Chr.1: 197510227-197510227 on Build GRCh38
rs73073636 Chr.1: 197510320-197510320 on Build GRCh38
rs200236372 Chr.1: 197510593-197510593 on Build GRCh38
rs764759028 Chr.1: 197510443-197510443 on Build GRCh38
rs141712038 Chr.1: 197510469-197510469 on Build GRCh38
rs773433361 Chr.1: 197510645-197510645 on Build GRCh38
rs199787690 Chr.1: 197510419-197510419 on Build GRCh38
rs140620253 Chr.1: 197510626-197510626 on Build GRCh38
rs779363166 Chr.1: 197510503-197510503 on Build GRCh38
rs529423137 Chr.1: 197510379-197510379 on Build GRCh38

View Full Product Details