Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 13087678-13088200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSG1
Entrez Gene ID
83445
Gene Name
germ cell associated 1
Gene Aliases
-
UniGene
Hs.240053
Gene Chromosome Location
Chr.12: 13083537-13103720 on Build GRCh38
Transcripts
NM_001080554.2 NM_001080555.2 NM_001206842.1 NM_001206843.1 NM_001206845.1 NM_031289.3 NM_153823.3 XM_005253493.2 XM_005253495.1 XM_011520858.1 XM_011520859.1 XM_011520860.1 XM_011520861.1 XM_011520862.1 XM_011520863.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4146844 c.694C>G p.H232D Chr.12: 13087971-13087971 on Build GRCh38
3772324 c.490G>C p.D164H Chr.12: 13087982-13087982 on Build GRCh38
3458205 c.639C>T p.L213L Chr.12: 13087918-13087918 on Build GRCh38
5374654 c.644C>T p.P215L Chr.12: 13087913-13087913 on Build GRCh38
3458203 c.554C>T p.S185F Chr.12: 13087918-13087918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1022946980 Chr.12: 13088120-13088120 on Build GRCh38
rs779133829 Chr.12: 13088064-13088064 on Build GRCh38
rs759050824 Chr.12: 13087954-13087954 on Build GRCh38
rs748866035 Chr.12: 13088090-13088090 on Build GRCh38
rs185497885 Chr.12: 13088153-13088153 on Build GRCh38
rs999461387 Chr.12: 13087748-13087748 on Build GRCh38
rs973899937 Chr.12: 13088177-13088177 on Build GRCh38
rs933336414 Chr.12: 13087843-13087843 on Build GRCh38
rs1015502049 Chr.12: 13088152-13088152 on Build GRCh38
rs974345384 Chr.12: 13087902-13087902 on Build GRCh38
rs1044345490 Chr.12: 13087888-13087888 on Build GRCh38
rs757979765 Chr.12: 13088087-13088087 on Build GRCh38
rs950880640 Chr.12: 13087708-13087708 on Build GRCh38
rs919768301 Chr.12: 13088178-13088178 on Build GRCh38
rs150092885 Chr.12: 13088157-13088157 on Build GRCh38
rs779248084 Chr.12: 13087984-13087987 on Build GRCh38
rs767700047 Chr.12: 13088021-13088021 on Build GRCh38
rs748756709 Chr.12: 13088070-13088070 on Build GRCh38
rs752546274 Chr.12: 13087860-13087860 on Build GRCh38
rs981469623 Chr.12: 13088150-13088150 on Build GRCh38
rs576580471 Chr.12: 13087841-13087841 on Build GRCh38
rs745397183 Chr.12: 13088079-13088079 on Build GRCh38
rs750700209 Chr.12: 13087887-13087887 on Build GRCh38
rs748581818 Chr.12: 13087949-13087949 on Build GRCh38
rs751182064 Chr.12: 13088137-13088137 on Build GRCh38
rs535884054 Chr.12: 13087844-13087844 on Build GRCh38
rs138229660 Chr.12: 13087816-13087816 on Build GRCh38
rs1053359637 Chr.12: 13087877-13087877 on Build GRCh38
rs769586584 Chr.12: 13087957-13087957 on Build GRCh38
rs1013016904 Chr.12: 13088118-13088118 on Build GRCh38
rs780181672 Chr.12: 13087912-13087912 on Build GRCh38
rs199682106 Chr.12: 13087886-13087886 on Build GRCh38
rs769392025 Chr.12: 13088086-13088086 on Build GRCh38
rs369468787 Chr.12: 13088043-13088043 on Build GRCh38
rs760379909 Chr.12: 13087837-13087838 on Build GRCh38
rs750001321 Chr.12: 13088019-13088019 on Build GRCh38
rs200316271 Chr.12: 13087882-13087882 on Build GRCh38
rs903335668 Chr.12: 13087731-13087731 on Build GRCh38
rs529034043 Chr.12: 13087732-13087732 on Build GRCh38
rs115931320 Chr.12: 13087935-13087935 on Build GRCh38
rs750502868 Chr.12: 13088027-13088027 on Build GRCh38
rs553580521 Chr.12: 13088072-13088072 on Build GRCh38
rs775237533 Chr.12: 13087959-13087959 on Build GRCh38
rs764117975 Chr.12: 13087872-13087872 on Build GRCh38
rs991447434 Chr.12: 13087845-13087845 on Build GRCh38
rs113026236 Chr.12: 13088062-13088062 on Build GRCh38
rs763769757 Chr.12: 13087977-13087977 on Build GRCh38
rs1033092125 Chr.12: 13087836-13087836 on Build GRCh38
rs886186654 Chr.12: 13087955-13087955 on Build GRCh38
rs200576038 Chr.12: 13088092-13088092 on Build GRCh38
rs1022978236 Chr.12: 13087854-13087854 on Build GRCh38
rs919646957 Chr.12: 13087910-13087910 on Build GRCh38
rs569643105 Chr.12: 13087907-13087907 on Build GRCh38
rs768853667 Chr.12: 13087937-13087937 on Build GRCh38
rs372652289 Chr.12: 13087868-13087868 on Build GRCh38
rs780274796 Chr.12: 13088035-13088035 on Build GRCh38
rs140866058 Chr.12: 13088123-13088123 on Build GRCh38
rs762780657 Chr.12: 13087863-13087863 on Build GRCh38
rs756127404 Chr.12: 13088034-13088034 on Build GRCh38
rs376340949 Chr.12: 13087938-13087938 on Build GRCh38
rs567395894 Chr.12: 13087953-13087953 on Build GRCh38
rs575257876 Chr.12: 13088133-13088133 on Build GRCh38
rs770532410 Chr.12: 13087952-13087952 on Build GRCh38
rs539330924 Chr.12: 13088116-13088116 on Build GRCh38
rs138940005 Chr.12: 13088018-13088018 on Build GRCh38
rs762876993 Chr.12: 13087974-13087974 on Build GRCh38
rs751415527 Chr.12: 13087881-13087881 on Build GRCh38
rs369932802 Chr.12: 13088078-13088078 on Build GRCh38
rs984018964 Chr.12: 13087729-13087729 on Build GRCh38
rs770594971 Chr.12: 13088055-13088055 on Build GRCh38
rs932401901 Chr.12: 13087922-13087922 on Build GRCh38

View Full Product Details