Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49642948-49643547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMNL3
Entrez Gene ID
91010
Gene Name
formin like 3
Gene Aliases
FHOD3, FRL2, WBP-3, WBP3
UniGene
Hs.179838
Gene Chromosome Location
Chr.12: 49636499-49707414 on Build GRCh38
Transcripts
NM_175736.4 NM_198900.2 XM_005269218.2 XM_011538968.2 XM_011538969.2 XM_011538970.2 XM_011538971.2 XM_011538972.2 XM_011538973.2 XM_011538974.2 NM_001031698.2 NM_012272.2 XM_006719324.3 XM_006719325.3 XM_011538135.2 XM_011538136.2 XM_011538137.2 XM_011538138.2 XM_011538139.2 XM_011538140.2 XM_011538141.1 XM_011538143.2 XM_011538144.2 XM_017019135.1 XM_017019136.1 XM_017019137.1 XM_017019138.1 XM_017019139.1

Gene Symbol
PRPF40B
Entrez Gene ID
25766
Gene Name
pre-mRNA processing factor 40 homolog B
Gene Aliases
HYPC
UniGene
Hs.706827
Gene Chromosome Location
Chr.12: 49622717-49644669 on Build GRCh38
Transcripts
NM_175736.4 NM_198900.2 XM_005269218.2 XM_011538968.2 XM_011538969.2 XM_011538970.2 XM_011538971.2 XM_011538972.2 XM_011538973.2 XM_011538974.2 NM_001031698.2 NM_012272.2 XM_006719324.3 XM_006719325.3 XM_011538135.2 XM_011538136.2 XM_011538137.2 XM_011538138.2 XM_011538139.2 XM_011538140.2 XM_011538141.1 XM_011538143.2 XM_011538144.2 XM_017019135.1 XM_017019136.1 XM_017019137.1 XM_017019138.1 XM_017019139.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6878670 c.2131C>A p.H711N Chr.12: 49643005-49643005 on Build GRCh38
1222171 c.2284C>T p.R762W Chr.12: 49643364-49643364 on Build GRCh38
1362144 c.2206C>T p.R736W Chr.12: 49643286-49643286 on Build GRCh38
1362143 c.2126G>A p.R709H Chr.12: 49643000-49643000 on Build GRCh38
4421873 c.2087G>A p.R696Q Chr.12: 49642961-49642961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565537155 Chr.12: 49643122-49643122 on Build GRCh38
rs762089237 Chr.12: 49643169-49643169 on Build GRCh38
rs145387088 Chr.12: 49643364-49643364 on Build GRCh38
rs751840640 Chr.12: 49643000-49643000 on Build GRCh38
rs752833704 Chr.12: 49642984-49642984 on Build GRCh38
rs531143131 Chr.12: 49643152-49643152 on Build GRCh38
rs755576038 Chr.12: 49643315-49643315 on Build GRCh38
rs768180891 Chr.12: 49643055-49643055 on Build GRCh38
rs200654119 Chr.12: 49642983-49642983 on Build GRCh38
rs374374885 Chr.12: 49643417-49643417 on Build GRCh38
rs1013236825 Chr.12: 49643218-49643218 on Build GRCh38
rs181509300 Chr.12: 49643456-49643456 on Build GRCh38
rs751539802 Chr.12: 49643228-49643228 on Build GRCh38
rs535872438 Chr.12: 49643312-49643312 on Build GRCh38
rs1045189056 Chr.12: 49643368-49643368 on Build GRCh38
rs997620361 Chr.12: 49643154-49643154 on Build GRCh38
rs202231084 Chr.12: 49643180-49643180 on Build GRCh38
rs974998628 Chr.12: 49643518-49643518 on Build GRCh38
rs773103374 Chr.12: 49643241-49643243 on Build GRCh38
rs758453530 Chr.12: 49642986-49642986 on Build GRCh38
rs186484553 Chr.12: 49643479-49643479 on Build GRCh38
rs773377400 Chr.12: 49643415-49643415 on Build GRCh38
rs973992003 Chr.12: 49643281-49643281 on Build GRCh38
rs762096751 Chr.12: 49643237-49643237 on Build GRCh38
rs1053969673 Chr.12: 49643463-49643463 on Build GRCh38
rs971148221 Chr.12: 49643026-49643026 on Build GRCh38
rs139701487 Chr.12: 49643108-49643108 on Build GRCh38
rs770746193 Chr.12: 49643273-49643273 on Build GRCh38
rs759800090 Chr.12: 49643427-49643427 on Build GRCh38
rs777227612 Chr.12: 49643332-49643332 on Build GRCh38
rs142161604 Chr.12: 49643240-49643240 on Build GRCh38
rs1032528815 Chr.12: 49643158-49643158 on Build GRCh38
rs914867773 Chr.12: 49643116-49643116 on Build GRCh38
rs770905357 Chr.12: 49643190-49643190 on Build GRCh38
rs764258978 Chr.12: 49643224-49643224 on Build GRCh38
rs892762254 Chr.12: 49643473-49643473 on Build GRCh38
rs904837183 Chr.12: 49643143-49643143 on Build GRCh38
rs189768718 Chr.12: 49643064-49643064 on Build GRCh38
rs761783061 Chr.12: 49643287-49643287 on Build GRCh38
rs550527975 Chr.12: 49643153-49643153 on Build GRCh38
rs769865188 Chr.12: 49643199-49643200 on Build GRCh38
rs780689981 Chr.12: 49643039-49643039 on Build GRCh38
rs936308388 Chr.12: 49643438-49643438 on Build GRCh38
rs760937022 Chr.12: 49643301-49643301 on Build GRCh38
rs777016432 Chr.12: 49643140-49643140 on Build GRCh38
rs1006706988 Chr.12: 49643326-49643326 on Build GRCh38
rs778244389 Chr.12: 49643057-49643057 on Build GRCh38
rs771815824 Chr.12: 49643271-49643271 on Build GRCh38
rs1051966927 Chr.12: 49643516-49643516 on Build GRCh38
rs567191228 Chr.12: 49643160-49643160 on Build GRCh38
rs772307302 Chr.12: 49643414-49643414 on Build GRCh38
rs990290046 Chr.12: 49643429-49643429 on Build GRCh38
rs752302137 Chr.12: 49643325-49643325 on Build GRCh38
rs756747246 Chr.12: 49643036-49643036 on Build GRCh38
rs114477497 Chr.12: 49643314-49643314 on Build GRCh38
rs767374932 Chr.12: 49642968-49642968 on Build GRCh38
rs775264410 Chr.12: 49643386-49643386 on Build GRCh38
rs201973529 Chr.12: 49643167-49643167 on Build GRCh38
rs759534743 Chr.12: 49643278-49643278 on Build GRCh38
rs371833248 Chr.12: 49643252-49643252 on Build GRCh38
rs757816284 Chr.12: 49643330-49643330 on Build GRCh38
rs773059866 Chr.12: 49643185-49643185 on Build GRCh38
rs1002534180 Chr.12: 49643411-49643411 on Build GRCh38
rs369382252 Chr.12: 49643201-49643201 on Build GRCh38
rs760612746 Chr.12: 49642978-49642978 on Build GRCh38
rs1022807456 Chr.12: 49643384-49643386 on Build GRCh38
rs754424825 Chr.12: 49643265-49643265 on Build GRCh38
rs764267443 Chr.12: 49642999-49642999 on Build GRCh38
rs115544158 Chr.12: 49643288-49643288 on Build GRCh38
rs765814015 Chr.12: 49643444-49643444 on Build GRCh38
rs1021814629 Chr.12: 49643446-49643446 on Build GRCh38
rs112086658 Chr.12: 49643282-49643282 on Build GRCh38
rs150633132 Chr.12: 49643027-49643027 on Build GRCh38
rs140773805 Chr.12: 49643321-49643321 on Build GRCh38
rs772935028 Chr.12: 49642977-49642977 on Build GRCh38
rs1009769829 Chr.12: 49643482-49643482 on Build GRCh38
rs760990489 Chr.12: 49643416-49643416 on Build GRCh38
rs774553692 Chr.12: 49643283-49643283 on Build GRCh38
rs971013304 Chr.12: 49643234-49643236 on Build GRCh38
rs755389795 Chr.12: 49643266-49643266 on Build GRCh38
rs769823189 Chr.12: 49643384-49643384 on Build GRCh38
rs543045643 Chr.12: 49643369-49643369 on Build GRCh38
rs780633306 Chr.12: 49643023-49643024 on Build GRCh38
rs149203454 Chr.12: 49643389-49643389 on Build GRCh38
rs151181775 Chr.12: 49643272-49643272 on Build GRCh38
rs369599079 Chr.12: 49643365-49643365 on Build GRCh38
rs748856142 Chr.12: 49643043-49643043 on Build GRCh38
rs748049386 Chr.12: 49643042-49643042 on Build GRCh38
rs199808082 Chr.12: 49642967-49642967 on Build GRCh38
rs114821665 Chr.12: 49643344-49643344 on Build GRCh38
rs775966493 Chr.12: 49643268-49643268 on Build GRCh38
rs754497081 Chr.12: 49643020-49643023 on Build GRCh38
rs114746574 Chr.12: 49643249-49643249 on Build GRCh38
rs750640824 Chr.12: 49643291-49643291 on Build GRCh38
rs376434775 Chr.12: 49643262-49643262 on Build GRCh38
rs200775632 Chr.12: 49643277-49643277 on Build GRCh38
rs192891339 Chr.12: 49643188-49643188 on Build GRCh38
rs566253760 Chr.12: 49643244-49643244 on Build GRCh38

View Full Product Details