Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37944579-37945173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C22orf23
Entrez Gene ID
84645
Gene Name
chromosome 22 open reading frame 23
Gene Aliases
EVG1, dJ1039K5.6
UniGene
Hs.517612
Gene Chromosome Location
Chr.22: 37943050-37953669 on Build GRCh38
Transcripts
NM_001207062.1 NM_032561.4 XM_005261781.1 XM_005261782.3 XM_005261783.2 XM_005261784.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1034079 c.374G>T p.R125I Chr.22: 37945149-37945149 on Build GRCh38
5921834 c.398G>A p.G133E Chr.22: 37945125-37945125 on Build GRCh38
3405665 c.440G>A p.R147Q Chr.22: 37945083-37945083 on Build GRCh38
6211047 c.431C>A p.P144H Chr.22: 37945092-37945092 on Build GRCh38
6389691 c.416G>A p.R139Q Chr.22: 37945107-37945107 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759822248 Chr.22: 37944627-37944627 on Build GRCh38
rs140055236 Chr.22: 37945075-37945075 on Build GRCh38
rs753561557 Chr.22: 37944626-37944626 on Build GRCh38
rs982349267 Chr.22: 37944735-37944735 on Build GRCh38
rs928224887 Chr.22: 37944776-37944776 on Build GRCh38
rs769600840 Chr.22: 37945140-37945140 on Build GRCh38
rs1033740767 Chr.22: 37944744-37944744 on Build GRCh38
rs760480322 Chr.22: 37945014-37945014 on Build GRCh38
rs571492654 Chr.22: 37944856-37944856 on Build GRCh38
rs6000947 Chr.22: 37944714-37944714 on Build GRCh38
rs772906646 Chr.22: 37945012-37945012 on Build GRCh38
rs766060211 Chr.22: 37945070-37945071 on Build GRCh38
rs745609325 Chr.22: 37945137-37945137 on Build GRCh38
rs888406855 Chr.22: 37944900-37944900 on Build GRCh38
rs1005269248 Chr.22: 37944702-37944702 on Build GRCh38
rs146578717 Chr.22: 37945053-37945053 on Build GRCh38
rs779071455 Chr.22: 37945107-37945107 on Build GRCh38
rs111556100 Chr.22: 37944991-37944991 on Build GRCh38
rs551250567 Chr.22: 37944899-37944900 on Build GRCh38
rs781332781 Chr.22: 37944873-37944873 on Build GRCh38
rs899800560 Chr.22: 37944866-37944866 on Build GRCh38
rs567298605 Chr.22: 37944818-37944818 on Build GRCh38
rs750258970 Chr.22: 37945039-37945040 on Build GRCh38
rs564782353 Chr.22: 37945054-37945054 on Build GRCh38
rs575089465 Chr.22: 37944899-37944899 on Build GRCh38
rs999408701 Chr.22: 37944899-37944902 on Build GRCh38
rs756465652 Chr.22: 37945036-37945036 on Build GRCh38
rs780301136 Chr.22: 37945041-37945041 on Build GRCh38
rs112866100 Chr.22: 37944790-37944790 on Build GRCh38
rs915374237 Chr.22: 37944768-37944768 on Build GRCh38
rs557080438 Chr.22: 37944891-37944891 on Build GRCh38
rs1012858270 Chr.22: 37944682-37944682 on Build GRCh38
rs998360411 Chr.22: 37945048-37945048 on Build GRCh38
rs779723763 Chr.22: 37944903-37944904 on Build GRCh38
rs374143684 Chr.22: 37944629-37944629 on Build GRCh38
rs553009373 Chr.22: 37944850-37944850 on Build GRCh38
rs748372730 Chr.22: 37944923-37944923 on Build GRCh38
rs776578637 Chr.22: 37945128-37945128 on Build GRCh38
rs534585370 Chr.22: 37944848-37944848 on Build GRCh38
rs3830772 Chr.22: 37944902-37944902 on Build GRCh38
rs954270464 Chr.22: 37944765-37944765 on Build GRCh38
rs1039775912 Chr.22: 37945064-37945064 on Build GRCh38
rs764924637 Chr.22: 37945083-37945083 on Build GRCh38
rs752186354 Chr.22: 37945029-37945029 on Build GRCh38
rs376250915 Chr.22: 37945099-37945099 on Build GRCh38
rs1010627629 Chr.22: 37944903-37944903 on Build GRCh38
rs200988522 Chr.22: 37945010-37945010 on Build GRCh38
rs771938191 Chr.22: 37945056-37945056 on Build GRCh38
rs542756193 Chr.22: 37944937-37944937 on Build GRCh38
rs769824320 Chr.22: 37945081-37945081 on Build GRCh38
rs571572959 Chr.22: 37944622-37944622 on Build GRCh38
rs370662580 Chr.22: 37944900-37944903 on Build GRCh38
rs958130767 Chr.22: 37944900-37944907 on Build GRCh38
rs1051611031 Chr.22: 37944880-37944880 on Build GRCh38
rs1047294777 Chr.22: 37944797-37944797 on Build GRCh38
rs5756860 Chr.22: 37944785-37944785 on Build GRCh38
rs565555420 Chr.22: 37944636-37944636 on Build GRCh38
rs1056446217 Chr.22: 37944857-37944857 on Build GRCh38
rs764834634 Chr.22: 37945028-37945028 on Build GRCh38
rs528399922 Chr.22: 37944639-37944639 on Build GRCh38
rs916598575 Chr.22: 37944864-37944864 on Build GRCh38
rs961455804 Chr.22: 37944718-37944718 on Build GRCh38
rs753520745 Chr.22: 37945016-37945016 on Build GRCh38
rs1040209534 Chr.22: 37944860-37944860 on Build GRCh38
rs976864364 Chr.22: 37944618-37944618 on Build GRCh38
rs184907810 Chr.22: 37944947-37944947 on Build GRCh38
rs34689374 Chr.22: 37944824-37944825 on Build GRCh38
rs750781074 Chr.22: 37945032-37945032 on Build GRCh38
rs766737991 Chr.22: 37945095-37945095 on Build GRCh38
rs756490588 Chr.22: 37945090-37945090 on Build GRCh38
rs750897255 Chr.22: 37944668-37944668 on Build GRCh38
rs978705059 Chr.22: 37944842-37944842 on Build GRCh38
rs976433511 Chr.22: 37944725-37944725 on Build GRCh38
rs1005378451 Chr.22: 37944942-37944942 on Build GRCh38
rs762481079 Chr.22: 37945086-37945086 on Build GRCh38
rs577420086 Chr.22: 37945084-37945084 on Build GRCh38
rs763162729 Chr.22: 37944717-37944717 on Build GRCh38
rs565460682 Chr.22: 37945060-37945060 on Build GRCh38
rs932932001 Chr.22: 37944623-37944623 on Build GRCh38
rs544488312 Chr.22: 37945096-37945096 on Build GRCh38
rs753374508 Chr.22: 37945137-37945138 on Build GRCh38
rs551124417 Chr.22: 37944658-37944658 on Build GRCh38
rs893628293 Chr.22: 37945145-37945145 on Build GRCh38
rs946771621 Chr.22: 37944633-37944633 on Build GRCh38
rs559246908 Chr.22: 37944628-37944628 on Build GRCh38
rs143633550 Chr.22: 37945108-37945108 on Build GRCh38
rs968189072 Chr.22: 37944772-37944772 on Build GRCh38
rs888288745 Chr.22: 37944701-37944701 on Build GRCh38
rs1037303398 Chr.22: 37944997-37944997 on Build GRCh38
rs199822431 Chr.22: 37945031-37945031 on Build GRCh38
rs983034278 Chr.22: 37945037-37945037 on Build GRCh38
rs758874854 Chr.22: 37944688-37944688 on Build GRCh38
rs765987764 Chr.22: 37945015-37945015 on Build GRCh38
rs371291489 Chr.22: 37945050-37945050 on Build GRCh38
rs375392356 Chr.22: 37945088-37945088 on Build GRCh38
rs902472777 Chr.22: 37944650-37944650 on Build GRCh38
rs35562630 Chr.22: 37945117-37945117 on Build GRCh38
rs538460585 Chr.22: 37944869-37944869 on Build GRCh38
rs113584643 Chr.22: 37944727-37944727 on Build GRCh38
rs569653281 Chr.22: 37944681-37944681 on Build GRCh38
rs139859 Chr.22: 37945127-37945127 on Build GRCh38
rs939363845 Chr.22: 37944655-37944655 on Build GRCh38
rs985687580 Chr.22: 37944767-37944767 on Build GRCh38
rs969063597 Chr.22: 37944988-37944988 on Build GRCh38
rs368393234 Chr.22: 37945101-37945104 on Build GRCh38
rs896883303 Chr.22: 37945004-37945004 on Build GRCh38
rs530569931 Chr.22: 37945064-37945065 on Build GRCh38
rs1022776795 Chr.22: 37944696-37944696 on Build GRCh38
rs759177237 Chr.22: 37945020-37945020 on Build GRCh38
rs746830275 Chr.22: 37945068-37945068 on Build GRCh38

View Full Product Details