Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 75281771-75282323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOS
Entrez Gene ID
2353
Gene Name
Fos proto-oncogene, AP-1 transcription factor subunit
Gene Aliases
AP-1, C-FOS, p55
UniGene
Hs.25647
Gene Chromosome Location
Chr.14: 75278778-75282234 on Build GRCh38
Transcripts
NM_005252.3

SNP Detail

SNP ID Chromosome Location
rs14371 Chr.14: 75282090-75282090 on Build GRCh38
rs551883379 Chr.14: 75281926-75281929 on Build GRCh38
rs45480193 Chr.14: 75281848-75281848 on Build GRCh38
rs1016286262 Chr.14: 75282190-75282190 on Build GRCh38
rs974909691 Chr.14: 75282235-75282235 on Build GRCh38
rs1011298990 Chr.14: 75281972-75281972 on Build GRCh38
rs554779201 Chr.14: 75281969-75281969 on Build GRCh38
rs950360985 Chr.14: 75282176-75282177 on Build GRCh38
rs779419408 Chr.14: 75282153-75282153 on Build GRCh38
rs3180688 Chr.14: 75282126-75282126 on Build GRCh38
rs754384786 Chr.14: 75281843-75281843 on Build GRCh38
rs972129041 Chr.14: 75282212-75282212 on Build GRCh38
rs986499909 Chr.14: 75282106-75282106 on Build GRCh38
rs1001600506 Chr.14: 75281906-75281906 on Build GRCh38
rs1046040071 Chr.14: 75281901-75281901 on Build GRCh38
rs967501573 Chr.14: 75281996-75281996 on Build GRCh38
rs1018631515 Chr.14: 75282229-75282229 on Build GRCh38
rs945416867 Chr.14: 75281807-75281807 on Build GRCh38
rs546553450 Chr.14: 75281838-75281838 on Build GRCh38
rs1004587215 Chr.14: 75282199-75282199 on Build GRCh38
rs963430060 Chr.14: 75282207-75282207 on Build GRCh38
rs1030993202 Chr.14: 75282082-75282082 on Build GRCh38
rs965749093 Chr.14: 75282234-75282234 on Build GRCh38
rs538767037 Chr.14: 75282030-75282030 on Build GRCh38
rs750879724 Chr.14: 75282250-75282250 on Build GRCh38
rs4645858 Chr.14: 75281905-75281905 on Build GRCh38
rs538000217 Chr.14: 75281924-75281924 on Build GRCh38
rs112926383 Chr.14: 75282231-75282231 on Build GRCh38
rs45628236 Chr.14: 75281840-75281840 on Build GRCh38
rs558838917 Chr.14: 75281861-75281861 on Build GRCh38
rs1055471768 Chr.14: 75281911-75281911 on Build GRCh38
rs550302148 Chr.14: 75282068-75282071 on Build GRCh38
rs892847222 Chr.14: 75281971-75281971 on Build GRCh38
rs957454529 Chr.14: 75282299-75282299 on Build GRCh38
rs1028345650 Chr.14: 75282115-75282115 on Build GRCh38
rs560765566 Chr.14: 75282089-75282089 on Build GRCh38
rs909257232 Chr.14: 75281810-75281810 on Build GRCh38
rs921772620 Chr.14: 75282254-75282254 on Build GRCh38
rs1020021896 Chr.14: 75281973-75281973 on Build GRCh38
rs919362214 Chr.14: 75282221-75282221 on Build GRCh38

View Full Product Details