Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 96285163-96285757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG2B
Entrez Gene ID
55102
Gene Name
autophagy related 2B
Gene Aliases
C14orf103
UniGene
Hs.168241
Gene Chromosome Location
Chr.14: 96281258-96363341 on Build GRCh38
Transcripts
XM_006720187.2

SNP Detail

SNP ID Chromosome Location
rs953249060 Chr.14: 96285479-96285479 on Build GRCh38
rs549268233 Chr.14: 96285460-96285460 on Build GRCh38
rs759663369 Chr.14: 96285596-96285596 on Build GRCh38
rs966216694 Chr.14: 96285530-96285530 on Build GRCh38
rs756127809 Chr.14: 96285280-96285280 on Build GRCh38
rs777268430 Chr.14: 96285721-96285721 on Build GRCh38
rs895906800 Chr.14: 96285371-96285371 on Build GRCh38
rs1040331389 Chr.14: 96285231-96285231 on Build GRCh38
rs376424066 Chr.14: 96285410-96285410 on Build GRCh38
rs1027582997 Chr.14: 96285465-96285465 on Build GRCh38
rs111415888 Chr.14: 96285319-96285319 on Build GRCh38
rs758041780 Chr.14: 96285716-96285716 on Build GRCh38
rs951450401 Chr.14: 96285610-96285610 on Build GRCh38
rs900315914 Chr.14: 96285271-96285271 on Build GRCh38
rs912552338 Chr.14: 96285702-96285702 on Build GRCh38
rs913597829 Chr.14: 96285674-96285674 on Build GRCh38
rs551139387 Chr.14: 96285421-96285421 on Build GRCh38
rs747506119 Chr.14: 96285707-96285708 on Build GRCh38
rs999105278 Chr.14: 96285411-96285412 on Build GRCh38
rs1018773697 Chr.14: 96285501-96285501 on Build GRCh38
rs150778607 Chr.14: 96285261-96285261 on Build GRCh38
rs987901104 Chr.14: 96285654-96285654 on Build GRCh38
rs969946910 Chr.14: 96285529-96285529 on Build GRCh38
rs1014319431 Chr.14: 96285386-96285386 on Build GRCh38
rs1014250512 Chr.14: 96285477-96285477 on Build GRCh38
rs979031570 Chr.14: 96285713-96285713 on Build GRCh38
rs1043287887 Chr.14: 96285283-96285283 on Build GRCh38
rs568317814 Chr.14: 96285599-96285599 on Build GRCh38
rs750350330 Chr.14: 96285194-96285194 on Build GRCh38
rs919969711 Chr.14: 96285562-96285562 on Build GRCh38
rs547515428 Chr.14: 96285678-96285678 on Build GRCh38
rs1015365244 Chr.14: 96285411-96285411 on Build GRCh38
rs746443554 Chr.14: 96285732-96285732 on Build GRCh38
rs988236396 Chr.14: 96285225-96285225 on Build GRCh38
rs937518284 Chr.14: 96285327-96285327 on Build GRCh38
rs1007419159 Chr.14: 96285482-96285482 on Build GRCh38
rs367842673 Chr.14: 96285554-96285555 on Build GRCh38
rs907760348 Chr.14: 96285223-96285223 on Build GRCh38
rs779939503 Chr.14: 96285424-96285424 on Build GRCh38
rs563164460 Chr.14: 96285449-96285449 on Build GRCh38
rs913555554 Chr.14: 96285253-96285253 on Build GRCh38
rs77540378 Chr.14: 96285457-96285457 on Build GRCh38
rs944450575 Chr.14: 96285222-96285222 on Build GRCh38
rs149387332 Chr.14: 96285653-96285653 on Build GRCh38
rs1055859482 Chr.14: 96285345-96285345 on Build GRCh38

View Full Product Details