Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 6779747-6780333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO39
Entrez Gene ID
162517
Gene Name
F-box protein 39
Gene Aliases
CT144, Fbx39
UniGene
Hs.368364
Gene Chromosome Location
Chr.17: 6776184-6798929 on Build GRCh38
Transcripts
NM_153230.2 XM_011523697.1 XM_017024283.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5039679 c.226G>A p.E76K Chr.17: 6780094-6780094 on Build GRCh38
2975080 c.369C>G p.N123K Chr.17: 6780237-6780237 on Build GRCh38
6556077 c.413G>T p.R138L Chr.17: 6780281-6780281 on Build GRCh38
2975082 c.403G>T p.E135* Chr.17: 6780271-6780271 on Build GRCh38
5443043 c.222A>C p.E74D Chr.17: 6780090-6780090 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759399371 Chr.17: 6780089-6780089 on Build GRCh38
rs780290851 Chr.17: 6780143-6780143 on Build GRCh38
rs747313581 Chr.17: 6780232-6780232 on Build GRCh38
rs774414477 Chr.17: 6779822-6779822 on Build GRCh38
rs991454223 Chr.17: 6780160-6780160 on Build GRCh38
rs746828339 Chr.17: 6780225-6780225 on Build GRCh38
rs747260135 Chr.17: 6780151-6780151 on Build GRCh38
rs764332418 Chr.17: 6779848-6779848 on Build GRCh38
rs926560791 Chr.17: 6780144-6780144 on Build GRCh38
rs542378940 Chr.17: 6780019-6780019 on Build GRCh38
rs1006244933 Chr.17: 6779919-6779919 on Build GRCh38
rs749711767 Chr.17: 6779909-6779909 on Build GRCh38
rs780924976 Chr.17: 6779992-6779992 on Build GRCh38
rs774739920 Chr.17: 6779912-6779912 on Build GRCh38
rs751318642 Chr.17: 6779976-6779976 on Build GRCh38
rs758859170 Chr.17: 6780220-6780220 on Build GRCh38
rs779943192 Chr.17: 6780113-6780123 on Build GRCh38
rs747137501 Chr.17: 6780048-6780048 on Build GRCh38
rs748190539 Chr.17: 6780169-6780169 on Build GRCh38
rs753332309 Chr.17: 6780127-6780127 on Build GRCh38
rs373411596 Chr.17: 6780205-6780205 on Build GRCh38
rs759700825 Chr.17: 6779917-6779917 on Build GRCh38
rs769894791 Chr.17: 6780244-6780244 on Build GRCh38
rs776729788 Chr.17: 6780061-6780061 on Build GRCh38
rs772078290 Chr.17: 6780289-6780289 on Build GRCh38
rs763720404 Chr.17: 6780109-6780109 on Build GRCh38
rs972471972 Chr.17: 6780285-6780285 on Build GRCh38
rs78983367 Chr.17: 6779929-6779929 on Build GRCh38
rs754915722 Chr.17: 6779793-6779793 on Build GRCh38
rs973567392 Chr.17: 6780036-6780036 on Build GRCh38
rs139725230 Chr.17: 6780280-6780280 on Build GRCh38
rs143419082 Chr.17: 6779872-6779872 on Build GRCh38
rs61753286 Chr.17: 6779874-6779874 on Build GRCh38
rs767526568 Chr.17: 6779833-6779833 on Build GRCh38
rs771724408 Chr.17: 6780180-6780180 on Build GRCh38
rs536953500 Chr.17: 6780281-6780281 on Build GRCh38
rs749792809 Chr.17: 6780026-6780026 on Build GRCh38
rs150271375 Chr.17: 6780128-6780128 on Build GRCh38
rs201069945 Chr.17: 6779928-6779928 on Build GRCh38
rs1030033745 Chr.17: 6780209-6780209 on Build GRCh38
rs775377350 Chr.17: 6780203-6780203 on Build GRCh38
rs768645681 Chr.17: 6780287-6780288 on Build GRCh38
rs763737793 Chr.17: 6780206-6780206 on Build GRCh38
rs367925502 Chr.17: 6779820-6779820 on Build GRCh38
rs761755341 Chr.17: 6779859-6779859 on Build GRCh38
rs765096770 Chr.17: 6779863-6779863 on Build GRCh38
rs768687307 Chr.17: 6780060-6780060 on Build GRCh38
rs769937090 Chr.17: 6780076-6780076 on Build GRCh38
rs200344565 Chr.17: 6780242-6780242 on Build GRCh38
rs556849207 Chr.17: 6780136-6780136 on Build GRCh38
rs774331786 Chr.17: 6780189-6780189 on Build GRCh38
rs780415756 Chr.17: 6780222-6780222 on Build GRCh38
rs887378122 Chr.17: 6779916-6779916 on Build GRCh38
rs376189972 Chr.17: 6779817-6779817 on Build GRCh38
rs770428705 Chr.17: 6779899-6779899 on Build GRCh38
rs138977169 Chr.17: 6780218-6780218 on Build GRCh38
rs758744565 Chr.17: 6780142-6780142 on Build GRCh38
rs889703951 Chr.17: 6780241-6780241 on Build GRCh38
rs377353189 Chr.17: 6780194-6780194 on Build GRCh38
rs866162711 Chr.17: 6779812-6779812 on Build GRCh38
rs368045455 Chr.17: 6779875-6779875 on Build GRCh38
rs146724926 Chr.17: 6779885-6779885 on Build GRCh38
rs755585129 Chr.17: 6779851-6779851 on Build GRCh38
rs755195237 Chr.17: 6780238-6780238 on Build GRCh38
rs759704081 Chr.17: 6779829-6779829 on Build GRCh38
rs752366449 Chr.17: 6780103-6780103 on Build GRCh38
rs1055114273 Chr.17: 6780125-6780125 on Build GRCh38
rs368869386 Chr.17: 6779838-6779838 on Build GRCh38
rs549708601 Chr.17: 6780135-6780135 on Build GRCh38
rs4796555 Chr.17: 6779896-6779896 on Build GRCh38
rs778465960 Chr.17: 6780025-6780025 on Build GRCh38
rs766116765 Chr.17: 6779971-6779971 on Build GRCh38
rs751519823 Chr.17: 6779778-6779778 on Build GRCh38
rs781471298 Chr.17: 6780240-6780240 on Build GRCh38
rs754599697 Chr.17: 6779982-6779982 on Build GRCh38
rs906542279 Chr.17: 6779776-6779776 on Build GRCh38
rs772988820 Chr.17: 6780213-6780213 on Build GRCh38
rs200419578 Chr.17: 6780037-6780037 on Build GRCh38
rs756781541 Chr.17: 6780215-6780215 on Build GRCh38
rs778419218 Chr.17: 6779902-6779902 on Build GRCh38
rs758244915 Chr.17: 6780077-6780084 on Build GRCh38
rs373018115 Chr.17: 6780138-6780138 on Build GRCh38
rs780619106 Chr.17: 6779801-6779801 on Build GRCh38
rs140674090 Chr.17: 6780038-6780038 on Build GRCh38
rs200968555 Chr.17: 6780196-6780196 on Build GRCh38
rs753360062 Chr.17: 6780211-6780211 on Build GRCh38
rs770840323 Chr.17: 6780181-6780181 on Build GRCh38
rs376499600 Chr.17: 6780049-6780049 on Build GRCh38
rs150934591 Chr.17: 6779945-6779945 on Build GRCh38
rs564185783 Chr.17: 6780107-6780107 on Build GRCh38
rs569543359 Chr.17: 6780164-6780164 on Build GRCh38
rs1016536562 Chr.17: 6780267-6780267 on Build GRCh38
rs369991116 Chr.17: 6780047-6780047 on Build GRCh38
rs762819097 Chr.17: 6780062-6780062 on Build GRCh38
rs199871206 Chr.17: 6780091-6780091 on Build GRCh38
rs768813963 Chr.17: 6780162-6780162 on Build GRCh38
rs755664200 Chr.17: 6780012-6780012 on Build GRCh38
rs539676107 Chr.17: 6779807-6779807 on Build GRCh38
rs576606128 Chr.17: 6779993-6779993 on Build GRCh38
rs766279425 Chr.17: 6780068-6780068 on Build GRCh38
rs964089461 Chr.17: 6779978-6779978 on Build GRCh38
rs146370414 Chr.17: 6779941-6779941 on Build GRCh38
rs61753143 Chr.17: 6779890-6779890 on Build GRCh38
rs1010770497 Chr.17: 6779871-6779871 on Build GRCh38
rs772227770 Chr.17: 6779925-6779925 on Build GRCh38
rs774019882 Chr.17: 6780081-6780081 on Build GRCh38

View Full Product Details