Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 76502730-76503324 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCNT1
Entrez Gene ID
2650
Gene Name
glucosaminyl (N-acetyl) transferase 1, core 2
Gene Aliases
C2GNT, C2GNT-L, C2GNT1, G6NT, NACGT2, NAGCT2
UniGene
Hs.521568
Gene Chromosome Location
Chr.9: 76441666-76507416 on Build GRCh38
Transcripts
NM_001097633.1 NM_001097634.1 NM_001097635.1 NM_001097636.1 NM_001490.4 XM_017014620.1 XM_017014621.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1674565 c.658G>A p.D220N Chr.9: 76503039-76503039 on Build GRCh38
4903703 c.903G>A p.Q301Q Chr.9: 76503284-76503284 on Build GRCh38
6193910 c.608T>G p.L203R Chr.9: 76502989-76502989 on Build GRCh38
754211 c.749A>G p.H250R Chr.9: 76503130-76503130 on Build GRCh38
1625106 c.657G>A p.M219I Chr.9: 76503038-76503038 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759615039 Chr.9: 76502769-76502769 on Build GRCh38
rs150529677 Chr.9: 76502787-76502787 on Build GRCh38
rs780811950 Chr.9: 76502834-76502834 on Build GRCh38
rs751651523 Chr.9: 76503086-76503086 on Build GRCh38
rs1055690516 Chr.9: 76503298-76503298 on Build GRCh38
rs758387360 Chr.9: 76502806-76502806 on Build GRCh38
rs376287292 Chr.9: 76503118-76503118 on Build GRCh38
rs367942265 Chr.9: 76503188-76503188 on Build GRCh38
rs913007014 Chr.9: 76503187-76503187 on Build GRCh38
rs865794464 Chr.9: 76503254-76503254 on Build GRCh38
rs755785501 Chr.9: 76502931-76502931 on Build GRCh38
rs758040032 Chr.9: 76502812-76502812 on Build GRCh38
rs762732983 Chr.9: 76502901-76502901 on Build GRCh38
rs781163105 Chr.9: 76503249-76503249 on Build GRCh38
rs747081945 Chr.9: 76503080-76503080 on Build GRCh38
rs141640466 Chr.9: 76502779-76502779 on Build GRCh38
rs775469409 Chr.9: 76503129-76503129 on Build GRCh38
rs777941377 Chr.9: 76502933-76502933 on Build GRCh38
rs140433187 Chr.9: 76502796-76502796 on Build GRCh38
rs756302676 Chr.9: 76503256-76503256 on Build GRCh38
rs774511006 Chr.9: 76502767-76502767 on Build GRCh38
rs758903174 Chr.9: 76503196-76503196 on Build GRCh38
rs546314870 Chr.9: 76502948-76502948 on Build GRCh38
rs200042546 Chr.9: 76503100-76503100 on Build GRCh38
rs762795842 Chr.9: 76502970-76502970 on Build GRCh38
rs539128019 Chr.9: 76502930-76502930 on Build GRCh38
rs770683006 Chr.9: 76502974-76502974 on Build GRCh38
rs656106 Chr.9: 76503216-76503216 on Build GRCh38
rs758991404 Chr.9: 76503047-76503047 on Build GRCh38
rs754231347 Chr.9: 76502938-76502938 on Build GRCh38
rs745654689 Chr.9: 76503288-76503288 on Build GRCh38
rs779101936 Chr.9: 76503233-76503233 on Build GRCh38
rs774224861 Chr.9: 76502852-76502852 on Build GRCh38
rs71509884 Chr.9: 76502963-76502963 on Build GRCh38
rs781455120 Chr.9: 76503090-76503090 on Build GRCh38
rs754550197 Chr.9: 76502815-76502815 on Build GRCh38
rs772001066 Chr.9: 76503128-76503128 on Build GRCh38
rs778611976 Chr.9: 76503269-76503269 on Build GRCh38
rs41288759 Chr.9: 76503176-76503176 on Build GRCh38
rs1011502292 Chr.9: 76503272-76503272 on Build GRCh38
rs749708931 Chr.9: 76502846-76502846 on Build GRCh38
rs376503627 Chr.9: 76503165-76503165 on Build GRCh38
rs756433009 Chr.9: 76503057-76503057 on Build GRCh38
rs772333502 Chr.9: 76502855-76502855 on Build GRCh38
rs766964977 Chr.9: 76502992-76502992 on Build GRCh38
rs372561524 Chr.9: 76503164-76503164 on Build GRCh38
rs373599348 Chr.9: 76502814-76502814 on Build GRCh38
rs763794562 Chr.9: 76503005-76503005 on Build GRCh38
rs778230598 Chr.9: 76502844-76502844 on Build GRCh38
rs750290107 Chr.9: 76502798-76502798 on Build GRCh38
rs759498821 Chr.9: 76502988-76502988 on Build GRCh38
rs751511655 Chr.9: 76503133-76503146 on Build GRCh38
rs556048059 Chr.9: 76503195-76503195 on Build GRCh38
rs770875325 Chr.9: 76502760-76502760 on Build GRCh38
rs369641865 Chr.9: 76503255-76503255 on Build GRCh38
rs775826802 Chr.9: 76502856-76502856 on Build GRCh38
rs149948942 Chr.9: 76502955-76502955 on Build GRCh38
rs774315358 Chr.9: 76502977-76502977 on Build GRCh38
rs371200631 Chr.9: 76503022-76503022 on Build GRCh38
rs2282683 Chr.9: 76502835-76502835 on Build GRCh38
rs757625842 Chr.9: 76502941-76502941 on Build GRCh38
rs367773332 Chr.9: 76503010-76503010 on Build GRCh38
rs771478745 Chr.9: 76502849-76502849 on Build GRCh38
rs368265529 Chr.9: 76503153-76503153 on Build GRCh38
rs766336364 Chr.9: 76502920-76502920 on Build GRCh38
rs565929874 Chr.9: 76503037-76503037 on Build GRCh38
rs776785969 Chr.9: 76502969-76502969 on Build GRCh38
rs773075818 Chr.9: 76502889-76502889 on Build GRCh38
rs767852982 Chr.9: 76502777-76502777 on Build GRCh38
rs766979831 Chr.9: 76503208-76503208 on Build GRCh38
rs145116432 Chr.9: 76503036-76503036 on Build GRCh38
rs11546569 Chr.9: 76502854-76502854 on Build GRCh38
rs151060743 Chr.9: 76503222-76503222 on Build GRCh38
rs759026554 Chr.9: 76502923-76502923 on Build GRCh38
rs746423733 Chr.9: 76502945-76502945 on Build GRCh38
rs749840186 Chr.9: 76502753-76502753 on Build GRCh38
rs750824838 Chr.9: 76503194-76503194 on Build GRCh38
rs975587504 Chr.9: 76502761-76502761 on Build GRCh38
rs757290320 Chr.9: 76503198-76503199 on Build GRCh38
rs573750966 Chr.9: 76502986-76502986 on Build GRCh38
rs928724563 Chr.9: 76503112-76503112 on Build GRCh38
rs752216368 Chr.9: 76503003-76503003 on Build GRCh38
rs769007670 Chr.9: 76502967-76502967 on Build GRCh38
rs925569384 Chr.9: 76503235-76503235 on Build GRCh38
rs756437872 Chr.9: 76502842-76502842 on Build GRCh38
rs757616309 Chr.9: 76503020-76503020 on Build GRCh38
rs570491038 Chr.9: 76502922-76502922 on Build GRCh38
rs745746805 Chr.9: 76503124-76503124 on Build GRCh38
rs375124342 Chr.9: 76503215-76503215 on Build GRCh38
rs1043099556 Chr.9: 76502828-76502828 on Build GRCh38
rs774153143 Chr.9: 76503119-76503119 on Build GRCh38
rs139675992 Chr.9: 76502886-76502886 on Build GRCh38
rs761011045 Chr.9: 76502868-76502868 on Build GRCh38
rs148923569 Chr.9: 76503084-76503084 on Build GRCh38
rs763744548 Chr.9: 76502928-76502929 on Build GRCh38
rs555199442 Chr.9: 76503113-76503113 on Build GRCh38
rs779459124 Chr.9: 76502944-76502944 on Build GRCh38
rs780219492 Chr.9: 76503048-76503048 on Build GRCh38
rs760250807 Chr.9: 76503004-76503004 on Build GRCh38
rs764292850 Chr.9: 76502993-76502993 on Build GRCh38

View Full Product Details