Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 45163789-45164339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf36
Entrez Gene ID
79742
Gene Name
chromosome X open reading frame 36
Gene Aliases
4930578C19Rik, DIA1R, EPQL1862, PRO3743, bA435K1.1
UniGene
Hs.98321
Gene Chromosome Location
Chr.X: 45148373-45200901 on Build GRCh38
Transcripts
NM_024689.2 NM_176819.3 XM_005272670.1 XM_006724559.1

SNP Detail

SNP ID Chromosome Location
rs779422834 Chr.X: 45164023-45164023 on Build GRCh38
rs7061984 Chr.X: 45163907-45163907 on Build GRCh38
rs748573448 Chr.X: 45164281-45164281 on Build GRCh38
rs781747454 Chr.X: 45164273-45164273 on Build GRCh38
rs766549240 Chr.X: 45164283-45164283 on Build GRCh38
rs774077493 Chr.X: 45164174-45164174 on Build GRCh38
rs769717141 Chr.X: 45163868-45163868 on Build GRCh38
rs753555891 Chr.X: 45164225-45164225 on Build GRCh38
rs914565977 Chr.X: 45164224-45164224 on Build GRCh38
rs748151816 Chr.X: 45163848-45163848 on Build GRCh38
rs771077188 Chr.X: 45164301-45164301 on Build GRCh38
rs772516195 Chr.X: 45164214-45164214 on Build GRCh38
rs942805162 Chr.X: 45163910-45163910 on Build GRCh38
rs763123276 Chr.X: 45164148-45164148 on Build GRCh38
rs925767972 Chr.X: 45163909-45163909 on Build GRCh38
rs933952046 Chr.X: 45164122-45164122 on Build GRCh38
rs373234614 Chr.X: 45164165-45164165 on Build GRCh38
rs1001295085 Chr.X: 45164237-45164237 on Build GRCh38
rs774602478 Chr.X: 45164307-45164307 on Build GRCh38
rs1038569067 Chr.X: 45164169-45164169 on Build GRCh38
rs752560792 Chr.X: 45164206-45164206 on Build GRCh38
rs922614222 Chr.X: 45163971-45163971 on Build GRCh38
rs1040152056 Chr.X: 45163945-45163945 on Build GRCh38
rs773870194 Chr.X: 45164230-45164230 on Build GRCh38
rs768175030 Chr.X: 45164085-45164085 on Build GRCh38
rs759478354 Chr.X: 45164179-45164179 on Build GRCh38
rs749297774 Chr.X: 45164216-45164216 on Build GRCh38
rs777667020 Chr.X: 45164293-45164293 on Build GRCh38
rs774900272 Chr.X: 45164095-45164095 on Build GRCh38
rs763478570 Chr.X: 45164223-45164223 on Build GRCh38
rs368888119 Chr.X: 45164260-45164260 on Build GRCh38
rs375902907 Chr.X: 45164242-45164242 on Build GRCh38
rs73206205 Chr.X: 45164082-45164082 on Build GRCh38
rs749528341 Chr.X: 45164295-45164295 on Build GRCh38
rs181848759 Chr.X: 45163966-45163966 on Build GRCh38
rs776549001 Chr.X: 45164318-45164318 on Build GRCh38
rs965244298 Chr.X: 45163965-45163965 on Build GRCh38
rs976962522 Chr.X: 45164059-45164059 on Build GRCh38
rs755388583 Chr.X: 45164264-45164264 on Build GRCh38
rs767512971 Chr.X: 45164184-45164184 on Build GRCh38
rs762304577 Chr.X: 45164134-45164134 on Build GRCh38
rs781125046 Chr.X: 45163866-45163866 on Build GRCh38
rs988800714 Chr.X: 45164213-45164213 on Build GRCh38

View Full Product Details