Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112427233-112427771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALPK1
Entrez Gene ID
80216
Gene Name
alpha kinase 1
Gene Aliases
8430410J10Rik, LAK
UniGene
Hs.652825
Gene Chromosome Location
Chr.4: 112286420-112442618 on Build GRCh38
Transcripts
NM_001102406.1 NM_001253884.1 NM_025144.3 XM_005263245.4 XM_005263246.4 XM_006714326.3 XM_017008633.1 XM_017008634.1 XM_017008635.1 XM_017008636.1 XM_017008637.1 XM_017008638.1 XM_017008639.1 XM_017008640.1 XM_017008641.1 XM_017008642.1 XM_017008643.1 XM_017008644.1 XM_017008645.1 XM_017008646.1 XM_017008647.1 XM_017008648.1 XM_017008649.1 XM_017008650.1 XM_017008651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4557203 c.718G>A p.G240S Chr.4: 112427588-112427588 on Build GRCh38
731960 c.747G>C p.M249I Chr.4: 112427617-112427617 on Build GRCh38
5974961 c.762T>C p.Y254Y Chr.4: 112427632-112427632 on Build GRCh38
5526534 c.705C>T p.L235L Chr.4: 112427575-112427575 on Build GRCh38
6488330 c.725T>G p.L242R Chr.4: 112427595-112427595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1015446756 Chr.4: 112427433-112427433 on Build GRCh38
rs200440410 Chr.4: 112427355-112427355 on Build GRCh38
rs752240064 Chr.4: 112427711-112427711 on Build GRCh38
rs763837838 Chr.4: 112427717-112427717 on Build GRCh38
rs199756961 Chr.4: 112427351-112427351 on Build GRCh38
rs879199633 Chr.4: 112427737-112427737 on Build GRCh38
rs75100887 Chr.4: 112427350-112427413 on Build GRCh38
rs747432717 Chr.4: 112427687-112427687 on Build GRCh38
rs199927672 Chr.4: 112427353-112427353 on Build GRCh38
rs986267825 Chr.4: 112427715-112427715 on Build GRCh38
rs764667727 Chr.4: 112427573-112427573 on Build GRCh38
rs113977807 Chr.4: 112427697-112427697 on Build GRCh38
rs777786049 Chr.4: 112427626-112427626 on Build GRCh38
rs772422578 Chr.4: 112427556-112427556 on Build GRCh38
rs1040821997 Chr.4: 112427602-112427602 on Build GRCh38
rs371765193 Chr.4: 112427709-112427709 on Build GRCh38
rs962338091 Chr.4: 112427313-112427313 on Build GRCh38
rs56997105 Chr.4: 112427351-112427354 on Build GRCh38
rs757233009 Chr.4: 112427634-112427634 on Build GRCh38
rs114465146 Chr.4: 112427497-112427497 on Build GRCh38
rs150823391 Chr.4: 112427619-112427619 on Build GRCh38
rs773865358 Chr.4: 112427703-112427703 on Build GRCh38
rs762548610 Chr.4: 112427583-112427583 on Build GRCh38
rs772436859 Chr.4: 112427660-112427660 on Build GRCh38
rs775509415 Chr.4: 112427684-112427684 on Build GRCh38
rs779204222 Chr.4: 112427649-112427649 on Build GRCh38
rs372563699 Chr.4: 112427738-112427739 on Build GRCh38
rs370712989 Chr.4: 112427543-112427543 on Build GRCh38
rs949111409 Chr.4: 112427406-112427434 on Build GRCh38
rs752537126 Chr.4: 112427616-112427616 on Build GRCh38
rs201382625 Chr.4: 112427410-112427410 on Build GRCh38
rs527857871 Chr.4: 112427690-112427690 on Build GRCh38
rs992355440 Chr.4: 112427728-112427728 on Build GRCh38
rs767856 Chr.4: 112427260-112427260 on Build GRCh38
rs749235785 Chr.4: 112427627-112427627 on Build GRCh38
rs952203327 Chr.4: 112427714-112427714 on Build GRCh38
rs952311990 Chr.4: 112427314-112427314 on Build GRCh38
rs917760604 Chr.4: 112427388-112427416 on Build GRCh38
rs201720771 Chr.4: 112427603-112427603 on Build GRCh38
rs1054397730 Chr.4: 112427555-112427555 on Build GRCh38
rs755986149 Chr.4: 112427349-112427352 on Build GRCh38
rs200480019 Chr.4: 112427402-112427402 on Build GRCh38
rs374448312 Chr.4: 112427349-112427368 on Build GRCh38
rs564997220 Chr.4: 112427415-112427415 on Build GRCh38
rs767094062 Chr.4: 112427610-112427610 on Build GRCh38
rs987971484 Chr.4: 112427371-112427385 on Build GRCh38
rs559793823 Chr.4: 112427506-112427506 on Build GRCh38
rs779504755 Chr.4: 112427638-112427638 on Build GRCh38
rs367588400 Chr.4: 112427351-112427412 on Build GRCh38
rs1004954749 Chr.4: 112427385-112427385 on Build GRCh38
rs200707092 Chr.4: 112427406-112427406 on Build GRCh38
rs767554894 Chr.4: 112427567-112427567 on Build GRCh38
rs1015533389 Chr.4: 112427363-112427363 on Build GRCh38
rs869115383 Chr.4: 112427352-112427413 on Build GRCh38
rs762292079 Chr.4: 112427694-112427694 on Build GRCh38
rs993022776 Chr.4: 112427496-112427496 on Build GRCh38
rs753520922 Chr.4: 112427735-112427735 on Build GRCh38
rs761672587 Chr.4: 112427507-112427507 on Build GRCh38
rs548784891 Chr.4: 112427488-112427488 on Build GRCh38
rs768855740 Chr.4: 112427688-112427688 on Build GRCh38
rs199701157 Chr.4: 112427412-112427412 on Build GRCh38
rs538982218 Chr.4: 112427358-112427415 on Build GRCh38
rs972175551 Chr.4: 112427328-112427328 on Build GRCh38
rs370003301 Chr.4: 112427413-112427414 on Build GRCh38
rs935926577 Chr.4: 112427571-112427571 on Build GRCh38
rs371617435 Chr.4: 112427372-112427373 on Build GRCh38
rs1052954321 Chr.4: 112427580-112427580 on Build GRCh38
rs200354459 Chr.4: 112427354-112427354 on Build GRCh38
rs201195172 Chr.4: 112427406-112427407 on Build GRCh38
rs200375768 Chr.4: 112427387-112427387 on Build GRCh38
rs372841187 Chr.4: 112427584-112427584 on Build GRCh38
rs996136705 Chr.4: 112427269-112427269 on Build GRCh38
rs760351476 Chr.4: 112427712-112427712 on Build GRCh38
rs928166960 Chr.4: 112427294-112427295 on Build GRCh38
rs373860176 Chr.4: 112427597-112427597 on Build GRCh38
rs930834615 Chr.4: 112427369-112427369 on Build GRCh38
rs201401104 Chr.4: 112427581-112427581 on Build GRCh38
rs146312094 Chr.4: 112427276-112427276 on Build GRCh38
rs377436691 Chr.4: 112427528-112427528 on Build GRCh38
rs199719196 Chr.4: 112427538-112427538 on Build GRCh38
rs745934168 Chr.4: 112427652-112427652 on Build GRCh38
rs1005588517 Chr.4: 112427655-112427655 on Build GRCh38
rs776059783 Chr.4: 112427557-112427557 on Build GRCh38
rs753035436 Chr.4: 112427334-112427335 on Build GRCh38
rs200049582 Chr.4: 112427352-112427352 on Build GRCh38
rs377361072 Chr.4: 112427539-112427539 on Build GRCh38
rs572343131 Chr.4: 112427270-112427270 on Build GRCh38
rs577329112 Chr.4: 112427486-112427486 on Build GRCh38
rs149307398 Chr.4: 112427737-112427738 on Build GRCh38
rs1022687023 Chr.4: 112427593-112427593 on Build GRCh38
rs200602800 Chr.4: 112427413-112427413 on Build GRCh38
rs888012804 Chr.4: 112427646-112427646 on Build GRCh38

View Full Product Details