Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 39119178-39119716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FYB
Entrez Gene ID
2533
Gene Name
FYN binding protein
Gene Aliases
ADAP, PRO0823, SLAP-130, SLAP130
UniGene
Hs.370503
Gene Chromosome Location
Chr.5: 39105252-39274523 on Build GRCh38
Transcripts
NM_001243093.1 NM_001465.4 NM_199335.3 XM_006714464.2 XM_006714465.2 XM_006714466.2 XM_011514008.2 XM_011514009.1 XM_011514010.1 XM_011514011.2 XM_017009316.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2689421 c.2139-7G>A p.? Chr.5: 39119641-39119641 on Build GRCh38
3849673 c.2190G>A p.K730K Chr.5: 39119583-39119583 on Build GRCh38
2689413 c.2226G>A p.R742R Chr.5: 39119547-39119547 on Build GRCh38
3849670 c.2190G>A p.K730K Chr.5: 39119583-39119583 on Build GRCh38
1437491 c.2224A>G p.R742G Chr.5: 39119549-39119549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916461780 Chr.5: 39119362-39119362 on Build GRCh38
rs893947067 Chr.5: 39119404-39119404 on Build GRCh38
rs764246818 Chr.5: 39119631-39119631 on Build GRCh38
rs772586087 Chr.5: 39119629-39119629 on Build GRCh38
rs267600623 Chr.5: 39119571-39119571 on Build GRCh38
rs909734857 Chr.5: 39119222-39119222 on Build GRCh38
rs909696006 Chr.5: 39119246-39119246 on Build GRCh38
rs959255180 Chr.5: 39119479-39119479 on Build GRCh38
rs1039282224 Chr.5: 39119368-39119368 on Build GRCh38
rs772894906 Chr.5: 39119554-39119554 on Build GRCh38
rs1016729948 Chr.5: 39119580-39119580 on Build GRCh38
rs373011379 Chr.5: 39119315-39119315 on Build GRCh38
rs757566808 Chr.5: 39119642-39119642 on Build GRCh38
rs1036874759 Chr.5: 39119294-39119294 on Build GRCh38
rs7723939 Chr.5: 39119686-39119686 on Build GRCh38
rs267600622 Chr.5: 39119570-39119570 on Build GRCh38
rs377225808 Chr.5: 39119441-39119441 on Build GRCh38
rs566079561 Chr.5: 39119221-39119221 on Build GRCh38
rs776449251 Chr.5: 39119603-39119603 on Build GRCh38
rs539916458 Chr.5: 39119321-39119321 on Build GRCh38
rs890677911 Chr.5: 39119339-39119339 on Build GRCh38
rs770697804 Chr.5: 39119561-39119561 on Build GRCh38
rs763252867 Chr.5: 39119598-39119598 on Build GRCh38
rs394930 Chr.5: 39119459-39119459 on Build GRCh38
rs1003981896 Chr.5: 39119455-39119455 on Build GRCh38
rs1039234746 Chr.5: 39119391-39119391 on Build GRCh38
rs1052205884 Chr.5: 39119326-39119326 on Build GRCh38
rs972979570 Chr.5: 39119575-39119575 on Build GRCh38
rs765156296 Chr.5: 39119618-39119618 on Build GRCh38
rs746219056 Chr.5: 39119505-39119505 on Build GRCh38
rs772645698 Chr.5: 39119496-39119497 on Build GRCh38
rs775842784 Chr.5: 39119666-39119669 on Build GRCh38
rs902757992 Chr.5: 39119450-39119450 on Build GRCh38
rs1007784028 Chr.5: 39119341-39119341 on Build GRCh38
rs918259223 Chr.5: 39119301-39119301 on Build GRCh38
rs762998354 Chr.5: 39119622-39119622 on Build GRCh38
rs751982479 Chr.5: 39119672-39119672 on Build GRCh38
rs201868597 Chr.5: 39119501-39119501 on Build GRCh38
rs959286209 Chr.5: 39119203-39119203 on Build GRCh38
rs902265050 Chr.5: 39119458-39119459 on Build GRCh38
rs750802327 Chr.5: 39119679-39119679 on Build GRCh38
rs574770122 Chr.5: 39119663-39119663 on Build GRCh38
rs1011167822 Chr.5: 39119408-39119408 on Build GRCh38
rs1020698405 Chr.5: 39119434-39119434 on Build GRCh38
rs990834933 Chr.5: 39119514-39119514 on Build GRCh38
rs751764845 Chr.5: 39119641-39119641 on Build GRCh38
rs1037778674 Chr.5: 39119309-39119309 on Build GRCh38
rs752245761 Chr.5: 39119566-39119566 on Build GRCh38
rs372533033 Chr.5: 39119620-39119620 on Build GRCh38
rs781561057 Chr.5: 39119655-39119655 on Build GRCh38
rs551537481 Chr.5: 39119417-39119417 on Build GRCh38
rs1035113619 Chr.5: 39119465-39119465 on Build GRCh38
rs569304618 Chr.5: 39119279-39119279 on Build GRCh38
rs1016889982 Chr.5: 39119527-39119527 on Build GRCh38
rs759531391 Chr.5: 39119616-39119616 on Build GRCh38
rs760422276 Chr.5: 39119557-39119557 on Build GRCh38
rs889560644 Chr.5: 39119345-39119345 on Build GRCh38
rs942405869 Chr.5: 39119381-39119381 on Build GRCh38
rs987466392 Chr.5: 39119629-39119629 on Build GRCh38
rs866013407 Chr.5: 39119582-39119582 on Build GRCh38
rs933689352 Chr.5: 39119311-39119311 on Build GRCh38
rs941198584 Chr.5: 39119253-39119253 on Build GRCh38
rs771887836 Chr.5: 39119547-39119547 on Build GRCh38
rs941294563 Chr.5: 39119261-39119261 on Build GRCh38
rs570634593 Chr.5: 39119454-39119454 on Build GRCh38
rs77239304 Chr.5: 39119369-39119369 on Build GRCh38
rs379707 Chr.5: 39119621-39119621 on Build GRCh38

View Full Product Details