Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 41933761-41934294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO4
Entrez Gene ID
26272
Gene Name
F-box protein 4
Gene Aliases
FBX4
UniGene
Hs.165575
Gene Chromosome Location
Chr.5: 41925249-42040230 on Build GRCh38
Transcripts
NM_001297437.1 NM_012176.2 NM_033484.2 XM_006714470.3 XM_011514026.2 XM_011514027.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5468265 c.834G>A p.Q278Q Chr.5: 41934244-41934244 on Build GRCh38
6662250 c.863G>T p.G288V Chr.5: 41934273-41934273 on Build GRCh38
53411 c.724A>G p.K242E Chr.5: 41934134-41934134 on Build GRCh38
5436495 c.838C>G p.Q280E Chr.5: 41934248-41934248 on Build GRCh38
6662249 c.863G>T p.G288V Chr.5: 41934273-41934273 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757886981 Chr.5: 41934120-41934121 on Build GRCh38
rs139568068 Chr.5: 41934192-41934192 on Build GRCh38
rs754980016 Chr.5: 41934045-41934045 on Build GRCh38
rs778704842 Chr.5: 41934158-41934158 on Build GRCh38
rs551162449 Chr.5: 41933842-41933842 on Build GRCh38
rs780017102 Chr.5: 41934035-41934035 on Build GRCh38
rs985827417 Chr.5: 41934121-41934121 on Build GRCh38
rs745717635 Chr.5: 41933917-41933917 on Build GRCh38
rs569029311 Chr.5: 41933864-41933864 on Build GRCh38
rs558849254 Chr.5: 41934018-41934018 on Build GRCh38
rs749064912 Chr.5: 41934038-41934038 on Build GRCh38
rs368369930 Chr.5: 41933803-41933803 on Build GRCh38
rs758307375 Chr.5: 41934169-41934169 on Build GRCh38
rs149448164 Chr.5: 41934120-41934120 on Build GRCh38
rs766966528 Chr.5: 41934137-41934137 on Build GRCh38
rs754320022 Chr.5: 41934108-41934111 on Build GRCh38
rs769722567 Chr.5: 41933923-41933923 on Build GRCh38
rs993769148 Chr.5: 41934022-41934022 on Build GRCh38
rs766094042 Chr.5: 41934152-41934152 on Build GRCh38
rs750103142 Chr.5: 41934030-41934030 on Build GRCh38
rs573042538 Chr.5: 41933943-41933943 on Build GRCh38
rs375021046 Chr.5: 41933801-41933801 on Build GRCh38
rs766764697 Chr.5: 41933998-41933998 on Build GRCh38
rs766992689 Chr.5: 41934026-41934026 on Build GRCh38
rs890636087 Chr.5: 41934056-41934056 on Build GRCh38
rs559438390 Chr.5: 41934183-41934183 on Build GRCh38
rs768661903 Chr.5: 41933969-41933969 on Build GRCh38
rs927875199 Chr.5: 41934167-41934167 on Build GRCh38
rs780423266 Chr.5: 41934050-41934051 on Build GRCh38
rs749269494 Chr.5: 41933962-41933962 on Build GRCh38
rs1052294653 Chr.5: 41933827-41933827 on Build GRCh38
rs997677302 Chr.5: 41934075-41934075 on Build GRCh38
rs755787042 Chr.5: 41934032-41934032 on Build GRCh38
rs536309737 Chr.5: 41933873-41933873 on Build GRCh38
rs528723765 Chr.5: 41934198-41934198 on Build GRCh38
rs577189527 Chr.5: 41934021-41934021 on Build GRCh38
rs915189994 Chr.5: 41933809-41933809 on Build GRCh38
rs748315150 Chr.5: 41934086-41934086 on Build GRCh38
rs748068857 Chr.5: 41934165-41934165 on Build GRCh38
rs763388630 Chr.5: 41934195-41934195 on Build GRCh38
rs141675593 Chr.5: 41934008-41934008 on Build GRCh38
rs866451226 Chr.5: 41934130-41934130 on Build GRCh38
rs146729073 Chr.5: 41934237-41934237 on Build GRCh38
rs747064227 Chr.5: 41934172-41934172 on Build GRCh38
rs774535471 Chr.5: 41933978-41933978 on Build GRCh38
rs753752449 Chr.5: 41934154-41934154 on Build GRCh38
rs772402597 Chr.5: 41933984-41933984 on Build GRCh38
rs773239522 Chr.5: 41934096-41934096 on Build GRCh38
rs137884688 Chr.5: 41934057-41934057 on Build GRCh38
rs752619607 Chr.5: 41933858-41933858 on Build GRCh38
rs571634489 Chr.5: 41934204-41934204 on Build GRCh38
rs989533257 Chr.5: 41933802-41933802 on Build GRCh38
rs749902930 Chr.5: 41934138-41934138 on Build GRCh38
rs768320778 Chr.5: 41934064-41934064 on Build GRCh38
rs149729841 Chr.5: 41934225-41934225 on Build GRCh38
rs201857049 Chr.5: 41934121-41934121 on Build GRCh38
rs532637639 Chr.5: 41934206-41934206 on Build GRCh38
rs538680318 Chr.5: 41933841-41933841 on Build GRCh38
rs201909279 Chr.5: 41934131-41934131 on Build GRCh38
rs374957047 Chr.5: 41934170-41934170 on Build GRCh38
rs948775010 Chr.5: 41933925-41933925 on Build GRCh38
rs780704969 Chr.5: 41933915-41933915 on Build GRCh38
rs559988406 Chr.5: 41934191-41934191 on Build GRCh38
rs747258299 Chr.5: 41934113-41934113 on Build GRCh38
rs758308528 Chr.5: 41934247-41934247 on Build GRCh38
rs367757201 Chr.5: 41934123-41934123 on Build GRCh38
rs771124157 Chr.5: 41934119-41934119 on Build GRCh38
rs571836740 Chr.5: 41933825-41933825 on Build GRCh38
rs964803854 Chr.5: 41934114-41934114 on Build GRCh38
rs541861883 Chr.5: 41934176-41934176 on Build GRCh38
rs751149174 Chr.5: 41934020-41934020 on Build GRCh38
rs773322764 Chr.5: 41933985-41933985 on Build GRCh38
rs761764703 Chr.5: 41933983-41933983 on Build GRCh38
rs768937992 Chr.5: 41934142-41934145 on Build GRCh38
rs765832234 Chr.5: 41934122-41934122 on Build GRCh38
rs771388488 Chr.5: 41934230-41934230 on Build GRCh38
rs192202249 Chr.5: 41934033-41934033 on Build GRCh38
rs756975452 Chr.5: 41933900-41933900 on Build GRCh38
rs112194697 Chr.5: 41933898-41933898 on Build GRCh38
rs201093238 Chr.5: 41934199-41934199 on Build GRCh38
rs754714358 Chr.5: 41934155-41934155 on Build GRCh38
rs748990341 Chr.5: 41934063-41934063 on Build GRCh38
rs772052410 Chr.5: 41934089-41934089 on Build GRCh38
rs751161590 Chr.5: 41933928-41933929 on Build GRCh38
rs755875450 Chr.5: 41934143-41934143 on Build GRCh38
rs762976512 Chr.5: 41933999-41933999 on Build GRCh38
rs753557222 Chr.5: 41934212-41934212 on Build GRCh38
rs1030627300 Chr.5: 41934084-41934084 on Build GRCh38
rs776824175 Chr.5: 41934178-41934178 on Build GRCh38
rs767010932 Chr.5: 41933973-41933974 on Build GRCh38
rs760823297 Chr.5: 41933989-41933989 on Build GRCh38
rs770055685 Chr.5: 41934187-41934187 on Build GRCh38
rs372005683 Chr.5: 41933872-41933872 on Build GRCh38
rs746430469 Chr.5: 41933950-41933952 on Build GRCh38

View Full Product Details