Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 178208794-178209352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPAB
Entrez Gene ID
3182
Gene Name
heterogeneous nuclear ribonucleoprotein A/B
Gene Aliases
ABBP1, HNRPAB
UniGene
Hs.591731
Gene Chromosome Location
Chr.5: 178204507-178211183 on Build GRCh38
Transcripts
NM_004499.3 NM_031266.2 NM_001278346.1 NM_153373.3 NR_103508.1 NR_103509.1 NR_103510.1 XM_006714925.2 XM_011534669.2 XM_011534670.2 XM_011534671.2 XM_011534672.2 XM_011534674.2 XM_011534676.2 XM_017009996.1 XM_017009997.1 XM_017009998.1 XM_017009999.1

Gene Symbol
PHYKPL
Entrez Gene ID
85007
Gene Name
5-phosphohydroxy-L-lysine phospho-lyase
Gene Aliases
AGXT2L2, PHLU
UniGene
Hs.248746
Gene Chromosome Location
Chr.5: 178208474-178232822 on Build GRCh38
Transcripts
NM_004499.3 NM_031266.2 NM_001278346.1 NM_153373.3 NR_103508.1 NR_103509.1 NR_103510.1 XM_006714925.2 XM_011534669.2 XM_011534670.2 XM_011534671.2 XM_011534672.2 XM_011534674.2 XM_011534676.2 XM_017009996.1 XM_017009997.1 XM_017009998.1 XM_017009999.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5910457 c.670-3T>C p.? Chr.5: 178209327-178209327 on Build GRCh38
5868266 c.671G>A p.C224Y Chr.5: 178209331-178209331 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs943276833 Chr.5: 178208953-178208953 on Build GRCh38
rs896760797 Chr.5: 178209093-178209093 on Build GRCh38
rs376126518 Chr.5: 178209226-178209226 on Build GRCh38
rs544382207 Chr.5: 178209269-178209269 on Build GRCh38
rs1009545480 Chr.5: 178208820-178208820 on Build GRCh38
rs976846611 Chr.5: 178209186-178209186 on Build GRCh38
rs1037319788 Chr.5: 178208948-178208948 on Build GRCh38
rs1016333975 Chr.5: 178209170-178209170 on Build GRCh38
rs992293951 Chr.5: 178208871-178208871 on Build GRCh38
rs892553913 Chr.5: 178208819-178208819 on Build GRCh38
rs1045835843 Chr.5: 178209032-178209032 on Build GRCh38
rs1020925520 Chr.5: 178209126-178209126 on Build GRCh38
rs563537995 Chr.5: 178208818-178208819 on Build GRCh38
rs577226985 Chr.5: 178209265-178209265 on Build GRCh38
rs976541472 Chr.5: 178208857-178208857 on Build GRCh38
rs773874091 Chr.5: 178208909-178208909 on Build GRCh38
rs909721684 Chr.5: 178209253-178209253 on Build GRCh38
rs35728946 Chr.5: 178209136-178209137 on Build GRCh38
rs950451477 Chr.5: 178209133-178209133 on Build GRCh38
rs908899922 Chr.5: 178208944-178208944 on Build GRCh38
rs985202246 Chr.5: 178209238-178209238 on Build GRCh38
rs564163483 Chr.5: 178209256-178209256 on Build GRCh38
rs774585068 Chr.5: 178208929-178208931 on Build GRCh38
rs776100195 Chr.5: 178209176-178209176 on Build GRCh38
rs892527438 Chr.5: 178209020-178209020 on Build GRCh38
rs941295156 Chr.5: 178208941-178208941 on Build GRCh38
rs537587266 Chr.5: 178209047-178209047 on Build GRCh38
rs923189834 Chr.5: 178208966-178208966 on Build GRCh38
rs918478475 Chr.5: 178209261-178209261 on Build GRCh38
rs1034261646 Chr.5: 178208837-178208837 on Build GRCh38
rs567038153 Chr.5: 178208983-178208983 on Build GRCh38
rs988631304 Chr.5: 178209200-178209200 on Build GRCh38
rs562872353 Chr.5: 178209275-178209275 on Build GRCh38
rs114379982 Chr.5: 178209040-178209040 on Build GRCh38
rs950483976 Chr.5: 178208876-178208876 on Build GRCh38
rs748859714 Chr.5: 178209306-178209306 on Build GRCh38
rs548919221 Chr.5: 178208832-178208832 on Build GRCh38
rs146320282 Chr.5: 178209295-178209295 on Build GRCh38
rs574100275 Chr.5: 178209114-178209114 on Build GRCh38
rs966744088 Chr.5: 178208856-178208856 on Build GRCh38
rs964373544 Chr.5: 178209171-178209171 on Build GRCh38
rs1033902829 Chr.5: 178209085-178209085 on Build GRCh38
rs904091422 Chr.5: 178208827-178208827 on Build GRCh38
rs772697723 Chr.5: 178208825-178208825 on Build GRCh38
rs778787282 Chr.5: 178209282-178209283 on Build GRCh38
rs756887503 Chr.5: 178209315-178209315 on Build GRCh38
rs142333197 Chr.5: 178209005-178209005 on Build GRCh38
rs1055060682 Chr.5: 178209264-178209264 on Build GRCh38
rs367966177 Chr.5: 178209293-178209293 on Build GRCh38
rs909706926 Chr.5: 178208940-178208940 on Build GRCh38
rs78891792 Chr.5: 178209081-178209081 on Build GRCh38
rs1052961425 Chr.5: 178208997-178208997 on Build GRCh38
rs778444767 Chr.5: 178209316-178209316 on Build GRCh38
rs960895595 Chr.5: 178208866-178208866 on Build GRCh38
rs974720162 Chr.5: 178208957-178208957 on Build GRCh38
rs558915156 Chr.5: 178209030-178209030 on Build GRCh38
rs897164698 Chr.5: 178208958-178208958 on Build GRCh38
rs371517909 Chr.5: 178208882-178208882 on Build GRCh38
rs777311444 Chr.5: 178209300-178209300 on Build GRCh38
rs370145181 Chr.5: 178209180-178209180 on Build GRCh38
rs903893711 Chr.5: 178209080-178209080 on Build GRCh38
rs984595571 Chr.5: 178209155-178209155 on Build GRCh38
rs1025996169 Chr.5: 178208875-178208875 on Build GRCh38
rs545973254 Chr.5: 178209255-178209255 on Build GRCh38
rs535338401 Chr.5: 178209143-178209143 on Build GRCh38
rs1043851495 Chr.5: 178209252-178209252 on Build GRCh38
rs530368385 Chr.5: 178208815-178208815 on Build GRCh38
rs1033175014 Chr.5: 178209080-178209080 on Build GRCh38
rs545315011 Chr.5: 178209281-178209281 on Build GRCh38
rs974814366 Chr.5: 178209173-178209173 on Build GRCh38
rs1059367 Chr.5: 178209198-178209198 on Build GRCh38
rs1026468948 Chr.5: 178209125-178209125 on Build GRCh38
rs748130778 Chr.5: 178209323-178209324 on Build GRCh38
rs747332559 Chr.5: 178209318-178209318 on Build GRCh38
rs771413364 Chr.5: 178208927-178208927 on Build GRCh38
rs945450749 Chr.5: 178209240-178209240 on Build GRCh38
rs768968337 Chr.5: 178209323-178209323 on Build GRCh38
rs1013678943 Chr.5: 178209107-178209107 on Build GRCh38
rs553632816 Chr.5: 178209169-178209169 on Build GRCh38
rs923162217 Chr.5: 178209194-178209194 on Build GRCh38
rs913095024 Chr.5: 178209207-178209207 on Build GRCh38

View Full Product Details