Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 97851999-97852546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASNS
Entrez Gene ID
440
Gene Name
asparagine synthetase (glutamine-hydrolyzing)
Gene Aliases
ASNSD, TS11
UniGene
Hs.489207
Gene Chromosome Location
Chr.7: 97852117-97872542 on Build GRCh38
Transcripts
NM_001178075.1 NM_001178076.1 NM_001178077.1 NM_001673.4 NM_133436.3 NM_183356.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4424052 c.1634C>T p.T545I Chr.7: 97852311-97852311 on Build GRCh38
5738029 c.1648C>T p.R550C Chr.7: 97852297-97852297 on Build GRCh38
3768585 c.1640C>A p.P547H Chr.7: 97852305-97852305 on Build GRCh38
4467947 c.1516C>T p.P506S Chr.7: 97852429-97852429 on Build GRCh38
2865883 c.1496C>T p.A499V Chr.7: 97852449-97852449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749741028 Chr.7: 97852241-97852241 on Build GRCh38
rs767700335 Chr.7: 97852331-97852331 on Build GRCh38
rs147448684 Chr.7: 97852463-97852463 on Build GRCh38
rs750880787 Chr.7: 97852481-97852481 on Build GRCh38
rs201432154 Chr.7: 97852352-97852352 on Build GRCh38
rs752951804 Chr.7: 97852335-97852335 on Build GRCh38
rs115270247 Chr.7: 97852179-97852179 on Build GRCh38
rs751859211 Chr.7: 97852216-97852216 on Build GRCh38
rs771660102 Chr.7: 97852256-97852256 on Build GRCh38
rs780904537 Chr.7: 97852344-97852345 on Build GRCh38
rs779360130 Chr.7: 97852360-97852360 on Build GRCh38
rs371278355 Chr.7: 97852219-97852219 on Build GRCh38
rs756447895 Chr.7: 97852351-97852351 on Build GRCh38
rs752871601 Chr.7: 97852429-97852429 on Build GRCh38
rs897877891 Chr.7: 97852307-97852307 on Build GRCh38
rs745559720 Chr.7: 97852255-97852255 on Build GRCh38
rs775200046 Chr.7: 97852259-97852259 on Build GRCh38
rs561841113 Chr.7: 97852191-97852191 on Build GRCh38
rs369978566 Chr.7: 97852233-97852233 on Build GRCh38
rs367894312 Chr.7: 97852212-97852212 on Build GRCh38
rs527374271 Chr.7: 97852200-97852200 on Build GRCh38
rs770968295 Chr.7: 97852285-97852304 on Build GRCh38
rs543190840 Chr.7: 97852459-97852459 on Build GRCh38
rs552452349 Chr.7: 97852296-97852296 on Build GRCh38
rs398122974 Chr.7: 97852297-97852297 on Build GRCh38
rs372844492 Chr.7: 97852436-97852436 on Build GRCh38
rs757846773 Chr.7: 97852489-97852489 on Build GRCh38
rs547403927 Chr.7: 97852229-97852229 on Build GRCh38
rs879056597 Chr.7: 97852269-97852269 on Build GRCh38
rs766586629 Chr.7: 97852300-97852300 on Build GRCh38
rs140975053 Chr.7: 97852358-97852358 on Build GRCh38
rs913613977 Chr.7: 97852163-97852163 on Build GRCh38
rs747005354 Chr.7: 97852284-97852285 on Build GRCh38
rs773279607 Chr.7: 97852415-97852415 on Build GRCh38
rs767577039 Chr.7: 97852426-97852426 on Build GRCh38
rs374715282 Chr.7: 97852357-97852357 on Build GRCh38
rs896304129 Chr.7: 97852239-97852239 on Build GRCh38
rs761677126 Chr.7: 97852410-97852410 on Build GRCh38
rs764468161 Chr.7: 97852439-97852439 on Build GRCh38
rs75198492 Chr.7: 97852154-97852154 on Build GRCh38
rs765308672 Chr.7: 97852414-97852414 on Build GRCh38
rs757715023 Chr.7: 97852359-97852359 on Build GRCh38
rs563917352 Chr.7: 97852287-97852287 on Build GRCh38
rs755055878 Chr.7: 97852321-97852321 on Build GRCh38
rs376247830 Chr.7: 97852419-97852419 on Build GRCh38
rs760440838 Chr.7: 97852271-97852271 on Build GRCh38
rs913657474 Chr.7: 97852484-97852484 on Build GRCh38
rs910711922 Chr.7: 97852081-97852081 on Build GRCh38
rs767444019 Chr.7: 97852090-97852090 on Build GRCh38
rs751735389 Chr.7: 97852317-97852317 on Build GRCh38
rs773814355 Chr.7: 97852248-97852248 on Build GRCh38
rs10486011 Chr.7: 97852500-97852500 on Build GRCh38
rs576307404 Chr.7: 97852159-97852159 on Build GRCh38
rs201209327 Chr.7: 97852367-97852367 on Build GRCh38
rs993460441 Chr.7: 97852353-97852353 on Build GRCh38
rs770435446 Chr.7: 97852247-97852247 on Build GRCh38
rs532710533 Chr.7: 97852293-97852293 on Build GRCh38
rs373078034 Chr.7: 97852375-97852375 on Build GRCh38
rs777391418 Chr.7: 97852260-97852261 on Build GRCh38
rs779451673 Chr.7: 97852471-97852471 on Build GRCh38
rs773526843 Chr.7: 97852093-97852093 on Build GRCh38
rs374219443 Chr.7: 97852220-97852220 on Build GRCh38
rs1043373356 Chr.7: 97852194-97852194 on Build GRCh38
rs376090391 Chr.7: 97852455-97852455 on Build GRCh38
rs768470556 Chr.7: 97852516-97852516 on Build GRCh38
rs546113283 Chr.7: 97852119-97852119 on Build GRCh38
rs746606098 Chr.7: 97852383-97852383 on Build GRCh38
rs1803163 Chr.7: 97852305-97852305 on Build GRCh38
rs568570377 Chr.7: 97852389-97852389 on Build GRCh38
rs757610802 Chr.7: 97852456-97852456 on Build GRCh38
rs746662528 Chr.7: 97852503-97852503 on Build GRCh38
rs771595135 Chr.7: 97852370-97852370 on Build GRCh38

View Full Product Details