Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94130504-94131103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH17
Entrez Gene ID
1015
Gene Name
cadherin 17
Gene Aliases
CDH16, HPT-1, HPT1
UniGene
Hs.591853
Gene Chromosome Location
Chr.8: 94127162-94217303 on Build GRCh38
Transcripts
NM_001144663.1 NM_004063.3 XM_011516790.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5424411 c.2216A>C p.E739A Chr.8: 94130944-94130944 on Build GRCh38
6252682 c.2363C>A p.A788E Chr.8: 94130661-94130661 on Build GRCh38
4420389 c.2275T>C p.S759P Chr.8: 94130885-94130885 on Build GRCh38
4635641 c.2234G>A p.R745H Chr.8: 94130926-94130926 on Build GRCh38
3763457 c.2202G>C p.E734D Chr.8: 94130958-94130958 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369741964 Chr.8: 94130825-94130825 on Build GRCh38
rs531694199 Chr.8: 94130790-94130790 on Build GRCh38
rs750641030 Chr.8: 94130873-94130873 on Build GRCh38
rs778961978 Chr.8: 94130742-94130742 on Build GRCh38
rs903345460 Chr.8: 94130562-94130563 on Build GRCh38
rs200480935 Chr.8: 94130662-94130662 on Build GRCh38
rs780779072 Chr.8: 94130763-94130763 on Build GRCh38
rs760902048 Chr.8: 94130729-94130729 on Build GRCh38
rs772002795 Chr.8: 94130684-94130684 on Build GRCh38
rs963705644 Chr.8: 94130792-94130792 on Build GRCh38
rs753239226 Chr.8: 94130726-94130726 on Build GRCh38
rs757457828 Chr.8: 94131041-94131042 on Build GRCh38
rs1041792402 Chr.8: 94130874-94130874 on Build GRCh38
rs199718614 Chr.8: 94130949-94130949 on Build GRCh38
rs937759442 Chr.8: 94130643-94130643 on Build GRCh38
rs150270154 Chr.8: 94130927-94130927 on Build GRCh38
rs776824647 Chr.8: 94130558-94130558 on Build GRCh38
rs192589622 Chr.8: 94130603-94130603 on Build GRCh38
rs971020266 Chr.8: 94131040-94131040 on Build GRCh38
rs760490043 Chr.8: 94130626-94130627 on Build GRCh38
rs146691258 Chr.8: 94130660-94130660 on Build GRCh38
rs761508463 Chr.8: 94130692-94130692 on Build GRCh38
rs377593854 Chr.8: 94130909-94130909 on Build GRCh38
rs771159015 Chr.8: 94130990-94130990 on Build GRCh38
rs570796332 Chr.8: 94130895-94130895 on Build GRCh38
rs926341114 Chr.8: 94130628-94130628 on Build GRCh38
rs1003466932 Chr.8: 94131068-94131068 on Build GRCh38
rs773474977 Chr.8: 94130924-94130924 on Build GRCh38
rs758232004 Chr.8: 94130875-94130875 on Build GRCh38
rs113977006 Chr.8: 94131005-94131006 on Build GRCh38
rs1033456983 Chr.8: 94131032-94131032 on Build GRCh38
rs746288985 Chr.8: 94130787-94130787 on Build GRCh38
rs992082797 Chr.8: 94131058-94131058 on Build GRCh38
rs982328099 Chr.8: 94130869-94130869 on Build GRCh38
rs762957284 Chr.8: 94130985-94130985 on Build GRCh38
rs145270119 Chr.8: 94130877-94130877 on Build GRCh38
rs571522164 Chr.8: 94130826-94130826 on Build GRCh38
rs534110861 Chr.8: 94130908-94130908 on Build GRCh38
rs767544550 Chr.8: 94130937-94130937 on Build GRCh38
rs138634549 Chr.8: 94130586-94130586 on Build GRCh38
rs199497492 Chr.8: 94130704-94130704 on Build GRCh38
rs776554687 Chr.8: 94130845-94130846 on Build GRCh38
rs769210721 Chr.8: 94130915-94130915 on Build GRCh38
rs1051624 Chr.8: 94130944-94130944 on Build GRCh38
rs16916585 Chr.8: 94130754-94130754 on Build GRCh38
rs544278571 Chr.8: 94130646-94130646 on Build GRCh38
rs750428578 Chr.8: 94131057-94131057 on Build GRCh38
rs766738946 Chr.8: 94130604-94130604 on Build GRCh38
rs993885654 Chr.8: 94131015-94131015 on Build GRCh38
rs770145671 Chr.8: 94130655-94130655 on Build GRCh38
rs776990944 Chr.8: 94130916-94130916 on Build GRCh38
rs947627544 Chr.8: 94130671-94130671 on Build GRCh38
rs972393855 Chr.8: 94130554-94130554 on Build GRCh38
rs899274618 Chr.8: 94131011-94131011 on Build GRCh38
rs769495156 Chr.8: 94130774-94130774 on Build GRCh38
rs139170804 Chr.8: 94130936-94130936 on Build GRCh38
rs141103178 Chr.8: 94130920-94130920 on Build GRCh38
rs767800258 Chr.8: 94130912-94130912 on Build GRCh38
rs754978008 Chr.8: 94130882-94130882 on Build GRCh38
rs765553891 Chr.8: 94130864-94130864 on Build GRCh38
rs759438649 Chr.8: 94131033-94131033 on Build GRCh38
rs747574291 Chr.8: 94130764-94130764 on Build GRCh38
rs774539012 Chr.8: 94131000-94131000 on Build GRCh38
rs774813679 Chr.8: 94130703-94130703 on Build GRCh38
rs571282275 Chr.8: 94130647-94130647 on Build GRCh38
rs773777280 Chr.8: 94130717-94130717 on Build GRCh38
rs972123498 Chr.8: 94130811-94130811 on Build GRCh38
rs950147046 Chr.8: 94130528-94130529 on Build GRCh38
rs751781106 Chr.8: 94131006-94131007 on Build GRCh38
rs762019852 Chr.8: 94130919-94130919 on Build GRCh38
rs779910161 Chr.8: 94130686-94130686 on Build GRCh38
rs776017133 Chr.8: 94130911-94130911 on Build GRCh38
rs760145848 Chr.8: 94130619-94130619 on Build GRCh38
rs776803179 Chr.8: 94130852-94130852 on Build GRCh38
rs900097047 Chr.8: 94130613-94130613 on Build GRCh38
rs768898733 Chr.8: 94130967-94130967 on Build GRCh38
rs770279346 Chr.8: 94130743-94130743 on Build GRCh38
rs780929487 Chr.8: 94130893-94130893 on Build GRCh38
rs117852687 Chr.8: 94130571-94130571 on Build GRCh38
rs772678822 Chr.8: 94130776-94130776 on Build GRCh38
rs746919625 Chr.8: 94130753-94130753 on Build GRCh38
rs769285229 Chr.8: 94130694-94130694 on Build GRCh38
rs16916583 Chr.8: 94130735-94130735 on Build GRCh38
rs554411229 Chr.8: 94130583-94130583 on Build GRCh38
rs2513797 Chr.8: 94130910-94130910 on Build GRCh38
rs751309904 Chr.8: 94130880-94130880 on Build GRCh38
rs555213077 Chr.8: 94130600-94130600 on Build GRCh38
rs748583626 Chr.8: 94130977-94130977 on Build GRCh38
rs773313879 Chr.8: 94130983-94130983 on Build GRCh38
rs997534611 Chr.8: 94130634-94130634 on Build GRCh38
rs374089949 Chr.8: 94130857-94130857 on Build GRCh38
rs752824274 Chr.8: 94130635-94130635 on Build GRCh38
rs774694145 Chr.8: 94130894-94130894 on Build GRCh38
rs187981885 Chr.8: 94130943-94130943 on Build GRCh38
rs757522033 Chr.8: 94130741-94130741 on Build GRCh38
rs757040010 Chr.8: 94130659-94130659 on Build GRCh38
rs867186116 Chr.8: 94130698-94130698 on Build GRCh38
rs1054687782 Chr.8: 94130612-94130612 on Build GRCh38
rs755469213 Chr.8: 94130730-94130730 on Build GRCh38
rs557873384 Chr.8: 94131036-94131036 on Build GRCh38
rs781743258 Chr.8: 94130971-94130971 on Build GRCh38
rs750231595 Chr.8: 94130743-94130743 on Build GRCh38
rs763018446 Chr.8: 94131006-94131006 on Build GRCh38
rs3192029 Chr.8: 94130638-94130638 on Build GRCh38
rs764457724 Chr.8: 94130842-94130842 on Build GRCh38
rs755562158 Chr.8: 94130899-94130899 on Build GRCh38
rs368869228 Chr.8: 94130723-94130723 on Build GRCh38
rs541787965 Chr.8: 94130597-94130597 on Build GRCh38
rs150741866 Chr.8: 94130786-94130786 on Build GRCh38
rs1051623 Chr.8: 94130958-94130958 on Build GRCh38
rs970691362 Chr.8: 94130851-94130851 on Build GRCh38
rs752058209 Chr.8: 94130610-94130610 on Build GRCh38
rs959074829 Chr.8: 94130889-94130889 on Build GRCh38
rs768102379 Chr.8: 94130621-94130621 on Build GRCh38
rs184072714 Chr.8: 94130752-94130752 on Build GRCh38
rs758176881 Chr.8: 94130745-94130745 on Build GRCh38
rs765077127 Chr.8: 94130980-94130980 on Build GRCh38
rs766247124 Chr.8: 94130633-94130634 on Build GRCh38
rs141222335 Chr.8: 94130690-94130690 on Build GRCh38

View Full Product Details