Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 114582625-114583224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NXPE2
Entrez Gene ID
120406
Gene Name
neurexophilin and PC-esterase domain family member 2
Gene Aliases
FAM55B
UniGene
Hs.446760
Gene Chromosome Location
Chr.11: 114464276-114813702 on Build GRCh38
Transcripts
XM_017017207.1 XM_017017208.1 XM_017017209.1 XM_017017210.1 NM_001077639.1 NM_017678.2 XM_006718865.3 XM_011542881.2 XM_011542882.2 XM_011542883.2

Gene Symbol
NXPE4
Entrez Gene ID
54827
Gene Name
neurexophilin and PC-esterase domain family member 4
Gene Aliases
C11orf33, FAM55D
UniGene
Hs.179100
Gene Chromosome Location
Chr.11: 114570591-114596215 on Build GRCh38
Transcripts
XM_017017207.1 XM_017017208.1 XM_017017209.1 XM_017017210.1 NM_001077639.1 NM_017678.2 XM_006718865.3 XM_011542881.2 XM_011542882.2 XM_011542883.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6189601 c.394C>T p.Q132* Chr.11: 114582724-114582724 on Build GRCh38
2103926 c.329C>T p.T110M Chr.11: 114582789-114582789 on Build GRCh38
1251919 c.449G>A p.G150D Chr.11: 114582669-114582669 on Build GRCh38
5453080 c.419C>T p.A140V Chr.11: 114582699-114582699 on Build GRCh38
3808403 c.307A>C p.T103P Chr.11: 114582811-114582811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774157031 Chr.11: 114582732-114582732 on Build GRCh38
rs369071049 Chr.11: 114582719-114582719 on Build GRCh38
rs754295764 Chr.11: 114582685-114582685 on Build GRCh38
rs1004302011 Chr.11: 114582660-114582660 on Build GRCh38
rs775188738 Chr.11: 114582750-114582750 on Build GRCh38
rs200385279 Chr.11: 114582711-114582711 on Build GRCh38
rs200121976 Chr.11: 114582764-114582764 on Build GRCh38
rs781187185 Chr.11: 114582949-114582950 on Build GRCh38
rs989874874 Chr.11: 114583177-114583177 on Build GRCh38
rs766889659 Chr.11: 114582911-114582911 on Build GRCh38
rs776429191 Chr.11: 114582860-114582860 on Build GRCh38
rs763316164 Chr.11: 114583021-114583021 on Build GRCh38
rs766921692 Chr.11: 114582749-114582749 on Build GRCh38
rs761351977 Chr.11: 114583015-114583015 on Build GRCh38
rs772086288 Chr.11: 114582981-114582981 on Build GRCh38
rs749870159 Chr.11: 114582654-114582654 on Build GRCh38
rs566166344 Chr.11: 114582977-114582977 on Build GRCh38
rs781404137 Chr.11: 114582805-114582805 on Build GRCh38
rs977586598 Chr.11: 114583196-114583196 on Build GRCh38
rs140208011 Chr.11: 114582887-114582887 on Build GRCh38
rs148043908 Chr.11: 114583098-114583098 on Build GRCh38
rs201549180 Chr.11: 114582789-114582789 on Build GRCh38
rs779311957 Chr.11: 114582704-114582704 on Build GRCh38
rs115348655 Chr.11: 114582696-114582696 on Build GRCh38
rs183638687 Chr.11: 114582937-114582937 on Build GRCh38
rs1006340721 Chr.11: 114583023-114583023 on Build GRCh38
rs192956484 Chr.11: 114582667-114582667 on Build GRCh38
rs192697424 Chr.11: 114583011-114583011 on Build GRCh38
rs758758998 Chr.11: 114582884-114582884 on Build GRCh38
rs377228260 Chr.11: 114582788-114582788 on Build GRCh38
rs201831822 Chr.11: 114582795-114582795 on Build GRCh38
rs780177321 Chr.11: 114582713-114582713 on Build GRCh38
rs370550628 Chr.11: 114582904-114582904 on Build GRCh38
rs752861867 Chr.11: 114583062-114583062 on Build GRCh38
rs1028019069 Chr.11: 114582878-114582878 on Build GRCh38
rs772742540 Chr.11: 114582924-114582924 on Build GRCh38
rs540593311 Chr.11: 114583191-114583191 on Build GRCh38
rs766253958 Chr.11: 114582666-114582666 on Build GRCh38
rs752055029 Chr.11: 114583039-114583039 on Build GRCh38
rs200548586 Chr.11: 114582681-114582681 on Build GRCh38
rs898305959 Chr.11: 114583051-114583051 on Build GRCh38
rs376142309 Chr.11: 114582830-114582830 on Build GRCh38
rs972637851 Chr.11: 114582829-114582829 on Build GRCh38
rs113839069 Chr.11: 114583138-114583138 on Build GRCh38
rs955299738 Chr.11: 114583114-114583114 on Build GRCh38
rs986494678 Chr.11: 114582842-114582842 on Build GRCh38
rs201081268 Chr.11: 114582986-114582986 on Build GRCh38
rs765709768 Chr.11: 114582779-114582779 on Build GRCh38
rs555017873 Chr.11: 114582994-114582994 on Build GRCh38
rs191556206 Chr.11: 114582834-114582834 on Build GRCh38
rs755139961 Chr.11: 114582803-114582803 on Build GRCh38
rs761596470 Chr.11: 114582880-114582880 on Build GRCh38
rs371127636 Chr.11: 114582848-114582848 on Build GRCh38
rs775100729 Chr.11: 114582847-114582847 on Build GRCh38
rs749055897 Chr.11: 114582721-114582721 on Build GRCh38
rs568501282 Chr.11: 114583001-114583001 on Build GRCh38
rs1007897727 Chr.11: 114583130-114583130 on Build GRCh38
rs377657848 Chr.11: 114582796-114582796 on Build GRCh38
rs565034018 Chr.11: 114582778-114582778 on Build GRCh38
rs374186429 Chr.11: 114583004-114583004 on Build GRCh38
rs758415512 Chr.11: 114582712-114582712 on Build GRCh38
rs761532668 Chr.11: 114582655-114582655 on Build GRCh38
rs187061568 Chr.11: 114582833-114582833 on Build GRCh38
rs371147789 Chr.11: 114582827-114582827 on Build GRCh38
rs760091707 Chr.11: 114583047-114583047 on Build GRCh38
rs766801718 Chr.11: 114583034-114583034 on Build GRCh38
rs1034366792 Chr.11: 114582742-114582742 on Build GRCh38
rs769575246 Chr.11: 114582656-114582656 on Build GRCh38
rs142540044 Chr.11: 114583160-114583160 on Build GRCh38
rs752391000 Chr.11: 114582680-114582680 on Build GRCh38
rs766029574 Chr.11: 114582770-114582771 on Build GRCh38
rs754207728 Chr.11: 114582772-114582772 on Build GRCh38
rs185749351 Chr.11: 114583020-114583020 on Build GRCh38
rs190553607 Chr.11: 114583133-114583133 on Build GRCh38
rs775589425 Chr.11: 114582651-114582651 on Build GRCh38
rs762842355 Chr.11: 114582662-114582662 on Build GRCh38
rs984837037 Chr.11: 114583082-114583082 on Build GRCh38
rs184273600 Chr.11: 114582715-114582715 on Build GRCh38
rs868456164 Chr.11: 114582913-114582913 on Build GRCh38
rs778259672 Chr.11: 114582684-114582684 on Build GRCh38
rs1025554955 Chr.11: 114583102-114583102 on Build GRCh38
rs752554204 Chr.11: 114582980-114582980 on Build GRCh38
rs900866642 Chr.11: 114583161-114583161 on Build GRCh38
rs758803514 Chr.11: 114582905-114582905 on Build GRCh38
rs751576523 Chr.11: 114582915-114582915 on Build GRCh38
rs772290293 Chr.11: 114582649-114582649 on Build GRCh38
rs112073332 Chr.11: 114582948-114582948 on Build GRCh38
rs754812838 Chr.11: 114582894-114582894 on Build GRCh38
rs893378957 Chr.11: 114582697-114582697 on Build GRCh38
rs763783603 Chr.11: 114582854-114582854 on Build GRCh38
rs111330181 Chr.11: 114582733-114582733 on Build GRCh38
rs200964488 Chr.11: 114582957-114582957 on Build GRCh38
rs181712337 Chr.11: 114583188-114583188 on Build GRCh38
rs773053522 Chr.11: 114582658-114582658 on Build GRCh38
rs866741765 Chr.11: 114582898-114582898 on Build GRCh38
rs763871832 Chr.11: 114582766-114582766 on Build GRCh38
rs767561811 Chr.11: 114583049-114583049 on Build GRCh38
rs767129579 Chr.11: 114582669-114582669 on Build GRCh38
rs73557425 Chr.11: 114582716-114582716 on Build GRCh38

View Full Product Details