Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119100028-119100589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPAGT1
Entrez Gene ID
1798
Gene Name
dolichyl-phosphate N-acetylglucosaminephosphotransferase 1
Gene Aliases
ALG7, CDG-Ij, CDG1J, CMS13, CMSTA2, D11S366, DGPT, DPAGT, DPAGT2, G1PT, GPT, UAGT, UGAT
UniGene
Hs.524081
Gene Chromosome Location
Chr.11: 119096503-119102414 on Build GRCh38
Transcripts
NM_001382.3 XM_005271422.2 XM_011542648.1 XM_017017293.1 XM_017017294.1 XM_017017295.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3985976 c.575G>A p.G192D Chr.11: 119100330-119100330 on Build GRCh38
6648758 c.296T>C p.I99T Chr.11: 119100288-119100288 on Build GRCh38
5547158 c.280delA p.I94fs*11 Chr.11: 119100304-119100304 on Build GRCh38
3935340 c.497G>T p.G166V Chr.11: 119100408-119100408 on Build GRCh38
6648757 c.617T>C p.I206T Chr.11: 119100288-119100288 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs946141210 Chr.11: 119100374-119100374 on Build GRCh38
rs915756308 Chr.11: 119100531-119100531 on Build GRCh38
rs968342032 Chr.11: 119100062-119100062 on Build GRCh38
rs75437642 Chr.11: 119100100-119100100 on Build GRCh38
rs765913027 Chr.11: 119100255-119100255 on Build GRCh38
rs755661117 Chr.11: 119100537-119100537 on Build GRCh38
rs536452844 Chr.11: 119100248-119100248 on Build GRCh38
rs80188413 Chr.11: 119100098-119100098 on Build GRCh38
rs370211356 Chr.11: 119100268-119100268 on Build GRCh38
rs1031264691 Chr.11: 119100448-119100448 on Build GRCh38
rs397515329 Chr.11: 119100402-119100402 on Build GRCh38
rs542884106 Chr.11: 119100476-119100476 on Build GRCh38
rs28934876 Chr.11: 119100396-119100396 on Build GRCh38
rs768464558 Chr.11: 119100331-119100331 on Build GRCh38
rs992739096 Chr.11: 119100489-119100489 on Build GRCh38
rs907065621 Chr.11: 119100443-119100443 on Build GRCh38
rs1025735507 Chr.11: 119100553-119100553 on Build GRCh38
rs764801785 Chr.11: 119100565-119100565 on Build GRCh38
rs775014419 Chr.11: 119100211-119100211 on Build GRCh38
rs767551785 Chr.11: 119100272-119100272 on Build GRCh38
rs746481248 Chr.11: 119100242-119100242 on Build GRCh38
rs751469003 Chr.11: 119100460-119100460 on Build GRCh38
rs201744612 Chr.11: 119100424-119100424 on Build GRCh38
rs879167295 Chr.11: 119100240-119100240 on Build GRCh38
rs532510371 Chr.11: 119100147-119100147 on Build GRCh38
rs763240506 Chr.11: 119100262-119100262 on Build GRCh38
rs200577447 Chr.11: 119100329-119100329 on Build GRCh38
rs918378436 Chr.11: 119100187-119100187 on Build GRCh38
rs778726543 Chr.11: 119100284-119100284 on Build GRCh38
rs766583767 Chr.11: 119100263-119100263 on Build GRCh38
rs746640458 Chr.11: 119100327-119100327 on Build GRCh38
rs755538085 Chr.11: 119100431-119100433 on Build GRCh38
rs770278685 Chr.11: 119100368-119100368 on Build GRCh38
rs997028828 Chr.11: 119100356-119100356 on Build GRCh38
rs759999337 Chr.11: 119100266-119100266 on Build GRCh38
rs951077823 Chr.11: 119100558-119100558 on Build GRCh38
rs761414084 Chr.11: 119100559-119100559 on Build GRCh38
rs556782038 Chr.11: 119100413-119100413 on Build GRCh38
rs561049226 Chr.11: 119100481-119100481 on Build GRCh38
rs760743805 Chr.11: 119100429-119100429 on Build GRCh38
rs977345953 Chr.11: 119100095-119100095 on Build GRCh38
rs751988655 Chr.11: 119100233-119100234 on Build GRCh38
rs603826 Chr.11: 119100168-119100168 on Build GRCh38
rs751784451 Chr.11: 119100570-119100571 on Build GRCh38
rs554742102 Chr.11: 119100249-119100249 on Build GRCh38
rs774754436 Chr.11: 119100260-119100260 on Build GRCh38
rs555565897 Chr.11: 119100144-119100144 on Build GRCh38
rs745647615 Chr.11: 119100231-119100231 on Build GRCh38
rs751396559 Chr.11: 119100493-119100493 on Build GRCh38
rs766806596 Chr.11: 119100231-119100231 on Build GRCh38
rs898521684 Chr.11: 119100205-119100205 on Build GRCh38
rs752838138 Chr.11: 119100274-119100274 on Build GRCh38
rs747719202 Chr.11: 119100347-119100347 on Build GRCh38
rs752325203 Chr.11: 119100235-119100235 on Build GRCh38
rs10892330 Chr.11: 119100165-119100165 on Build GRCh38
rs772626864 Chr.11: 119100301-119100301 on Build GRCh38
rs765185746 Chr.11: 119100434-119100434 on Build GRCh38
rs777262277 Chr.11: 119100451-119100453 on Build GRCh38
rs1024092849 Chr.11: 119100132-119100132 on Build GRCh38
rs863225088 Chr.11: 119100321-119100321 on Build GRCh38
rs767782520 Chr.11: 119100416-119100416 on Build GRCh38
rs185858377 Chr.11: 119100251-119100251 on Build GRCh38
rs974217692 Chr.11: 119100180-119100180 on Build GRCh38
rs141702694 Chr.11: 119100467-119100467 on Build GRCh38
rs1021369191 Chr.11: 119100387-119100387 on Build GRCh38
rs757171035 Chr.11: 119100275-119100275 on Build GRCh38
rs758161757 Chr.11: 119100316-119100316 on Build GRCh38
rs758649876 Chr.11: 119100225-119100233 on Build GRCh38
rs368867603 Chr.11: 119100451-119100451 on Build GRCh38
rs386758014 Chr.11: 119100098-119100100 on Build GRCh38
rs1008455980 Chr.11: 119100306-119100306 on Build GRCh38
rs768307038 Chr.11: 119100245-119100245 on Build GRCh38
rs773623306 Chr.11: 119100380-119100380 on Build GRCh38
rs758413791 Chr.11: 119100442-119100442 on Build GRCh38
rs750353546 Chr.11: 119100437-119100437 on Build GRCh38
rs201604403 Chr.11: 119100285-119100285 on Build GRCh38
rs34186993 Chr.11: 119100445-119100445 on Build GRCh38
rs62641715 Chr.11: 119100332-119100332 on Build GRCh38
rs1001422022 Chr.11: 119100150-119100150 on Build GRCh38
rs771175893 Chr.11: 119100412-119100412 on Build GRCh38
rs1010062032 Chr.11: 119100120-119100120 on Build GRCh38
rs750170790 Chr.11: 119100292-119100292 on Build GRCh38
rs649870 Chr.11: 119100541-119100541 on Build GRCh38
rs779710973 Chr.11: 119100317-119100317 on Build GRCh38

View Full Product Details