Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 112176240-112176620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCO2
Entrez Gene ID
83875
Gene Name
beta-carotene oxygenase 2
Gene Aliases
B-DIOX-II, BCDO2
UniGene
Hs.728325
Gene Chromosome Location
Chr.11: 112175485-112218929 on Build GRCh38
Transcripts
NM_001037290.2 NM_001256397.1 NM_001256398.1 NM_001256400.1 NM_031938.5

SNP Detail

SNP ID Chromosome Location
rs35450963 Chr.11: 112176518-112176519 on Build GRCh38
rs900687238 Chr.11: 112176309-112176309 on Build GRCh38
rs1026746818 Chr.11: 112176520-112176521 on Build GRCh38
rs1039140165 Chr.11: 112176296-112176296 on Build GRCh38
rs533653956 Chr.11: 112176306-112176306 on Build GRCh38
rs970588781 Chr.11: 112176406-112176406 on Build GRCh38
rs386374915 Chr.11: 112176527-112176528 on Build GRCh38
rs952058016 Chr.11: 112176522-112176523 on Build GRCh38
rs1006209704 Chr.11: 112176523-112176524 on Build GRCh38
rs868160576 Chr.11: 112176485-112176485 on Build GRCh38
rs1016358484 Chr.11: 112176343-112176343 on Build GRCh38
rs10431036 Chr.11: 112176338-112176338 on Build GRCh38
rs997716912 Chr.11: 112176318-112176318 on Build GRCh38
rs779391079 Chr.11: 112176514-112176515 on Build GRCh38
rs565669028 Chr.11: 112176384-112176384 on Build GRCh38
rs772731770 Chr.11: 112176517-112176518 on Build GRCh38
rs767243053 Chr.11: 112176554-112176555 on Build GRCh38
rs970124398 Chr.11: 112176537-112176537 on Build GRCh38
rs981595547 Chr.11: 112176349-112176349 on Build GRCh38
rs536775800 Chr.11: 112176522-112176522 on Build GRCh38
rs923908532 Chr.11: 112176496-112176496 on Build GRCh38
rs1030624699 Chr.11: 112176333-112176333 on Build GRCh38
rs893121423 Chr.11: 112176564-112176564 on Build GRCh38
rs554611857 Chr.11: 112176439-112176439 on Build GRCh38
rs1023086025 Chr.11: 112176524-112176525 on Build GRCh38
rs1049490784 Chr.11: 112176453-112176453 on Build GRCh38
rs947156197 Chr.11: 112176436-112176436 on Build GRCh38
rs1055138500 Chr.11: 112176536-112176536 on Build GRCh38
rs773111894 Chr.11: 112176486-112176486 on Build GRCh38
rs984846441 Chr.11: 112176325-112176325 on Build GRCh38
rs886088395 Chr.11: 112176334-112176334 on Build GRCh38
rs947493268 Chr.11: 112176280-112176280 on Build GRCh38
rs1023463675 Chr.11: 112176402-112176402 on Build GRCh38
rs61899683 Chr.11: 112176567-112176567 on Build GRCh38
rs1008049069 Chr.11: 112176519-112176520 on Build GRCh38
rs71060227 Chr.11: 112176528-112176529 on Build GRCh38
rs201677811 Chr.11: 112176517-112176517 on Build GRCh38
rs796777571 Chr.11: 112176518-112176518 on Build GRCh38
rs1026276337 Chr.11: 112176299-112176301 on Build GRCh38
rs186567844 Chr.11: 112176394-112176394 on Build GRCh38
rs567834015 Chr.11: 112176516-112176516 on Build GRCh38
rs914211966 Chr.11: 112176421-112176421 on Build GRCh38
rs934888138 Chr.11: 112176388-112176388 on Build GRCh38
rs981608474 Chr.11: 112176581-112176581 on Build GRCh38

View Full Product Details