Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 22469349-22469948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2CD5
Entrez Gene ID
9847
Gene Name
C2 calcium dependent domain containing 5
Gene Aliases
CDP138, KIAA0528
UniGene
Hs.271014
Gene Chromosome Location
Chr.12: 22448546-22544546 on Build GRCh38
Transcripts
NM_001286173.1 NM_001286174.1 NM_001286175.1 NM_001286176.1 NM_001286177.1 NM_014802.2 XM_005253538.2 XM_005253539.3 XM_006719170.2 XM_017020264.1 XM_017020265.1 XM_017020266.1 XM_017020267.1 XM_017020268.1 XM_017020269.1 XM_017020270.1 XM_017020271.1 XM_017020272.1 XM_017020273.1 XM_017020274.1 XM_017020275.1 XM_017020276.1 XM_017020277.1 XM_017020278.1 XM_017020279.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4481372 c.1946C>T p.S649L Chr.12: 22469741-22469741 on Build GRCh38
6615763 c.1961C>A p.P654H Chr.12: 22469726-22469726 on Build GRCh38
4947352 c.1900G>C p.D634H Chr.12: 22469787-22469787 on Build GRCh38
5840737 c.2533G>A p.A845T Chr.12: 22469709-22469709 on Build GRCh38
6615762 c.2516C>A p.P839H Chr.12: 22469726-22469726 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766688891 Chr.12: 22469832-22469832 on Build GRCh38
rs1023532239 Chr.12: 22469558-22469558 on Build GRCh38
rs767790227 Chr.12: 22469675-22469675 on Build GRCh38
rs927692893 Chr.12: 22469640-22469640 on Build GRCh38
rs969354510 Chr.12: 22469570-22469570 on Build GRCh38
rs527729055 Chr.12: 22469885-22469885 on Build GRCh38
rs775218647 Chr.12: 22469684-22469685 on Build GRCh38
rs773554823 Chr.12: 22469680-22469680 on Build GRCh38
rs751464436 Chr.12: 22469740-22469740 on Build GRCh38
rs768736681 Chr.12: 22469661-22469661 on Build GRCh38
rs951769839 Chr.12: 22469656-22469656 on Build GRCh38
rs745361913 Chr.12: 22469739-22469739 on Build GRCh38
rs533709099 Chr.12: 22469817-22469817 on Build GRCh38
rs775125870 Chr.12: 22469839-22469839 on Build GRCh38
rs781195596 Chr.12: 22469744-22469744 on Build GRCh38
rs749489666 Chr.12: 22469796-22469796 on Build GRCh38
rs184928160 Chr.12: 22469761-22469761 on Build GRCh38
rs578194078 Chr.12: 22469531-22469531 on Build GRCh38
rs974443569 Chr.12: 22469631-22469631 on Build GRCh38
rs143380735 Chr.12: 22469385-22469385 on Build GRCh38
rs897785785 Chr.12: 22469910-22469911 on Build GRCh38
rs552250605 Chr.12: 22469482-22469482 on Build GRCh38
rs771746368 Chr.12: 22469679-22469680 on Build GRCh38
rs373817352 Chr.12: 22469667-22469667 on Build GRCh38
rs768626962 Chr.12: 22469797-22469797 on Build GRCh38
rs936122603 Chr.12: 22469877-22469877 on Build GRCh38
rs952489586 Chr.12: 22469453-22469453 on Build GRCh38
rs1044517329 Chr.12: 22469474-22469474 on Build GRCh38
rs373426233 Chr.12: 22469858-22469858 on Build GRCh38
rs946863303 Chr.12: 22469766-22469766 on Build GRCh38
rs759253598 Chr.12: 22469683-22469683 on Build GRCh38
rs1014317841 Chr.12: 22469438-22469438 on Build GRCh38
rs1000207241 Chr.12: 22469486-22469486 on Build GRCh38
rs768372563 Chr.12: 22469517-22469517 on Build GRCh38
rs891820587 Chr.12: 22469916-22469916 on Build GRCh38
rs1033460194 Chr.12: 22469518-22469518 on Build GRCh38
rs189734026 Chr.12: 22469804-22469804 on Build GRCh38
rs1037182046 Chr.12: 22469408-22469408 on Build GRCh38
rs910144562 Chr.12: 22469383-22469383 on Build GRCh38
rs955981403 Chr.12: 22469520-22469520 on Build GRCh38
rs1018721115 Chr.12: 22469582-22469582 on Build GRCh38
rs748661728 Chr.12: 22469812-22469812 on Build GRCh38
rs16924973 Chr.12: 22469419-22469419 on Build GRCh38
rs956578275 Chr.12: 22469645-22469647 on Build GRCh38
rs367545136 Chr.12: 22469743-22469743 on Build GRCh38
rs780448873 Chr.12: 22469790-22469790 on Build GRCh38
rs773608267 Chr.12: 22469828-22469828 on Build GRCh38
rs984467882 Chr.12: 22469660-22469660 on Build GRCh38
rs761007224 Chr.12: 22469830-22469830 on Build GRCh38
rs764838485 Chr.12: 22469717-22469717 on Build GRCh38
rs1005399732 Chr.12: 22469477-22469477 on Build GRCh38
rs897896386 Chr.12: 22469412-22469412 on Build GRCh38
rs762317405 Chr.12: 22469672-22469672 on Build GRCh38
rs181942366 Chr.12: 22469588-22469588 on Build GRCh38
rs573046182 Chr.12: 22469925-22469925 on Build GRCh38
rs763590515 Chr.12: 22469736-22469736 on Build GRCh38
rs1050578091 Chr.12: 22469789-22469789 on Build GRCh38
rs376587653 Chr.12: 22469636-22469636 on Build GRCh38
rs112136054 Chr.12: 22469918-22469918 on Build GRCh38
rs774869482 Chr.12: 22469730-22469730 on Build GRCh38
rs202196779 Chr.12: 22469751-22469751 on Build GRCh38
rs756450806 Chr.12: 22469763-22469763 on Build GRCh38
rs752296378 Chr.12: 22469722-22469722 on Build GRCh38
rs945010180 Chr.12: 22469831-22469831 on Build GRCh38
rs917396485 Chr.12: 22469732-22469732 on Build GRCh38
rs760469891 Chr.12: 22469832-22469835 on Build GRCh38
rs774220193 Chr.12: 22469533-22469533 on Build GRCh38

View Full Product Details