Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 67298799-67299276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAND1
Entrez Gene ID
55832
Gene Name
cullin associated and neddylation dissociated 1
Gene Aliases
TIP120, TIP120A
UniGene
Hs.546407
Gene Chromosome Location
Chr.12: 67269281-67314692 on Build GRCh38
Transcripts
NM_018448.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1747205 c.877C>G p.H293D Chr.12: 67298972-67298972 on Build GRCh38
1199493 c.969G>T p.M323I Chr.12: 67299064-67299064 on Build GRCh38
6356828 c.877C>T p.H293Y Chr.12: 67298972-67298972 on Build GRCh38
5623790 c.929A>G p.N310S Chr.12: 67299024-67299024 on Build GRCh38
1705906 c.905T>C p.L302P Chr.12: 67299000-67299000 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs957487908 Chr.12: 67298924-67298924 on Build GRCh38
rs749693989 Chr.12: 67299075-67299075 on Build GRCh38
rs768058823 Chr.12: 67299115-67299115 on Build GRCh38
rs190843503 Chr.12: 67298906-67298906 on Build GRCh38
rs1009323270 Chr.12: 67299195-67299195 on Build GRCh38
rs751743076 Chr.12: 67299145-67299145 on Build GRCh38
rs1020802666 Chr.12: 67299199-67299199 on Build GRCh38
rs981874425 Chr.12: 67299222-67299222 on Build GRCh38
rs747689240 Chr.12: 67299035-67299040 on Build GRCh38
rs1007858948 Chr.12: 67298868-67298868 on Build GRCh38
rs996656856 Chr.12: 67298905-67298905 on Build GRCh38
rs374907934 Chr.12: 67298944-67298944 on Build GRCh38
rs918948679 Chr.12: 67298843-67298843 on Build GRCh38
rs776275317 Chr.12: 67299121-67299121 on Build GRCh38
rs910297373 Chr.12: 67298892-67298892 on Build GRCh38
rs1032973522 Chr.12: 67299170-67299170 on Build GRCh38
rs201748001 Chr.12: 67299015-67299015 on Build GRCh38
rs751760787 Chr.12: 67298978-67298978 on Build GRCh38
rs776449443 Chr.12: 67298907-67298907 on Build GRCh38
rs762868843 Chr.12: 67299139-67299139 on Build GRCh38
rs777666465 Chr.12: 67299130-67299130 on Build GRCh38
rs770504095 Chr.12: 67299204-67299204 on Build GRCh38
rs1000611527 Chr.12: 67299137-67299137 on Build GRCh38
rs1041971880 Chr.12: 67299109-67299109 on Build GRCh38
rs183461516 Chr.12: 67299180-67299180 on Build GRCh38
rs138846584 Chr.12: 67299007-67299007 on Build GRCh38
rs778233399 Chr.12: 67299055-67299055 on Build GRCh38
rs773534586 Chr.12: 67299038-67299043 on Build GRCh38
rs1012995687 Chr.12: 67299234-67299234 on Build GRCh38
rs780510457 Chr.12: 67298903-67298905 on Build GRCh38
rs971960873 Chr.12: 67298839-67298839 on Build GRCh38
rs767916183 Chr.12: 67298972-67298972 on Build GRCh38
rs893645442 Chr.12: 67299191-67299191 on Build GRCh38
rs746618717 Chr.12: 67299081-67299081 on Build GRCh38
rs367644026 Chr.12: 67299034-67299034 on Build GRCh38
rs769898396 Chr.12: 67299136-67299136 on Build GRCh38
rs200800485 Chr.12: 67298902-67298904 on Build GRCh38
rs1024173324 Chr.12: 67299239-67299243 on Build GRCh38
rs771246650 Chr.12: 67299078-67299078 on Build GRCh38
rs943719962 Chr.12: 67299122-67299122 on Build GRCh38
rs766223664 Chr.12: 67299144-67299144 on Build GRCh38
rs367933735 Chr.12: 67299130-67299130 on Build GRCh38
rs963969627 Chr.12: 67298889-67298889 on Build GRCh38
rs1047259251 Chr.12: 67299099-67299099 on Build GRCh38
rs937558251 Chr.12: 67299052-67299052 on Build GRCh38
rs566777202 Chr.12: 67298914-67298914 on Build GRCh38
rs996709226 Chr.12: 67299174-67299174 on Build GRCh38
rs913278935 Chr.12: 67298965-67298965 on Build GRCh38
rs562008785 Chr.12: 67298844-67298844 on Build GRCh38
rs35272662 Chr.12: 67299063-67299064 on Build GRCh38
rs886831914 Chr.12: 67299111-67299111 on Build GRCh38
rs1035930096 Chr.12: 67298893-67298893 on Build GRCh38
rs990193174 Chr.12: 67298947-67298947 on Build GRCh38
rs748606273 Chr.12: 67299013-67299013 on Build GRCh38
rs971201166 Chr.12: 67299249-67299249 on Build GRCh38
rs931429618 Chr.12: 67299093-67299093 on Build GRCh38
rs756709210 Chr.12: 67299035-67299035 on Build GRCh38
rs1019961087 Chr.12: 67298888-67298888 on Build GRCh38
rs758242738 Chr.12: 67298969-67298969 on Build GRCh38
rs755034334 Chr.12: 67298981-67298981 on Build GRCh38
rs11176676 Chr.12: 67299138-67299138 on Build GRCh38

View Full Product Details