Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 68481600-68482186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEPDC1
Entrez Gene ID
55635
Gene Name
DEP domain containing 1
Gene Aliases
DEP.8, DEPDC1-V2, DEPDC1A, SDP35
UniGene
Hs.445098
Gene Chromosome Location
Chr.1: 68474152-68497221 on Build GRCh38
Transcripts
NM_001114120.2 NM_017779.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4009698 c.1695C>T p.C565C Chr.1: 68482113-68482113 on Build GRCh38
1344099 c.1730T>G p.L577R Chr.1: 68482078-68482078 on Build GRCh38
5847270 c.1703C>T p.T568I Chr.1: 68482105-68482105 on Build GRCh38
5992893 c.1699A>C p.S567R Chr.1: 68482109-68482109 on Build GRCh38
4677065 c.1654G>A p.E552K Chr.1: 68482154-68482154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758311460 Chr.1: 68482027-68482027 on Build GRCh38
rs974513767 Chr.1: 68482010-68482010 on Build GRCh38
rs764099361 Chr.1: 68482075-68482075 on Build GRCh38
rs74704835 Chr.1: 68481635-68481635 on Build GRCh38
rs746267128 Chr.1: 68482115-68482115 on Build GRCh38
rs994045895 Chr.1: 68481942-68481942 on Build GRCh38
rs148276338 Chr.1: 68481896-68481896 on Build GRCh38
rs571907093 Chr.1: 68481749-68481749 on Build GRCh38
rs202089723 Chr.1: 68482163-68482163 on Build GRCh38
rs770589389 Chr.1: 68482104-68482104 on Build GRCh38
rs754677588 Chr.1: 68482061-68482061 on Build GRCh38
rs925228266 Chr.1: 68482111-68482111 on Build GRCh38
rs1033755616 Chr.1: 68482018-68482018 on Build GRCh38
rs1017316843 Chr.1: 68481983-68481983 on Build GRCh38
rs752668319 Chr.1: 68481906-68481906 on Build GRCh38
rs555155508 Chr.1: 68481902-68481902 on Build GRCh38
rs761756687 Chr.1: 68482164-68482164 on Build GRCh38
rs397816625 Chr.1: 68481640-68481641 on Build GRCh38
rs764992503 Chr.1: 68481858-68481858 on Build GRCh38
rs753696143 Chr.1: 68482053-68482053 on Build GRCh38
rs763034818 Chr.1: 68481650-68481650 on Build GRCh38
rs545409487 Chr.1: 68481824-68481824 on Build GRCh38
rs564267862 Chr.1: 68481965-68481965 on Build GRCh38
rs917764920 Chr.1: 68482088-68482088 on Build GRCh38
rs763446549 Chr.1: 68482149-68482149 on Build GRCh38
rs775162506 Chr.1: 68481705-68481706 on Build GRCh38
rs190519030 Chr.1: 68481993-68481993 on Build GRCh38
rs748028202 Chr.1: 68482068-68482068 on Build GRCh38
rs185594838 Chr.1: 68481951-68481951 on Build GRCh38
rs1038777691 Chr.1: 68481799-68481799 on Build GRCh38
rs572943175 Chr.1: 68482071-68482071 on Build GRCh38
rs558122294 Chr.1: 68481696-68481696 on Build GRCh38
rs776110439 Chr.1: 68482113-68482113 on Build GRCh38
rs917970477 Chr.1: 68481849-68481849 on Build GRCh38
rs868014999 Chr.1: 68481746-68481746 on Build GRCh38
rs560168786 Chr.1: 68482148-68482148 on Build GRCh38
rs968590614 Chr.1: 68481736-68481736 on Build GRCh38
rs954376860 Chr.1: 68481659-68481659 on Build GRCh38
rs777036652 Chr.1: 68481625-68481625 on Build GRCh38
rs917817389 Chr.1: 68481707-68481707 on Build GRCh38
rs1038354764 Chr.1: 68481911-68481912 on Build GRCh38
rs780284564 Chr.1: 68481634-68481635 on Build GRCh38
rs370058783 Chr.1: 68482001-68482001 on Build GRCh38
rs777412786 Chr.1: 68482099-68482099 on Build GRCh38
rs769997866 Chr.1: 68482143-68482143 on Build GRCh38
rs750477800 Chr.1: 68481627-68481628 on Build GRCh38
rs3833522 Chr.1: 68481635-68481636 on Build GRCh38
rs758420543 Chr.1: 68481628-68481629 on Build GRCh38
rs905458323 Chr.1: 68481895-68481895 on Build GRCh38
rs781466422 Chr.1: 68481641-68481642 on Build GRCh38
rs1017537295 Chr.1: 68481727-68481727 on Build GRCh38
rs891347306 Chr.1: 68481922-68481922 on Build GRCh38
rs746712086 Chr.1: 68482103-68482103 on Build GRCh38
rs200804950 Chr.1: 68482154-68482154 on Build GRCh38
rs766125125 Chr.1: 68482026-68482026 on Build GRCh38
rs758044681 Chr.1: 68482091-68482091 on Build GRCh38
rs143161368 Chr.1: 68481773-68481773 on Build GRCh38
rs553736867 Chr.1: 68481783-68481783 on Build GRCh38
rs945289153 Chr.1: 68481719-68481719 on Build GRCh38
rs1045004184 Chr.1: 68481865-68481865 on Build GRCh38
rs985771607 Chr.1: 68481687-68481687 on Build GRCh38
rs374973337 Chr.1: 68482064-68482064 on Build GRCh38
rs764095648 Chr.1: 68481651-68481651 on Build GRCh38
rs267598702 Chr.1: 68482086-68482086 on Build GRCh38
rs760003345 Chr.1: 68481632-68481632 on Build GRCh38
rs1007242336 Chr.1: 68481954-68481954 on Build GRCh38
rs377002285 Chr.1: 68481999-68481999 on Build GRCh38
rs575302323 Chr.1: 68481949-68481949 on Build GRCh38
rs775929453 Chr.1: 68481647-68481647 on Build GRCh38
rs1042041772 Chr.1: 68481745-68481745 on Build GRCh38

View Full Product Details