Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3548895-3549297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATRN
Entrez Gene ID
8455
Gene Name
attractin
Gene Aliases
DPPT-L, MGCA
UniGene
Hs.276252
Gene Chromosome Location
Chr.20: 3471018-3651122 on Build GRCh38
Transcripts
NM_001207047.2 NM_001323332.1 NM_139321.2 NM_139322.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3933857 c.992G>T p.R331L Chr.20: 3549218-3549218 on Build GRCh38
6278258 c.991C>T p.R331* Chr.20: 3549217-3549217 on Build GRCh38
1713110 c.980C>T p.S327L Chr.20: 3549206-3549206 on Build GRCh38
5883550 c.1036G>T p.A346S Chr.20: 3549262-3549262 on Build GRCh38
5732048 c.964G>C p.V322L Chr.20: 3549190-3549190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186249171 Chr.20: 3549060-3549060 on Build GRCh38
rs1028043917 Chr.20: 3549175-3549175 on Build GRCh38
rs533610281 Chr.20: 3549125-3549125 on Build GRCh38
rs550682204 Chr.20: 3549012-3549012 on Build GRCh38
rs760560964 Chr.20: 3549182-3549182 on Build GRCh38
rs961276222 Chr.20: 3549230-3549230 on Build GRCh38
rs772840717 Chr.20: 3549205-3549205 on Build GRCh38
rs377163365 Chr.20: 3549245-3549245 on Build GRCh38
rs1031294017 Chr.20: 3549098-3549098 on Build GRCh38
rs763251726 Chr.20: 3549231-3549231 on Build GRCh38
rs374861849 Chr.20: 3549094-3549095 on Build GRCh38
rs765324915 Chr.20: 3549188-3549188 on Build GRCh38
rs983913453 Chr.20: 3549184-3549184 on Build GRCh38
rs1031116629 Chr.20: 3548924-3548924 on Build GRCh38
rs528992747 Chr.20: 3548984-3548984 on Build GRCh38
rs773153051 Chr.20: 3549160-3549160 on Build GRCh38
rs183429110 Chr.20: 3549026-3549026 on Build GRCh38
rs374246567 Chr.20: 3549250-3549250 on Build GRCh38
rs751847658 Chr.20: 3549233-3549233 on Build GRCh38
rs748948672 Chr.20: 3549135-3549135 on Build GRCh38
rs764474867 Chr.20: 3549232-3549232 on Build GRCh38
rs564136600 Chr.20: 3548980-3548980 on Build GRCh38
rs750911567 Chr.20: 3549260-3549260 on Build GRCh38
rs771865961 Chr.20: 3549156-3549156 on Build GRCh38
rs954030033 Chr.20: 3549178-3549178 on Build GRCh38
rs761528436 Chr.20: 3549147-3549147 on Build GRCh38
rs909806675 Chr.20: 3549217-3549217 on Build GRCh38
rs998512930 Chr.20: 3549056-3549056 on Build GRCh38
rs955512859 Chr.20: 3548932-3548932 on Build GRCh38
rs757649933 Chr.20: 3549243-3549243 on Build GRCh38
rs965180330 Chr.20: 3549119-3549119 on Build GRCh38
rs942725309 Chr.20: 3548972-3548972 on Build GRCh38
rs919240372 Chr.20: 3549244-3549244 on Build GRCh38
rs750201064 Chr.20: 3549156-3549157 on Build GRCh38
rs986867766 Chr.20: 3548939-3548940 on Build GRCh38
rs774128912 Chr.20: 3549145-3549145 on Build GRCh38
rs779471638 Chr.20: 3549133-3549133 on Build GRCh38
rs779604268 Chr.20: 3549273-3549273 on Build GRCh38
rs911268231 Chr.20: 3548953-3548953 on Build GRCh38
rs147361324 Chr.20: 3549271-3549271 on Build GRCh38
rs756629370 Chr.20: 3549270-3549270 on Build GRCh38
rs563390966 Chr.20: 3549023-3549023 on Build GRCh38
rs752765173 Chr.20: 3549218-3549218 on Build GRCh38
rs768357426 Chr.20: 3549141-3549141 on Build GRCh38

View Full Product Details