Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 46366975-46367567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMO2
Entrez Gene ID
63916
Gene Name
engulfment and cell motility 2
Gene Aliases
CED-12, CED12, Ced-12A, ELMO-2
UniGene
Hs.210469
Gene Chromosome Location
Chr.20: 46366045-46407051 on Build GRCh38
Transcripts
NM_001318253.1 NM_133171.4 NM_182764.2 XM_005260496.3 XM_005260498.3 XM_005260499.3 XM_005260500.3 XM_005260501.4 XM_006723854.3 XM_017028009.1 XM_017028010.1 XM_017028011.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4680793 c.2032G>A p.D678N Chr.20: 46367491-46367491 on Build GRCh38
6289851 c.2017G>A p.E673K Chr.20: 46367506-46367506 on Build GRCh38
7319601 c.2100C>T p.P700P Chr.20: 46367423-46367423 on Build GRCh38
4098999 c.2047C>T p.L683L Chr.20: 46367476-46367476 on Build GRCh38
6334815 c.2068C>A p.R690R Chr.20: 46367455-46367455 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs200120295 Chr.20: 46367048-46367048 on Build GRCh38
rs141818894 Chr.20: 46367410-46367410 on Build GRCh38
rs36000025 Chr.20: 46367388-46367388 on Build GRCh38
rs772644424 Chr.20: 46367516-46367516 on Build GRCh38
rs752731509 Chr.20: 46367350-46367350 on Build GRCh38
rs367919241 Chr.20: 46367046-46367047 on Build GRCh38
rs921289345 Chr.20: 46367406-46367406 on Build GRCh38
rs890721579 Chr.20: 46367114-46367114 on Build GRCh38
rs928957256 Chr.20: 46367089-46367089 on Build GRCh38
rs759921910 Chr.20: 46367433-46367433 on Build GRCh38
rs34630674 Chr.20: 46367438-46367438 on Build GRCh38
rs202085930 Chr.20: 46367422-46367422 on Build GRCh38
rs1041056057 Chr.20: 46367054-46367054 on Build GRCh38
rs753155755 Chr.20: 46367443-46367443 on Build GRCh38
rs1002522772 Chr.20: 46367162-46367162 on Build GRCh38
rs992743946 Chr.20: 46367273-46367273 on Build GRCh38
rs746752883 Chr.20: 46367483-46367483 on Build GRCh38
rs2257495 Chr.20: 46367543-46367543 on Build GRCh38
rs1010986835 Chr.20: 46367170-46367170 on Build GRCh38
rs973141407 Chr.20: 46367037-46367037 on Build GRCh38
rs1009650309 Chr.20: 46367129-46367129 on Build GRCh38
rs149510828 Chr.20: 46367459-46367459 on Build GRCh38
rs188737696 Chr.20: 46367217-46367217 on Build GRCh38
rs754833560 Chr.20: 46367488-46367488 on Build GRCh38
rs777989540 Chr.20: 46367366-46367366 on Build GRCh38
rs13433084 Chr.20: 46367207-46367207 on Build GRCh38
rs781217194 Chr.20: 46367344-46367344 on Build GRCh38
rs946895887 Chr.20: 46367508-46367508 on Build GRCh38
rs1012845504 Chr.20: 46367250-46367250 on Build GRCh38
rs3179883 Chr.20: 46367039-46367039 on Build GRCh38
rs138167431 Chr.20: 46367101-46367101 on Build GRCh38
rs999949523 Chr.20: 46367209-46367209 on Build GRCh38
rs868613199 Chr.20: 46367517-46367517 on Build GRCh38
rs576573452 Chr.20: 46367287-46367287 on Build GRCh38
rs144969329 Chr.20: 46367361-46367361 on Build GRCh38
rs375262745 Chr.20: 46367449-46367449 on Build GRCh38
rs149135334 Chr.20: 46367213-46367213 on Build GRCh38
rs542123639 Chr.20: 46367495-46367495 on Build GRCh38
rs965437150 Chr.20: 46367355-46367355 on Build GRCh38
rs893980451 Chr.20: 46367119-46367119 on Build GRCh38
rs780047843 Chr.20: 46367397-46367397 on Build GRCh38
rs776733201 Chr.20: 46367409-46367409 on Build GRCh38
rs145079411 Chr.20: 46367456-46367456 on Build GRCh38
rs766720174 Chr.20: 46367340-46367340 on Build GRCh38
rs749472199 Chr.20: 46367375-46367375 on Build GRCh38
rs764140896 Chr.20: 46367455-46367455 on Build GRCh38
rs146276812 Chr.20: 46367430-46367430 on Build GRCh38
rs920298128 Chr.20: 46367060-46367060 on Build GRCh38
rs746916401 Chr.20: 46367398-46367398 on Build GRCh38
rs543867235 Chr.20: 46366994-46366994 on Build GRCh38
rs755210422 Chr.20: 46367016-46367016 on Build GRCh38
rs370400358 Chr.20: 46367432-46367432 on Build GRCh38
rs886037919 Chr.20: 46367443-46367443 on Build GRCh38
rs748938643 Chr.20: 46367519-46367519 on Build GRCh38
rs969876712 Chr.20: 46367291-46367291 on Build GRCh38
rs921538506 Chr.20: 46367077-46367077 on Build GRCh38
rs373827294 Chr.20: 46367343-46367343 on Build GRCh38
rs944959464 Chr.20: 46367115-46367115 on Build GRCh38
rs267605962 Chr.20: 46367423-46367423 on Build GRCh38
rs763312200 Chr.20: 46367329-46367329 on Build GRCh38
rs900672343 Chr.20: 46367159-46367159 on Build GRCh38
rs1039639226 Chr.20: 46367143-46367143 on Build GRCh38
rs767959037 Chr.20: 46367441-46367441 on Build GRCh38
rs756148777 Chr.20: 46367454-46367454 on Build GRCh38
rs902574608 Chr.20: 46367194-46367194 on Build GRCh38
rs144087106 Chr.20: 46367515-46367515 on Build GRCh38
rs536148604 Chr.20: 46366995-46366995 on Build GRCh38
rs566287464 Chr.20: 46367172-46367172 on Build GRCh38
rs748101681 Chr.20: 46367514-46367514 on Build GRCh38
rs960251833 Chr.20: 46367235-46367235 on Build GRCh38
rs770529461 Chr.20: 46367537-46367537 on Build GRCh38
rs902932690 Chr.20: 46367147-46367147 on Build GRCh38
rs771891555 Chr.20: 46367387-46367387 on Build GRCh38
rs6017798 Chr.20: 46367341-46367341 on Build GRCh38

View Full Product Details