Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 49372279-49372878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNB1
Entrez Gene ID
3745
Gene Name
potassium voltage-gated channel subfamily B member 1
Gene Aliases
DRK1, Kv2.1
UniGene
Hs.633143
Gene Chromosome Location
Chr.20: 49363877-49484033 on Build GRCh38
Transcripts
NM_004975.3 XM_006723784.3 XM_011528799.2

SNP Detail

SNP ID Chromosome Location
rs980464564 Chr.20: 49372394-49372394 on Build GRCh38
rs549177376 Chr.20: 49372848-49372848 on Build GRCh38
rs368326818 Chr.20: 49372311-49372311 on Build GRCh38
rs560878669 Chr.20: 49372494-49372494 on Build GRCh38
rs983154230 Chr.20: 49372602-49372602 on Build GRCh38
rs567724672 Chr.20: 49372851-49372851 on Build GRCh38
rs189296294 Chr.20: 49372665-49372665 on Build GRCh38
rs778391721 Chr.20: 49372393-49372393 on Build GRCh38
rs987891973 Chr.20: 49372345-49372345 on Build GRCh38
rs1051295 Chr.20: 49372368-49372368 on Build GRCh38
rs946100795 Chr.20: 49372803-49372803 on Build GRCh38
rs868736165 Chr.20: 49372524-49372524 on Build GRCh38
rs940132398 Chr.20: 49372781-49372781 on Build GRCh38
rs931837178 Chr.20: 49372719-49372719 on Build GRCh38
rs908702230 Chr.20: 49372735-49372735 on Build GRCh38
rs1014375569 Chr.20: 49372547-49372547 on Build GRCh38
rs768546168 Chr.20: 49372638-49372638 on Build GRCh38
rs547158736 Chr.20: 49372440-49372440 on Build GRCh38
rs144832197 Chr.20: 49372419-49372419 on Build GRCh38
rs756994408 Chr.20: 49372385-49372385 on Build GRCh38
rs763738558 Chr.20: 49372333-49372333 on Build GRCh38
rs912005056 Chr.20: 49372785-49372785 on Build GRCh38
rs557083250 Chr.20: 49372418-49372418 on Build GRCh38
rs956441453 Chr.20: 49372341-49372341 on Build GRCh38
rs183774230 Chr.20: 49372581-49372581 on Build GRCh38
rs887306310 Chr.20: 49372427-49372427 on Build GRCh38
rs1019193792 Chr.20: 49372632-49372632 on Build GRCh38
rs530869697 Chr.20: 49372838-49372838 on Build GRCh38
rs898584969 Chr.20: 49372485-49372485 on Build GRCh38
rs963334408 Chr.20: 49372637-49372637 on Build GRCh38
rs749314327 Chr.20: 49372700-49372700 on Build GRCh38
rs890538128 Chr.20: 49372439-49372439 on Build GRCh38
rs1008842173 Chr.20: 49372621-49372621 on Build GRCh38
rs1041734512 Chr.20: 49372810-49372810 on Build GRCh38
rs9798 Chr.20: 49372359-49372359 on Build GRCh38
rs926841662 Chr.20: 49372416-49372416 on Build GRCh38
rs746435805 Chr.20: 49372814-49372815 on Build GRCh38
rs917587375 Chr.20: 49372300-49372300 on Build GRCh38
rs193238145 Chr.20: 49372708-49372708 on Build GRCh38
rs771214876 Chr.20: 49372782-49372782 on Build GRCh38
rs777876952 Chr.20: 49372658-49372658 on Build GRCh38
rs1021959697 Chr.20: 49372668-49372668 on Build GRCh38
rs542847335 Chr.20: 49372481-49372481 on Build GRCh38
rs559724576 Chr.20: 49372648-49372648 on Build GRCh38
rs113974524 Chr.20: 49372544-49372544 on Build GRCh38
rs941916787 Chr.20: 49372376-49372376 on Build GRCh38
rs562061352 Chr.20: 49372536-49372536 on Build GRCh38
rs374585405 Chr.20: 49372684-49372684 on Build GRCh38
rs991102201 Chr.20: 49372788-49372788 on Build GRCh38
rs13516 Chr.20: 49372423-49372423 on Build GRCh38
rs973276126 Chr.20: 49372675-49372675 on Build GRCh38
rs967788171 Chr.20: 49372676-49372676 on Build GRCh38
rs551502484 Chr.20: 49372697-49372697 on Build GRCh38

View Full Product Details