Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38482955-38483550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX17
Entrez Gene ID
10521
Gene Name
DEAD-box helicase 17
Gene Aliases
P72, RH70
UniGene
Hs.528305
Gene Chromosome Location
Chr.22: 38483438-38506340 on Build GRCh38
Transcripts
NM_001098504.1 NM_006386.4 NM_006855.3

Gene Symbol
KDELR3
Entrez Gene ID
11015
Gene Name
KDEL endoplasmic reticulum protein retention receptor 3
Gene Aliases
ERD2L3
UniGene
Hs.745072
Gene Chromosome Location
Chr.22: 38468062-38483447 on Build GRCh38
Transcripts
NM_001098504.1 NM_006386.4 NM_006855.3

SNP Detail

SNP ID Chromosome Location
rs565112734 Chr.22: 38483392-38483392 on Build GRCh38
rs1045865406 Chr.22: 38483465-38483465 on Build GRCh38
rs573751035 Chr.22: 38483194-38483194 on Build GRCh38
rs773417435 Chr.22: 38483337-38483337 on Build GRCh38
rs144793274 Chr.22: 38483298-38483298 on Build GRCh38
rs527513609 Chr.22: 38483314-38483314 on Build GRCh38
rs901313626 Chr.22: 38483482-38483482 on Build GRCh38
rs1014826811 Chr.22: 38483036-38483036 on Build GRCh38
rs912852138 Chr.22: 38482997-38482997 on Build GRCh38
rs768097602 Chr.22: 38483216-38483217 on Build GRCh38
rs747419606 Chr.22: 38483056-38483056 on Build GRCh38
rs1039040008 Chr.22: 38483004-38483004 on Build GRCh38
rs761045418 Chr.22: 38483447-38483448 on Build GRCh38
rs16998989 Chr.22: 38483490-38483490 on Build GRCh38
rs950189055 Chr.22: 38483416-38483416 on Build GRCh38
rs940627786 Chr.22: 38483341-38483341 on Build GRCh38
rs918623897 Chr.22: 38483438-38483440 on Build GRCh38
rs545702537 Chr.22: 38483330-38483330 on Build GRCh38
rs1036660059 Chr.22: 38483398-38483398 on Build GRCh38
rs935965308 Chr.22: 38483351-38483351 on Build GRCh38
rs139900049 Chr.22: 38483071-38483071 on Build GRCh38
rs761621184 Chr.22: 38483116-38483116 on Build GRCh38
rs1020939994 Chr.22: 38483499-38483499 on Build GRCh38
rs958564754 Chr.22: 38483305-38483305 on Build GRCh38
rs779601089 Chr.22: 38482998-38482998 on Build GRCh38
rs927177799 Chr.22: 38483020-38483020 on Build GRCh38
rs889883759 Chr.22: 38483464-38483464 on Build GRCh38
rs2071887 Chr.22: 38483005-38483005 on Build GRCh38
rs185585718 Chr.22: 38483475-38483475 on Build GRCh38
rs775557002 Chr.22: 38483397-38483397 on Build GRCh38
rs917155296 Chr.22: 38483072-38483072 on Build GRCh38
rs769929594 Chr.22: 38483241-38483241 on Build GRCh38
rs539731433 Chr.22: 38482993-38482993 on Build GRCh38
rs780606028 Chr.22: 38483301-38483305 on Build GRCh38
rs925191597 Chr.22: 38483336-38483336 on Build GRCh38
rs942522264 Chr.22: 38483326-38483326 on Build GRCh38
rs995615316 Chr.22: 38483104-38483104 on Build GRCh38
rs1019963961 Chr.22: 38483262-38483263 on Build GRCh38
rs113770058 Chr.22: 38483232-38483232 on Build GRCh38
rs1053572048 Chr.22: 38483024-38483024 on Build GRCh38
rs555348091 Chr.22: 38483130-38483130 on Build GRCh38
rs773375646 Chr.22: 38483253-38483253 on Build GRCh38
rs972430088 Chr.22: 38483203-38483203 on Build GRCh38
rs898760768 Chr.22: 38483519-38483519 on Build GRCh38
rs891870656 Chr.22: 38483411-38483411 on Build GRCh38
rs564373179 Chr.22: 38483371-38483371 on Build GRCh38
rs1036995502 Chr.22: 38483087-38483089 on Build GRCh38
rs990392107 Chr.22: 38483226-38483227 on Build GRCh38
rs1048510164 Chr.22: 38483450-38483451 on Build GRCh38
rs891884800 Chr.22: 38483031-38483031 on Build GRCh38
rs1017155893 Chr.22: 38483170-38483170 on Build GRCh38
rs940577024 Chr.22: 38483075-38483075 on Build GRCh38
rs181487511 Chr.22: 38483096-38483096 on Build GRCh38
rs367596937 Chr.22: 38483195-38483195 on Build GRCh38
rs987035833 Chr.22: 38483262-38483262 on Build GRCh38
rs541173578 Chr.22: 38483277-38483277 on Build GRCh38
rs1017102237 Chr.22: 38483500-38483500 on Build GRCh38
rs748250557 Chr.22: 38483196-38483196 on Build GRCh38
rs111911559 Chr.22: 38483303-38483304 on Build GRCh38
rs936005015 Chr.22: 38483009-38483009 on Build GRCh38
rs371197046 Chr.22: 38483052-38483052 on Build GRCh38

View Full Product Details