Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 11455080-11455645 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E2F6
Entrez Gene ID
1876
Gene Name
E2F transcription factor 6
Gene Aliases
E2F-6
UniGene
Hs.603093
Gene Chromosome Location
Chr.2: 11444375-11466177 on Build GRCh38
Transcripts
NM_001278275.1 NM_001278276.1 NM_001278277.1 NM_001278278.1 NM_198256.3 NM_212540.2 NR_103490.1 XM_017003547.1 XM_017003548.1 XM_017003549.1

SNP Detail

SNP ID Chromosome Location
rs907778755 Chr.2: 11455586-11455586 on Build GRCh38
rs979364216 Chr.2: 11455250-11455251 on Build GRCh38
rs551576504 Chr.2: 11455544-11455544 on Build GRCh38
rs547410934 Chr.2: 11455487-11455487 on Build GRCh38
rs963546723 Chr.2: 11455234-11455234 on Build GRCh38
rs139355387 Chr.2: 11455522-11455522 on Build GRCh38
rs907747534 Chr.2: 11455308-11455308 on Build GRCh38
rs925175164 Chr.2: 11455271-11455271 on Build GRCh38
rs983370655 Chr.2: 11455585-11455585 on Build GRCh38
rs1018400019 Chr.2: 11455501-11455501 on Build GRCh38
rs1036774550 Chr.2: 11455415-11455415 on Build GRCh38
rs6736044 Chr.2: 11455570-11455570 on Build GRCh38
rs913761244 Chr.2: 11455206-11455206 on Build GRCh38
rs1021017567 Chr.2: 11455571-11455571 on Build GRCh38
rs528935023 Chr.2: 11455424-11455424 on Build GRCh38
rs906779567 Chr.2: 11455439-11455439 on Build GRCh38
rs1011063154 Chr.2: 11455493-11455493 on Build GRCh38
rs983251581 Chr.2: 11455300-11455300 on Build GRCh38
rs1033712210 Chr.2: 11455219-11455219 on Build GRCh38
rs894880183 Chr.2: 11455409-11455409 on Build GRCh38
rs760871893 Chr.2: 11455492-11455492 on Build GRCh38
rs917676311 Chr.2: 11455336-11455336 on Build GRCh38
rs549880808 Chr.2: 11455272-11455272 on Build GRCh38
rs561748006 Chr.2: 11455421-11455421 on Build GRCh38
rs1027844568 Chr.2: 11455119-11455119 on Build GRCh38
rs1046040850 Chr.2: 11455431-11455431 on Build GRCh38
rs898085061 Chr.2: 11455100-11455100 on Build GRCh38
rs370108359 Chr.2: 11455397-11455397 on Build GRCh38
rs76357606 Chr.2: 11455257-11455257 on Build GRCh38
rs778582384 Chr.2: 11455491-11455491 on Build GRCh38
rs764252302 Chr.2: 11455520-11455520 on Build GRCh38
rs545591643 Chr.2: 11455200-11455200 on Build GRCh38
rs188751596 Chr.2: 11455407-11455407 on Build GRCh38
rs901837765 Chr.2: 11455543-11455543 on Build GRCh38
rs73917203 Chr.2: 11455406-11455406 on Build GRCh38
rs941896800 Chr.2: 11455598-11455598 on Build GRCh38
rs116554320 Chr.2: 11455289-11455289 on Build GRCh38
rs911308952 Chr.2: 11455134-11455134 on Build GRCh38
rs77112633 Chr.2: 11455128-11455128 on Build GRCh38
rs975950453 Chr.2: 11455197-11455197 on Build GRCh38
rs181487270 Chr.2: 11455512-11455512 on Build GRCh38
rs966487449 Chr.2: 11455185-11455185 on Build GRCh38
rs186162773 Chr.2: 11455137-11455137 on Build GRCh38
rs534689796 Chr.2: 11455105-11455105 on Build GRCh38
rs916332143 Chr.2: 11455615-11455615 on Build GRCh38
rs1007488363 Chr.2: 11455136-11455136 on Build GRCh38
rs756753651 Chr.2: 11455474-11455474 on Build GRCh38
rs533378880 Chr.2: 11455154-11455154 on Build GRCh38
rs879149127 Chr.2: 11455502-11455502 on Build GRCh38
rs973180007 Chr.2: 11455613-11455613 on Build GRCh38
rs976767200 Chr.2: 11455196-11455196 on Build GRCh38

View Full Product Details