Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 25232846-25233417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNMT3A
Entrez Gene ID
1788
Gene Name
DNA methyltransferase 3 alpha
Gene Aliases
DNMT3A2, M.HsaIIIA, TBRS
UniGene
Hs.515840
Gene Chromosome Location
Chr.2: 25232961-25342590 on Build GRCh38
Transcripts
NM_001320893.1 NM_022552.4 NM_153759.3 NM_175629.2 NR_135490.1 XM_005264175.4 XM_005264177.4 XM_011532662.2 XM_011532663.2 XM_011532666.2 XM_011532667.2 XM_017003526.1 XM_017003527.1

SNP Detail

SNP ID Chromosome Location
rs768509440 Chr.2: 25233230-25233230 on Build GRCh38
rs540995300 Chr.2: 25233185-25233185 on Build GRCh38
rs1039414978 Chr.2: 25233105-25233105 on Build GRCh38
rs536862337 Chr.2: 25233276-25233276 on Build GRCh38
rs1006177558 Chr.2: 25232925-25232925 on Build GRCh38
rs564377554 Chr.2: 25233374-25233374 on Build GRCh38
rs886358873 Chr.2: 25233204-25233204 on Build GRCh38
rs948849301 Chr.2: 25233108-25233108 on Build GRCh38
rs900944391 Chr.2: 25232887-25232887 on Build GRCh38
rs1002179251 Chr.2: 25232959-25232962 on Build GRCh38
rs572153377 Chr.2: 25233275-25233275 on Build GRCh38
rs553948306 Chr.2: 25233207-25233207 on Build GRCh38
rs369187148 Chr.2: 25232878-25232878 on Build GRCh38
rs942322673 Chr.2: 25233081-25233081 on Build GRCh38
rs150488471 Chr.2: 25232890-25232890 on Build GRCh38
rs992613987 Chr.2: 25233054-25233056 on Build GRCh38
rs960808001 Chr.2: 25232989-25232991 on Build GRCh38
rs987087102 Chr.2: 25232926-25232926 on Build GRCh38
rs970450351 Chr.2: 25233311-25233311 on Build GRCh38
rs1031811636 Chr.2: 25233282-25233282 on Build GRCh38
rs567426281 Chr.2: 25232891-25232891 on Build GRCh38
rs186497151 Chr.2: 25233250-25233250 on Build GRCh38
rs1009570202 Chr.2: 25233258-25233258 on Build GRCh38
rs762214026 Chr.2: 25233129-25233129 on Build GRCh38
rs996307919 Chr.2: 25232897-25232897 on Build GRCh38
rs770167739 Chr.2: 25233241-25233241 on Build GRCh38
rs975070973 Chr.2: 25233288-25233288 on Build GRCh38
rs1028516947 Chr.2: 25232903-25232903 on Build GRCh38
rs1046813709 Chr.2: 25233169-25233169 on Build GRCh38
rs995971462 Chr.2: 25233225-25233225 on Build GRCh38
rs1021048431 Chr.2: 25233259-25233259 on Build GRCh38
rs900150331 Chr.2: 25233217-25233217 on Build GRCh38
rs922303874 Chr.2: 25233290-25233290 on Build GRCh38
rs775810747 Chr.2: 25233203-25233203 on Build GRCh38
rs1002148233 Chr.2: 25233209-25233209 on Build GRCh38
rs372931913 Chr.2: 25233362-25233362 on Build GRCh38
rs10538 Chr.2: 25233303-25233303 on Build GRCh38
rs980244463 Chr.2: 25233313-25233313 on Build GRCh38
rs867091878 Chr.2: 25233128-25233128 on Build GRCh38
rs180811074 Chr.2: 25233071-25233071 on Build GRCh38
rs538627138 Chr.2: 25233110-25233110 on Build GRCh38
rs990174409 Chr.2: 25233027-25233027 on Build GRCh38
rs1047675265 Chr.2: 25233181-25233181 on Build GRCh38
rs1038507064 Chr.2: 25233386-25233386 on Build GRCh38
rs980576116 Chr.2: 25233109-25233109 on Build GRCh38
rs1037383409 Chr.2: 25233210-25233210 on Build GRCh38
rs956289289 Chr.2: 25233295-25233295 on Build GRCh38
rs938727063 Chr.2: 25233340-25233340 on Build GRCh38
rs914529704 Chr.2: 25233072-25233072 on Build GRCh38
rs1035050112 Chr.2: 25232979-25232979 on Build GRCh38
rs537769164 Chr.2: 25232928-25232928 on Build GRCh38
rs145421172 Chr.2: 25233301-25233301 on Build GRCh38
rs928718784 Chr.2: 25233318-25233318 on Build GRCh38
rs1030488574 Chr.2: 25233226-25233226 on Build GRCh38
rs1035019440 Chr.2: 25233212-25233212 on Build GRCh38
rs747021738 Chr.2: 25233234-25233234 on Build GRCh38
rs955513570 Chr.2: 25232912-25232912 on Build GRCh38
rs1021650345 Chr.2: 25233310-25233310 on Build GRCh38
rs534641115 Chr.2: 25232898-25232898 on Build GRCh38
rs550011256 Chr.2: 25232954-25232954 on Build GRCh38

View Full Product Details