Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 98176009-98176598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FARP1
Entrez Gene ID
10160
Gene Name
FERM, ARH/RhoGEF and pleckstrin domain protein 1
Gene Aliases
CDEP, FARP1-IT1, PLEKHC2, PPP1R75
UniGene
Hs.403917
Gene Chromosome Location
Chr.13: 98142562-98449773 on Build GRCh38
Transcripts
NM_001001715.3 NM_001286839.1 NM_005766.3 XM_011521046.2 XM_017020312.1 NM_178861.4

Gene Symbol
RNF113B
Entrez Gene ID
140432
Gene Name
ring finger protein 113B
Gene Aliases
RNF161, ZNF183L1, bA10G5.1
UniGene
Hs.296045
Gene Chromosome Location
Chr.13: 98175785-98177267 on Build GRCh38
Transcripts
NM_001001715.3 NM_001286839.1 NM_005766.3 XM_011521046.2 XM_017020312.1 NM_178861.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6767220 c.910G>A p.A304T Chr.13: 98176327-98176327 on Build GRCh38
4721954 c.796C>T p.P266S Chr.13: 98176441-98176441 on Build GRCh38
6767221 c.962delA p.K321fs*>2 Chr.13: 98176170-98176170 on Build GRCh38
4048894 c.909C>T p.P303P Chr.13: 98176328-98176328 on Build GRCh38
4721952 c.938A>C p.K313T Chr.13: 98176299-98176299 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746445835 Chr.13: 98176268-98176268 on Build GRCh38
rs764494395 Chr.13: 98176139-98176139 on Build GRCh38
rs945497328 Chr.13: 98176114-98176114 on Build GRCh38
rs376932060 Chr.13: 98176244-98176244 on Build GRCh38
rs1043785861 Chr.13: 98176118-98176118 on Build GRCh38
rs575920246 Chr.13: 98176528-98176528 on Build GRCh38
rs774363097 Chr.13: 98176489-98176490 on Build GRCh38
rs760628427 Chr.13: 98176371-98176371 on Build GRCh38
rs564509369 Chr.13: 98176132-98176132 on Build GRCh38
rs758189740 Chr.13: 98176252-98176252 on Build GRCh38
rs761529542 Chr.13: 98176571-98176571 on Build GRCh38
rs201183546 Chr.13: 98176345-98176345 on Build GRCh38
rs768979308 Chr.13: 98176326-98176326 on Build GRCh38
rs748448691 Chr.13: 98176451-98176451 on Build GRCh38
rs1055268653 Chr.13: 98176186-98176187 on Build GRCh38
rs778236848 Chr.13: 98176526-98176526 on Build GRCh38
rs777223809 Chr.13: 98176409-98176409 on Build GRCh38
rs755629957 Chr.13: 98176519-98176519 on Build GRCh38
rs754767770 Chr.13: 98176237-98176237 on Build GRCh38
rs759621971 Chr.13: 98176536-98176536 on Build GRCh38
rs778664498 Chr.13: 98176141-98176141 on Build GRCh38
rs749811099 Chr.13: 98176386-98176386 on Build GRCh38
rs762063976 Chr.13: 98176194-98176194 on Build GRCh38
rs776077151 Chr.13: 98176174-98176174 on Build GRCh38
rs1024013513 Chr.13: 98176246-98176246 on Build GRCh38
rs780797199 Chr.13: 98176427-98176427 on Build GRCh38
rs772284861 Chr.13: 98176564-98176564 on Build GRCh38
rs747823372 Chr.13: 98176144-98176144 on Build GRCh38
rs115032993 Chr.13: 98176436-98176436 on Build GRCh38
rs753841984 Chr.13: 98176228-98176228 on Build GRCh38
rs375668710 Chr.13: 98176184-98176184 on Build GRCh38
rs538170646 Chr.13: 98176565-98176565 on Build GRCh38
rs993125147 Chr.13: 98176291-98176291 on Build GRCh38
rs574953217 Chr.13: 98176351-98176351 on Build GRCh38
rs774767836 Chr.13: 98176327-98176327 on Build GRCh38
rs754294962 Chr.13: 98176170-98176170 on Build GRCh38
rs757002188 Chr.13: 98176507-98176507 on Build GRCh38
rs373285066 Chr.13: 98176253-98176253 on Build GRCh38
rs151316967 Chr.13: 98176555-98176555 on Build GRCh38
rs748982430 Chr.13: 98176126-98176126 on Build GRCh38
rs765635600 Chr.13: 98176397-98176397 on Build GRCh38
rs759601974 Chr.13: 98176518-98176520 on Build GRCh38
rs961706108 Chr.13: 98176258-98176258 on Build GRCh38
rs751760297 Chr.13: 98176192-98176192 on Build GRCh38
rs762608112 Chr.13: 98176483-98176483 on Build GRCh38
rs759532026 Chr.13: 98176465-98176465 on Build GRCh38
rs778897644 Chr.13: 98176240-98176240 on Build GRCh38
rs1040801364 Chr.13: 98176103-98176103 on Build GRCh38
rs116765314 Chr.13: 98176562-98176562 on Build GRCh38
rs115127901 Chr.13: 98176095-98176095 on Build GRCh38
rs951974326 Chr.13: 98176066-98176066 on Build GRCh38
rs764464377 Chr.13: 98176186-98176186 on Build GRCh38
rs768572290 Chr.13: 98176133-98176133 on Build GRCh38
rs780669305 Chr.13: 98176513-98176513 on Build GRCh38
rs570754261 Chr.13: 98176169-98176169 on Build GRCh38
rs985947870 Chr.13: 98176088-98176088 on Build GRCh38
rs773880195 Chr.13: 98176157-98176160 on Build GRCh38
rs147722072 Chr.13: 98176415-98176415 on Build GRCh38
rs373084773 Chr.13: 98176142-98176142 on Build GRCh38
rs771241422 Chr.13: 98176533-98176533 on Build GRCh38
rs770024902 Chr.13: 98176541-98176541 on Build GRCh38
rs369128203 Chr.13: 98176248-98176248 on Build GRCh38
rs267603872 Chr.13: 98176292-98176292 on Build GRCh38
rs771437938 Chr.13: 98176419-98176419 on Build GRCh38
rs199776079 Chr.13: 98176473-98176473 on Build GRCh38
rs1055732127 Chr.13: 98176546-98176546 on Build GRCh38
rs752595311 Chr.13: 98176398-98176398 on Build GRCh38
rs775399101 Chr.13: 98176061-98176061 on Build GRCh38
rs377323551 Chr.13: 98176350-98176350 on Build GRCh38
rs182388789 Chr.13: 98176043-98176043 on Build GRCh38
rs775812030 Chr.13: 98176283-98176283 on Build GRCh38
rs376379120 Chr.13: 98176495-98176495 on Build GRCh38
rs200579942 Chr.13: 98176441-98176441 on Build GRCh38
rs771683618 Chr.13: 98176159-98176159 on Build GRCh38
rs762988535 Chr.13: 98176551-98176551 on Build GRCh38
rs369474969 Chr.13: 98176410-98176410 on Build GRCh38
rs766335721 Chr.13: 98176462-98176462 on Build GRCh38
rs779793046 Chr.13: 98176225-98176225 on Build GRCh38
rs749793696 Chr.13: 98176433-98176433 on Build GRCh38
rs910366947 Chr.13: 98176360-98176360 on Build GRCh38
rs115981649 Chr.13: 98176522-98176522 on Build GRCh38
rs749879415 Chr.13: 98176517-98176517 on Build GRCh38
rs779890630 Chr.13: 98176117-98176117 on Build GRCh38
rs373685549 Chr.13: 98176548-98176548 on Build GRCh38
rs770505949 Chr.13: 98176275-98176275 on Build GRCh38
rs553174924 Chr.13: 98176220-98176220 on Build GRCh38
rs753802105 Chr.13: 98176394-98176394 on Build GRCh38
rs767634227 Chr.13: 98176202-98176202 on Build GRCh38
rs540958770 Chr.13: 98176374-98176374 on Build GRCh38
rs910473874 Chr.13: 98176091-98176091 on Build GRCh38
rs548321071 Chr.13: 98176523-98176523 on Build GRCh38
rs777005575 Chr.13: 98176534-98176534 on Build GRCh38
rs142469809 Chr.13: 98176457-98176457 on Build GRCh38
rs751431106 Chr.13: 98176568-98176568 on Build GRCh38
rs772298086 Chr.13: 98176525-98176525 on Build GRCh38
rs199754785 Chr.13: 98176381-98176381 on Build GRCh38
rs749845859 Chr.13: 98176310-98176310 on Build GRCh38
rs751143273 Chr.13: 98176502-98176502 on Build GRCh38
rs913890188 Chr.13: 98176110-98176110 on Build GRCh38
rs925370874 Chr.13: 98176489-98176489 on Build GRCh38
rs199816159 Chr.13: 98176529-98176529 on Build GRCh38
rs772636294 Chr.13: 98176165-98176165 on Build GRCh38
rs758225579 Chr.13: 98176400-98176400 on Build GRCh38
rs773255110 Chr.13: 98176458-98176458 on Build GRCh38

View Full Product Details