Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 124289005-124289541 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM101A
Entrez Gene ID
144347
Gene Name
family with sequence similarity 101 member A
Gene Aliases
CFM2
UniGene
Hs.432901
Gene Chromosome Location
Chr.12: 124289164-124316024 on Build GRCh38
Transcripts
NM_181709.4 NM_001204299.1

Gene Symbol
ZNF664-FAM101A
Entrez Gene ID
100533183
Gene Name
filamin-interacting protein FAM101A
Gene Aliases
-
UniGene
Hs.432901
Gene Chromosome Location
Chr.12: 123973215-124316024 on Build GRCh38
Transcripts
NM_181709.4 NM_001204299.1

SNP Detail

SNP ID Chromosome Location
rs539979761 Chr.12: 124289349-124289349 on Build GRCh38
rs917905897 Chr.12: 124289106-124289106 on Build GRCh38
rs1053204922 Chr.12: 124289282-124289282 on Build GRCh38
rs932608221 Chr.12: 124289376-124289376 on Build GRCh38
rs552619950 Chr.12: 124289026-124289026 on Build GRCh38
rs1031361125 Chr.12: 124289083-124289083 on Build GRCh38
rs1013733763 Chr.12: 124289466-124289466 on Build GRCh38
rs909277495 Chr.12: 124289166-124289166 on Build GRCh38
rs972588284 Chr.12: 124289359-124289359 on Build GRCh38
rs11834869 Chr.12: 124289148-124289148 on Build GRCh38
rs950963045 Chr.12: 124289509-124289509 on Build GRCh38
rs139394460 Chr.12: 124289449-124289449 on Build GRCh38
rs1050811382 Chr.12: 124289210-124289210 on Build GRCh38
rs912451689 Chr.12: 124289388-124289388 on Build GRCh38
rs866191179 Chr.12: 124289324-124289324 on Build GRCh38
rs1001034217 Chr.12: 124289295-124289295 on Build GRCh38
rs959647012 Chr.12: 124289315-124289315 on Build GRCh38
rs773206967 Chr.12: 124289360-124289360 on Build GRCh38
rs919362303 Chr.12: 124289162-124289162 on Build GRCh38
rs921083394 Chr.12: 124289375-124289375 on Build GRCh38
rs1051471856 Chr.12: 124289385-124289385 on Build GRCh38
rs926237005 Chr.12: 124289406-124289406 on Build GRCh38
rs749046148 Chr.12: 124289264-124289264 on Build GRCh38
rs184102894 Chr.12: 124289404-124289404 on Build GRCh38
rs377531147 Chr.12: 124289510-124289510 on Build GRCh38
rs984014982 Chr.12: 124289122-124289122 on Build GRCh38
rs144165611 Chr.12: 124289487-124289487 on Build GRCh38
rs965081731 Chr.12: 124289090-124289090 on Build GRCh38
rs572685129 Chr.12: 124289347-124289347 on Build GRCh38
rs745584633 Chr.12: 124289163-124289163 on Build GRCh38
rs961170503 Chr.12: 124289135-124289136 on Build GRCh38
rs1043261695 Chr.12: 124289226-124289227 on Build GRCh38
rs1016836385 Chr.12: 124289345-124289345 on Build GRCh38
rs950797408 Chr.12: 124289335-124289335 on Build GRCh38
rs1033843513 Chr.12: 124289296-124289296 on Build GRCh38
rs889467575 Chr.12: 124289501-124289501 on Build GRCh38
rs1057449636 Chr.12: 124289189-124289189 on Build GRCh38
rs915911836 Chr.12: 124289400-124289400 on Build GRCh38
rs774199940 Chr.12: 124289343-124289343 on Build GRCh38
rs770185736 Chr.12: 124289099-124289103 on Build GRCh38
rs954139660 Chr.12: 124289120-124289120 on Build GRCh38
rs750257784 Chr.12: 124289205-124289205 on Build GRCh38
rs959570395 Chr.12: 124289065-124289065 on Build GRCh38
rs901302689 Chr.12: 124289439-124289439 on Build GRCh38
rs558614847 Chr.12: 124289403-124289403 on Build GRCh38
rs1000610959 Chr.12: 124289434-124289434 on Build GRCh38
rs146247657 Chr.12: 124289333-124289333 on Build GRCh38
rs973917458 Chr.12: 124289373-124289373 on Build GRCh38
rs890767317 Chr.12: 124289232-124289232 on Build GRCh38
rs368160235 Chr.12: 124289123-124289123 on Build GRCh38
rs546315454 Chr.12: 124289171-124289171 on Build GRCh38
rs374879371 Chr.12: 124289415-124289415 on Build GRCh38
rs143347067 Chr.12: 124289089-124289089 on Build GRCh38
rs755890734 Chr.12: 124289310-124289310 on Build GRCh38
rs78015427 Chr.12: 124289325-124289325 on Build GRCh38
rs191576774 Chr.12: 124289350-124289350 on Build GRCh38
rs186678873 Chr.12: 124289201-124289201 on Build GRCh38
rs903569149 Chr.12: 124289255-124289255 on Build GRCh38
rs780010890 Chr.12: 124289249-124289249 on Build GRCh38
rs941212756 Chr.12: 124289169-124289169 on Build GRCh38
rs373291459 Chr.12: 124289435-124289435 on Build GRCh38
rs1027981748 Chr.12: 124289108-124289108 on Build GRCh38
rs921183443 Chr.12: 124289190-124289190 on Build GRCh38
rs183455959 Chr.12: 124289170-124289170 on Build GRCh38
rs992213276 Chr.12: 124289081-124289081 on Build GRCh38
rs999003750 Chr.12: 124289261-124289261 on Build GRCh38
rs1009210927 Chr.12: 124289247-124289247 on Build GRCh38
rs1033936939 Chr.12: 124289049-124289049 on Build GRCh38
rs772107767 Chr.12: 124289344-124289344 on Build GRCh38

View Full Product Details