Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 49278664-49279260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBLN1
Entrez Gene ID
869
Gene Name
cerebellin 1 precursor
Gene Aliases
-
UniGene
Hs.458423
Gene Chromosome Location
Chr.16: 49277917-49281831 on Build GRCh38
Transcripts
NM_004352.3

SNP Detail

SNP ID Chromosome Location
rs921863050 Chr.16: 49278931-49278933 on Build GRCh38
rs367994175 Chr.16: 49278816-49278816 on Build GRCh38
rs963086892 Chr.16: 49279237-49279237 on Build GRCh38
rs1051515207 Chr.16: 49278732-49278732 on Build GRCh38
rs994080 Chr.16: 49278872-49278872 on Build GRCh38
rs773901681 Chr.16: 49278756-49278756 on Build GRCh38
rs775744661 Chr.16: 49278901-49278901 on Build GRCh38
rs748395079 Chr.16: 49279198-49279198 on Build GRCh38
rs78493955 Chr.16: 49278970-49278970 on Build GRCh38
rs540970970 Chr.16: 49278763-49278763 on Build GRCh38
rs771721834 Chr.16: 49278828-49278828 on Build GRCh38
rs1051589254 Chr.16: 49278955-49278955 on Build GRCh38
rs1047429615 Chr.16: 49278767-49278767 on Build GRCh38
rs890239918 Chr.16: 49278734-49278734 on Build GRCh38
rs997040356 Chr.16: 49278930-49278930 on Build GRCh38
rs995971127 Chr.16: 49278725-49278725 on Build GRCh38
rs1816580 Chr.16: 49278761-49278761 on Build GRCh38
rs890267687 Chr.16: 49278957-49278957 on Build GRCh38
rs1029376791 Chr.16: 49278936-49278936 on Build GRCh38
rs994079 Chr.16: 49278757-49278757 on Build GRCh38
rs540616212 Chr.16: 49279030-49279030 on Build GRCh38
rs963700142 Chr.16: 49278912-49278912 on Build GRCh38
rs904246155 Chr.16: 49279011-49279011 on Build GRCh38
rs949936237 Chr.16: 49279207-49279207 on Build GRCh38
rs941389283 Chr.16: 49278907-49278907 on Build GRCh38
rs1033981242 Chr.16: 49279037-49279037 on Build GRCh38
rs373717138 Chr.16: 49278979-49278980 on Build GRCh38
rs3743776 Chr.16: 49279214-49279214 on Build GRCh38
rs1041478228 Chr.16: 49278970-49278970 on Build GRCh38
rs866320018 Chr.16: 49278780-49278780 on Build GRCh38
rs566699016 Chr.16: 49278960-49278960 on Build GRCh38
rs988300011 Chr.16: 49279029-49279029 on Build GRCh38
rs41210 Chr.16: 49278762-49278762 on Build GRCh38
rs188920933 Chr.16: 49279121-49279121 on Build GRCh38
rs1014511118 Chr.16: 49279045-49279045 on Build GRCh38
rs1040073766 Chr.16: 49278930-49278931 on Build GRCh38
rs768979344 Chr.16: 49279041-49279042 on Build GRCh38
rs909972147 Chr.16: 49279236-49279236 on Build GRCh38
rs386790577 Chr.16: 49278761-49278762 on Build GRCh38
rs982874639 Chr.16: 49278896-49278896 on Build GRCh38
rs909929238 Chr.16: 49278898-49278898 on Build GRCh38
rs968292764 Chr.16: 49279039-49279039 on Build GRCh38
rs955738560 Chr.16: 49279021-49279021 on Build GRCh38
rs770635057 Chr.16: 49278741-49278741 on Build GRCh38
rs917412998 Chr.16: 49278843-49278843 on Build GRCh38
rs1016639109 Chr.16: 49278878-49278878 on Build GRCh38
rs917973986 Chr.16: 49279188-49279188 on Build GRCh38
rs189527594 Chr.16: 49279033-49279033 on Build GRCh38
rs565221438 Chr.16: 49278962-49278962 on Build GRCh38
rs1056953017 Chr.16: 49279173-49279173 on Build GRCh38
rs185504367 Chr.16: 49278916-49278916 on Build GRCh38
rs958904393 Chr.16: 49278792-49278792 on Build GRCh38
rs886995459 Chr.16: 49278822-49278822 on Build GRCh38
rs182271774 Chr.16: 49279105-49279105 on Build GRCh38
rs975475184 Chr.16: 49279238-49279238 on Build GRCh38
rs577167117 Chr.16: 49278969-49278970 on Build GRCh38
rs77591139 Chr.16: 49279042-49279042 on Build GRCh38
rs926658640 Chr.16: 49279077-49279077 on Build GRCh38
rs371886000 Chr.16: 49279057-49279057 on Build GRCh38
rs1009937519 Chr.16: 49278745-49278745 on Build GRCh38

View Full Product Details