Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 81262022-81262422 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCO1
Entrez Gene ID
53630
Gene Name
beta-carotene oxygenase 1
Gene Aliases
BCDO, BCDO1, BCMO, BCMO1, BCO
UniGene
Hs.212172
Gene Chromosome Location
Chr.16: 81238642-81291142 on Build GRCh38
Transcripts
NM_017429.2 XM_011523109.1 XM_017023286.1 XM_017023287.1 XM_017023288.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5851566 c.471+1G>A p.? Chr.16: 81262284-81262284 on Build GRCh38
396438 c.444C>A p.N148K Chr.16: 81262256-81262256 on Build GRCh38
2693250 c.466G>A p.E156K Chr.16: 81262278-81262278 on Build GRCh38
2693248 c.410C>T p.A137V Chr.16: 81262222-81262222 on Build GRCh38
1380069 c.408C>T p.Y136Y Chr.16: 81262220-81262220 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746790957 Chr.16: 81262207-81262207 on Build GRCh38
rs200428274 Chr.16: 81262222-81262222 on Build GRCh38
rs912504653 Chr.16: 81262173-81262173 on Build GRCh38
rs749838835 Chr.16: 81262283-81262283 on Build GRCh38
rs763233403 Chr.16: 81262315-81262315 on Build GRCh38
rs752480679 Chr.16: 81262164-81262164 on Build GRCh38
rs147092975 Chr.16: 81262108-81262108 on Build GRCh38
rs867803292 Chr.16: 81262194-81262194 on Build GRCh38
rs775757317 Chr.16: 81262309-81262309 on Build GRCh38
rs972542317 Chr.16: 81262185-81262185 on Build GRCh38
rs769523486 Chr.16: 81262120-81262120 on Build GRCh38
rs765487181 Chr.16: 81262233-81262233 on Build GRCh38
rs766347639 Chr.16: 81262147-81262147 on Build GRCh38
rs370376971 Chr.16: 81262224-81262224 on Build GRCh38
rs760655707 Chr.16: 81262339-81262339 on Build GRCh38
rs767322152 Chr.16: 81262335-81262335 on Build GRCh38
rs769062936 Chr.16: 81262319-81262319 on Build GRCh38
rs767505930 Chr.16: 81262274-81262274 on Build GRCh38
rs773962707 Chr.16: 81262320-81262320 on Build GRCh38
rs760677900 Chr.16: 81262085-81262085 on Build GRCh38
rs35683292 Chr.16: 81262175-81262175 on Build GRCh38
rs754613556 Chr.16: 81262346-81262346 on Build GRCh38
rs116347061 Chr.16: 81262306-81262306 on Build GRCh38
rs369543338 Chr.16: 81262114-81262114 on Build GRCh38
rs753414853 Chr.16: 81262343-81262343 on Build GRCh38
rs761594340 Chr.16: 81262156-81262158 on Build GRCh38
rs376471797 Chr.16: 81262189-81262189 on Build GRCh38
rs773740943 Chr.16: 81262133-81262133 on Build GRCh38
rs866662767 Chr.16: 81262269-81262269 on Build GRCh38
rs886153068 Chr.16: 81262201-81262201 on Build GRCh38
rs745894707 Chr.16: 81262305-81262305 on Build GRCh38
rs766585824 Chr.16: 81262089-81262089 on Build GRCh38
rs750400189 Chr.16: 81262337-81262337 on Build GRCh38
rs201662228 Chr.16: 81262242-81262242 on Build GRCh38
rs771283822 Chr.16: 81262298-81262298 on Build GRCh38
rs199834539 Chr.16: 81262220-81262220 on Build GRCh38
rs776813359 Chr.16: 81262153-81262153 on Build GRCh38
rs1025671846 Chr.16: 81262043-81262043 on Build GRCh38
rs373346551 Chr.16: 81262177-81262177 on Build GRCh38
rs930314881 Chr.16: 81262379-81262379 on Build GRCh38
rs987915412 Chr.16: 81262348-81262348 on Build GRCh38
rs561466042 Chr.16: 81262115-81262115 on Build GRCh38
rs920276056 Chr.16: 81262136-81262136 on Build GRCh38
rs752605503 Chr.16: 81262097-81262097 on Build GRCh38
rs1035764515 Chr.16: 81262070-81262070 on Build GRCh38
rs569702266 Chr.16: 81262287-81262287 on Build GRCh38
rs751909346 Chr.16: 81262237-81262238 on Build GRCh38
rs185021473 Chr.16: 81262096-81262096 on Build GRCh38
rs754031707 Chr.16: 81262092-81262092 on Build GRCh38
rs574989597 Chr.16: 81262383-81262383 on Build GRCh38
rs754667038 Chr.16: 81262291-81262291 on Build GRCh38
rs758396669 Chr.16: 81262101-81262101 on Build GRCh38
rs931079058 Chr.16: 81262193-81262193 on Build GRCh38
rs776764772 Chr.16: 81262253-81262253 on Build GRCh38
rs757134025 Chr.16: 81262273-81262273 on Build GRCh38
rs370355631 Chr.16: 81262202-81262202 on Build GRCh38
rs564498764 Chr.16: 81262076-81262076 on Build GRCh38
rs1031187081 Chr.16: 81262301-81262301 on Build GRCh38
rs751408559 Chr.16: 81262270-81262270 on Build GRCh38
rs775884304 Chr.16: 81262239-81262239 on Build GRCh38
rs758271353 Chr.16: 81262166-81262166 on Build GRCh38
rs764080372 Chr.16: 81262167-81262167 on Build GRCh38
rs773618146 Chr.16: 81262215-81262215 on Build GRCh38
rs149750747 Chr.16: 81262284-81262284 on Build GRCh38
rs748475104 Chr.16: 81262205-81262205 on Build GRCh38
rs780533156 Chr.16: 81262176-81262176 on Build GRCh38
rs376055699 Chr.16: 81262244-81262244 on Build GRCh38
rs772450083 Chr.16: 81262206-81262206 on Build GRCh38
rs965681998 Chr.16: 81262387-81262387 on Build GRCh38
rs781501121 Chr.16: 81262350-81262350 on Build GRCh38
rs759672104 Chr.16: 81262160-81262160 on Build GRCh38
rs769717305 Chr.16: 81262347-81262347 on Build GRCh38
rs1046638346 Chr.16: 81262221-81262221 on Build GRCh38
rs199710262 Chr.16: 81262138-81262138 on Build GRCh38
rs1010868122 Chr.16: 81262293-81262293 on Build GRCh38
rs757671439 Chr.16: 81262349-81262349 on Build GRCh38
rs764802608 Chr.16: 81262161-81262162 on Build GRCh38
rs761676041 Chr.16: 81262323-81262323 on Build GRCh38
rs772865830 Chr.16: 81262168-81262169 on Build GRCh38
rs138370116 Chr.16: 81262302-81262302 on Build GRCh38
rs970993374 Chr.16: 81262045-81262045 on Build GRCh38
rs983169170 Chr.16: 81262402-81262402 on Build GRCh38
rs745483499 Chr.16: 81262116-81262116 on Build GRCh38
rs181156113 Chr.16: 81262303-81262303 on Build GRCh38
rs765845201 Chr.16: 81262342-81262342 on Build GRCh38
rs1000138679 Chr.16: 81262057-81262057 on Build GRCh38
rs747993367 Chr.16: 81262079-81262079 on Build GRCh38

View Full Product Details