Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42909144-42909565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G6PC
Entrez Gene ID
2538
Gene Name
glucose-6-phosphatase catalytic subunit
Gene Aliases
G6PC1, G6PT, G6Pase, GSD1, GSD1a
UniGene
Hs.212293
Gene Chromosome Location
Chr.17: 42900797-42914433 on Build GRCh38
Transcripts
NM_000151.3 NM_001270397.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6668277 c.500G>A p.C167Y Chr.17: 42909356-42909356 on Build GRCh38
979694 c.559T>C p.S187P Chr.17: 42909415-42909415 on Build GRCh38
7184870 c.499_500delTG p.C167fs*36 Chr.17: 42909355-42909356 on Build GRCh38
3517836 c.556C>T p.L186L Chr.17: 42909412-42909412 on Build GRCh38
7140262 c.509G>T p.R170L Chr.17: 42909365-42909365 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs889507660 Chr.17: 42909221-42909221 on Build GRCh38
rs935358064 Chr.17: 42909178-42909178 on Build GRCh38
rs161623 Chr.17: 42909245-42909245 on Build GRCh38
rs751810814 Chr.17: 42909225-42909225 on Build GRCh38
rs750188110 Chr.17: 42909294-42909294 on Build GRCh38
rs771720935 Chr.17: 42909327-42909327 on Build GRCh38
rs935325575 Chr.17: 42909449-42909449 on Build GRCh38
rs780486456 Chr.17: 42909465-42909465 on Build GRCh38
rs1052952321 Chr.17: 42909199-42909199 on Build GRCh38
rs373345919 Chr.17: 42909364-42909364 on Build GRCh38
rs141376085 Chr.17: 42909414-42909414 on Build GRCh38
rs766230068 Chr.17: 42909409-42909409 on Build GRCh38
rs763543607 Chr.17: 42909388-42909388 on Build GRCh38
rs199505156 Chr.17: 42909428-42909428 on Build GRCh38
rs104894571 Chr.17: 42909353-42909353 on Build GRCh38
rs767885807 Chr.17: 42909287-42909287 on Build GRCh38
rs542754290 Chr.17: 42909204-42909204 on Build GRCh38
rs762917407 Chr.17: 42909267-42909267 on Build GRCh38
rs375921716 Chr.17: 42909405-42909405 on Build GRCh38
rs999908615 Chr.17: 42909476-42909476 on Build GRCh38
rs753856233 Chr.17: 42909306-42909306 on Build GRCh38
rs1022672212 Chr.17: 42909463-42909463 on Build GRCh38
rs747740866 Chr.17: 42909315-42909315 on Build GRCh38
rs549712764 Chr.17: 42909303-42909303 on Build GRCh38
rs756002909 Chr.17: 42909299-42909299 on Build GRCh38
rs1057516630 Chr.17: 42909355-42909356 on Build GRCh38
rs80356482 Chr.17: 42909418-42909418 on Build GRCh38
rs777552825 Chr.17: 42909356-42909356 on Build GRCh38
rs774585761 Chr.17: 42909390-42909390 on Build GRCh38
rs775826449 Chr.17: 42909374-42909374 on Build GRCh38
rs567279632 Chr.17: 42909184-42909184 on Build GRCh38
rs200905457 Chr.17: 42909415-42909415 on Build GRCh38
rs1050026736 Chr.17: 42909212-42909212 on Build GRCh38
rs746712553 Chr.17: 42909454-42909454 on Build GRCh38
rs770010331 Chr.17: 42909464-42909464 on Build GRCh38
rs71359588 Chr.17: 42909452-42909453 on Build GRCh38
rs978584776 Chr.17: 42909191-42909191 on Build GRCh38
rs757572132 Chr.17: 42909289-42909289 on Build GRCh38
rs979468295 Chr.17: 42909421-42909421 on Build GRCh38
rs987462071 Chr.17: 42909346-42909346 on Build GRCh38
rs1037751173 Chr.17: 42909417-42909417 on Build GRCh38
rs767362252 Chr.17: 42909290-42909290 on Build GRCh38
rs1035278041 Chr.17: 42909395-42909395 on Build GRCh38
rs104894569 Chr.17: 42909407-42909407 on Build GRCh38
rs778417581 Chr.17: 42909314-42909314 on Build GRCh38
rs191213146 Chr.17: 42909482-42909482 on Build GRCh38
rs1013917574 Chr.17: 42909458-42909458 on Build GRCh38
rs901119743 Chr.17: 42909202-42909202 on Build GRCh38
rs764307217 Chr.17: 42909278-42909278 on Build GRCh38
rs369892096 Chr.17: 42909309-42909309 on Build GRCh38
rs161622 Chr.17: 42909472-42909472 on Build GRCh38
rs180807007 Chr.17: 42909504-42909504 on Build GRCh38
rs750470654 Chr.17: 42909365-42909365 on Build GRCh38
rs374531217 Chr.17: 42909239-42909239 on Build GRCh38
rs969826245 Chr.17: 42909168-42909168 on Build GRCh38
rs767846924 Chr.17: 42909403-42909403 on Build GRCh38
rs776126016 Chr.17: 42909254-42909254 on Build GRCh38
rs751788357 Chr.17: 42909282-42909282 on Build GRCh38
rs560809795 Chr.17: 42909235-42909235 on Build GRCh38
rs777271010 Chr.17: 42909439-42909439 on Build GRCh38
rs768803329 Chr.17: 42909389-42909389 on Build GRCh38

View Full Product Details