Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 72727281-72727880 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BORA
Entrez Gene ID
79866
Gene Name
bora, aurora kinase A activator
Gene Aliases
C13orf34
UniGene
Hs.643464
Gene Chromosome Location
Chr.13: 72727739-72756198 on Build GRCh38
Transcripts
NM_001286747.1 NM_024808.3 XM_006719868.3 NM_001071775.2

Gene Symbol
MZT1
Entrez Gene ID
440145
Gene Name
mitotic spindle organizing protein 1
Gene Aliases
C13orf37, MOZART1
UniGene
Hs.28465
Gene Chromosome Location
Chr.13: 72708357-72727800 on Build GRCh38
Transcripts
NM_001286747.1 NM_024808.3 XM_006719868.3 NM_001071775.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6721298 c.30G>C p.A10A Chr.13: 72727573-72727573 on Build GRCh38
1659960 c.64C>T p.R22W Chr.13: 72727539-72727539 on Build GRCh38
6855675 c.61G>A p.V21M Chr.13: 72727542-72727542 on Build GRCh38
297763 c.36_38delGGC p.A16delA Chr.13: 72727565-72727567 on Build GRCh38
3753731 c.13A>G p.S5G Chr.13: 72727590-72727590 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs76288727 Chr.13: 72727800-72727800 on Build GRCh38
rs752130949 Chr.13: 72727555-72727566 on Build GRCh38
rs749609502 Chr.13: 72727786-72727786 on Build GRCh38
rs769938486 Chr.13: 72727574-72727590 on Build GRCh38
rs945629801 Chr.13: 72727806-72727806 on Build GRCh38
rs746444240 Chr.13: 72727618-72727618 on Build GRCh38
rs757772883 Chr.13: 72727588-72727588 on Build GRCh38
rs759695356 Chr.13: 72727571-72727571 on Build GRCh38
rs368142617 Chr.13: 72727630-72727630 on Build GRCh38
rs961274638 Chr.13: 72727731-72727731 on Build GRCh38
rs763915489 Chr.13: 72727516-72727516 on Build GRCh38
rs778115494 Chr.13: 72727553-72727553 on Build GRCh38
rs745918229 Chr.13: 72727589-72727589 on Build GRCh38
rs979849817 Chr.13: 72727773-72727773 on Build GRCh38
rs897889372 Chr.13: 72727769-72727769 on Build GRCh38
rs540496880 Chr.13: 72727715-72727715 on Build GRCh38
rs9652239 Chr.13: 72727342-72727342 on Build GRCh38
rs780734450 Chr.13: 72727525-72727525 on Build GRCh38
rs930812986 Chr.13: 72727385-72727385 on Build GRCh38
rs780120678 Chr.13: 72727623-72727623 on Build GRCh38
rs749891691 Chr.13: 72727585-72727585 on Build GRCh38
rs370419285 Chr.13: 72727613-72727613 on Build GRCh38
rs1028792579 Chr.13: 72727696-72727696 on Build GRCh38
rs753685535 Chr.13: 72727828-72727828 on Build GRCh38
rs770241397 Chr.13: 72727738-72727738 on Build GRCh38
rs774962876 Chr.13: 72727565-72727567 on Build GRCh38
rs1015526682 Chr.13: 72727719-72727719 on Build GRCh38
rs749579611 Chr.13: 72727593-72727593 on Build GRCh38
rs759104575 Chr.13: 72727617-72727617 on Build GRCh38
rs536324657 Chr.13: 72727612-72727612 on Build GRCh38
rs938354276 Chr.13: 72727621-72727621 on Build GRCh38
rs569158696 Chr.13: 72727366-72727366 on Build GRCh38
rs776991206 Chr.13: 72727505-72727505 on Build GRCh38
rs765571191 Chr.13: 72727508-72727508 on Build GRCh38
rs142438556 Chr.13: 72727582-72727585 on Build GRCh38
rs116470408 Chr.13: 72727483-72727483 on Build GRCh38
rs774125906 Chr.13: 72727597-72727597 on Build GRCh38
rs1043740815 Chr.13: 72727815-72727815 on Build GRCh38
rs975186535 Chr.13: 72727319-72727319 on Build GRCh38
rs768769963 Chr.13: 72727595-72727595 on Build GRCh38
rs749328128 Chr.13: 72727624-72727624 on Build GRCh38
rs377210022 Chr.13: 72727372-72727372 on Build GRCh38
rs773795636 Chr.13: 72727485-72727485 on Build GRCh38
rs939547195 Chr.13: 72727781-72727781 on Build GRCh38
rs748414458 Chr.13: 72727537-72727537 on Build GRCh38
rs529755573 Chr.13: 72727761-72727761 on Build GRCh38
rs908195089 Chr.13: 72727359-72727359 on Build GRCh38
rs764016435 Chr.13: 72727583-72727583 on Build GRCh38
rs866243564 Chr.13: 72727701-72727701 on Build GRCh38
rs772540987 Chr.13: 72727543-72727543 on Build GRCh38
rs770091272 Chr.13: 72727574-72727574 on Build GRCh38
rs931839814 Chr.13: 72727333-72727333 on Build GRCh38
rs968986652 Chr.13: 72727807-72727807 on Build GRCh38
rs1045316431 Chr.13: 72727702-72727702 on Build GRCh38
rs914351567 Chr.13: 72727609-72727609 on Build GRCh38
rs564249450 Chr.13: 72727665-72727665 on Build GRCh38
rs905479917 Chr.13: 72727434-72727434 on Build GRCh38
rs780273306 Chr.13: 72727550-72727590 on Build GRCh38
rs1465895 Chr.13: 72727666-72727666 on Build GRCh38
rs774455452 Chr.13: 72727643-72727643 on Build GRCh38
rs767396491 Chr.13: 72727598-72727598 on Build GRCh38
rs760701607 Chr.13: 72727779-72727779 on Build GRCh38
rs190351413 Chr.13: 72727311-72727311 on Build GRCh38
rs758733223 Chr.13: 72727546-72727548 on Build GRCh38
rs185837399 Chr.13: 72727782-72727782 on Build GRCh38
rs939649285 Chr.13: 72727365-72727365 on Build GRCh38
rs760651318 Chr.13: 72727495-72727495 on Build GRCh38
rs996879955 Chr.13: 72727679-72727679 on Build GRCh38
rs545767567 Chr.13: 72727638-72727638 on Build GRCh38
rs752620138 Chr.13: 72727714-72727714 on Build GRCh38
rs778259134 Chr.13: 72727587-72727587 on Build GRCh38
rs948447565 Chr.13: 72727603-72727603 on Build GRCh38
rs567141444 Chr.13: 72727374-72727374 on Build GRCh38
rs892682857 Chr.13: 72727648-72727648 on Build GRCh38
rs752499216 Chr.13: 72727514-72727514 on Build GRCh38
rs977236090 Chr.13: 72727811-72727811 on Build GRCh38
rs1465897 Chr.13: 72727517-72727517 on Build GRCh38
rs763892940 Chr.13: 72727783-72727783 on Build GRCh38
rs755693567 Chr.13: 72727536-72727536 on Build GRCh38
rs181034097 Chr.13: 72727567-72727567 on Build GRCh38
rs376939061 Chr.13: 72727520-72727520 on Build GRCh38
rs776796592 Chr.13: 72727568-72727568 on Build GRCh38
rs969675712 Chr.13: 72727763-72727763 on Build GRCh38
rs748546311 Chr.13: 72727479-72727479 on Build GRCh38
rs757744083 Chr.13: 72727615-72727615 on Build GRCh38
rs764804413 Chr.13: 72727610-72727610 on Build GRCh38
rs111320152 Chr.13: 72727416-72727416 on Build GRCh38
rs756248478 Chr.13: 72727622-72727622 on Build GRCh38
rs747619919 Chr.13: 72727561-72727561 on Build GRCh38
rs371680740 Chr.13: 72727582-72727583 on Build GRCh38
rs955238565 Chr.13: 72727698-72727698 on Build GRCh38
rs1465896 Chr.13: 72727590-72727590 on Build GRCh38
rs1022348848 Chr.13: 72727532-72727532 on Build GRCh38
rs761747873 Chr.13: 72727584-72727584 on Build GRCh38
rs896746668 Chr.13: 72727650-72727650 on Build GRCh38
rs568232835 Chr.13: 72727324-72727324 on Build GRCh38
rs1028156723 Chr.13: 72727559-72727559 on Build GRCh38
rs953325793 Chr.13: 72727784-72727784 on Build GRCh38
rs996658783 Chr.13: 72727544-72727544 on Build GRCh38
rs973759587 Chr.13: 72727734-72727734 on Build GRCh38
rs529420401 Chr.13: 72727339-72727339 on Build GRCh38
rs771084236 Chr.13: 72727498-72727498 on Build GRCh38
rs775828017 Chr.13: 72727582-72727582 on Build GRCh38
rs754602697 Chr.13: 72727607-72727607 on Build GRCh38
rs915599477 Chr.13: 72727851-72727851 on Build GRCh38
rs756262716 Chr.13: 72727738-72727739 on Build GRCh38
rs755448899 Chr.13: 72727555-72727569 on Build GRCh38
rs749946780 Chr.13: 72727529-72727529 on Build GRCh38
rs986493367 Chr.13: 72727797-72727797 on Build GRCh38
rs748174456 Chr.13: 72727564-72727565 on Build GRCh38
rs562993019 Chr.13: 72727825-72727825 on Build GRCh38
rs992495386 Chr.13: 72727845-72727845 on Build GRCh38
rs539815214 Chr.13: 72727367-72727367 on Build GRCh38
rs924367827 Chr.13: 72727813-72727813 on Build GRCh38
rs753572040 Chr.13: 72727586-72727586 on Build GRCh38
rs1056765575 Chr.13: 72727433-72727433 on Build GRCh38
rs1009550794 Chr.13: 72727662-72727662 on Build GRCh38
rs933906538 Chr.13: 72727338-72727338 on Build GRCh38
rs376877243 Chr.13: 72727515-72727515 on Build GRCh38
rs534153420 Chr.13: 72727426-72727426 on Build GRCh38
rs1041386472 Chr.13: 72727373-72727373 on Build GRCh38
rs374768667 Chr.13: 72727616-72727616 on Build GRCh38
rs367758166 Chr.13: 72727602-72727602 on Build GRCh38
rs200478166 Chr.13: 72727450-72727450 on Build GRCh38

View Full Product Details