Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 112168645-112169244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41L4A
Entrez Gene ID
64097
Gene Name
erythrocyte membrane protein band 4.1 like 4A
Gene Aliases
EPB41L4, NBL4
UniGene
Hs.584954
Gene Chromosome Location
Chr.5: 112141829-112419490 on Build GRCh38
Transcripts
NM_022140.3 XM_011543530.1 XM_011543531.2 XM_011543532.1 XM_011543533.1 XM_017009689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
735087 c.705G>A p.G235G Chr.5: 112169021-112169021 on Build GRCh38
6314898 c.1886C>A p.T629N Chr.5: 112168785-112168785 on Build GRCh38
3768086 c.709C>T p.R237* Chr.5: 112169017-112169017 on Build GRCh38
1432121 c.1779G>A p.S593S Chr.5: 112169066-112169066 on Build GRCh38
3768085 c.1828C>T p.R610* Chr.5: 112169017-112169017 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768025406 Chr.5: 112168756-112168756 on Build GRCh38
rs776607129 Chr.5: 112168791-112168791 on Build GRCh38
rs376136861 Chr.5: 112168973-112168973 on Build GRCh38
rs768869746 Chr.5: 112168805-112168805 on Build GRCh38
rs770590756 Chr.5: 112168871-112168875 on Build GRCh38
rs541836807 Chr.5: 112169188-112169188 on Build GRCh38
rs750945497 Chr.5: 112168761-112168761 on Build GRCh38
rs999271041 Chr.5: 112169174-112169174 on Build GRCh38
rs1022030946 Chr.5: 112168916-112168916 on Build GRCh38
rs922100221 Chr.5: 112169050-112169050 on Build GRCh38
rs751062021 Chr.5: 112169035-112169035 on Build GRCh38
rs757910803 Chr.5: 112168960-112168960 on Build GRCh38
rs939275291 Chr.5: 112169118-112169118 on Build GRCh38
rs547490755 Chr.5: 112169192-112169192 on Build GRCh38
rs201545233 Chr.5: 112169089-112169089 on Build GRCh38
rs773332244 Chr.5: 112169003-112169003 on Build GRCh38
rs756855321 Chr.5: 112169043-112169043 on Build GRCh38
rs757303695 Chr.5: 112168754-112168754 on Build GRCh38
rs750927508 Chr.5: 112168820-112168820 on Build GRCh38
rs533999192 Chr.5: 112169067-112169067 on Build GRCh38
rs763301958 Chr.5: 112168741-112168741 on Build GRCh38
rs766711566 Chr.5: 112168827-112168827 on Build GRCh38
rs966378312 Chr.5: 112168933-112168933 on Build GRCh38
rs1050399573 Chr.5: 112168676-112168676 on Build GRCh38
rs775841254 Chr.5: 112168740-112168740 on Build GRCh38
rs999219986 Chr.5: 112168952-112168952 on Build GRCh38
rs886443621 Chr.5: 112168724-112168724 on Build GRCh38
rs746910564 Chr.5: 112168966-112168966 on Build GRCh38
rs746617620 Chr.5: 112168856-112168868 on Build GRCh38
rs202038537 Chr.5: 112168979-112168979 on Build GRCh38
rs768050533 Chr.5: 112168702-112168702 on Build GRCh38
rs767005378 Chr.5: 112169142-112169142 on Build GRCh38
rs771013845 Chr.5: 112169066-112169066 on Build GRCh38
rs753933549 Chr.5: 112169154-112169154 on Build GRCh38
rs747025327 Chr.5: 112169063-112169063 on Build GRCh38
rs183686385 Chr.5: 112169033-112169033 on Build GRCh38
rs752536809 Chr.5: 112168986-112168986 on Build GRCh38
rs1045750828 Chr.5: 112169087-112169087 on Build GRCh38
rs761882734 Chr.5: 112168813-112168813 on Build GRCh38
rs778452899 Chr.5: 112168775-112168775 on Build GRCh38
rs752316139 Chr.5: 112168713-112168715 on Build GRCh38
rs200812889 Chr.5: 112169017-112169017 on Build GRCh38
rs766093340 Chr.5: 112168921-112168921 on Build GRCh38
rs1029222162 Chr.5: 112169208-112169208 on Build GRCh38
rs577849317 Chr.5: 112168911-112168911 on Build GRCh38
rs532518113 Chr.5: 112169185-112169185 on Build GRCh38
rs956394961 Chr.5: 112169194-112169194 on Build GRCh38
rs868429825 Chr.5: 112168759-112168759 on Build GRCh38
rs763428611 Chr.5: 112169096-112169096 on Build GRCh38
rs780468584 Chr.5: 112168769-112168769 on Build GRCh38
rs756058838 Chr.5: 112169153-112169153 on Build GRCh38
rs573085210 Chr.5: 112168898-112168898 on Build GRCh38
rs760481013 Chr.5: 112168715-112168715 on Build GRCh38
rs754307593 Chr.5: 112169016-112169016 on Build GRCh38
rs765096922 Chr.5: 112169000-112169000 on Build GRCh38
rs572106322 Chr.5: 112168712-112168712 on Build GRCh38
rs772113191 Chr.5: 112168996-112168996 on Build GRCh38
rs773454868 Chr.5: 112169062-112169062 on Build GRCh38
rs774623206 Chr.5: 112168980-112168980 on Build GRCh38
rs774466034 Chr.5: 112168810-112168810 on Build GRCh38
rs775272553 Chr.5: 112169007-112169007 on Build GRCh38
rs773316110 Chr.5: 112168714-112168714 on Build GRCh38
rs772818883 Chr.5: 112168724-112168727 on Build GRCh38
rs773955133 Chr.5: 112168705-112168705 on Build GRCh38
rs1025759724 Chr.5: 112169099-112169099 on Build GRCh38
rs777490218 Chr.5: 112168962-112168962 on Build GRCh38
rs755935788 Chr.5: 112169052-112169052 on Build GRCh38
rs1023828283 Chr.5: 112169181-112169181 on Build GRCh38
rs752307560 Chr.5: 112168954-112168954 on Build GRCh38
rs373252356 Chr.5: 112169143-112169143 on Build GRCh38
rs368861569 Chr.5: 112168776-112168776 on Build GRCh38
rs748912347 Chr.5: 112168770-112168770 on Build GRCh38
rs7703522 Chr.5: 112168782-112168782 on Build GRCh38
rs952324324 Chr.5: 112169222-112169222 on Build GRCh38
rs774082369 Chr.5: 112168832-112168832 on Build GRCh38
rs763795979 Chr.5: 112169127-112169127 on Build GRCh38
rs375125928 Chr.5: 112168709-112168709 on Build GRCh38
rs776366610 Chr.5: 112168965-112168966 on Build GRCh38
rs946537686 Chr.5: 112169008-112169008 on Build GRCh38
rs200738708 Chr.5: 112169093-112169093 on Build GRCh38
rs17319543 Chr.5: 112168706-112168706 on Build GRCh38
rs781207403 Chr.5: 112169044-112169044 on Build GRCh38
rs188205734 Chr.5: 112169162-112169162 on Build GRCh38
rs766288959 Chr.5: 112168725-112168725 on Build GRCh38
rs762990750 Chr.5: 112169123-112169123 on Build GRCh38
rs376941185 Chr.5: 112168825-112168825 on Build GRCh38
rs756767432 Chr.5: 112169140-112169140 on Build GRCh38
rs371127136 Chr.5: 112168848-112168848 on Build GRCh38
rs749000368 Chr.5: 112168690-112168690 on Build GRCh38
rs759328952 Chr.5: 112168969-112168984 on Build GRCh38
rs1018654084 Chr.5: 112169184-112169184 on Build GRCh38
rs200195011 Chr.5: 112169095-112169095 on Build GRCh38
rs902285251 Chr.5: 112169165-112169165 on Build GRCh38
rs766832104 Chr.5: 112169015-112169015 on Build GRCh38
rs751258543 Chr.5: 112169129-112169129 on Build GRCh38
rs762010062 Chr.5: 112168991-112168991 on Build GRCh38
rs749864627 Chr.5: 112168721-112168722 on Build GRCh38
rs771478027 Chr.5: 112168723-112168724 on Build GRCh38
rs772237814 Chr.5: 112169061-112169061 on Build GRCh38
rs756624587 Chr.5: 112168694-112168695 on Build GRCh38
rs764062685 Chr.5: 112169131-112169140 on Build GRCh38
rs370174086 Chr.5: 112169047-112169047 on Build GRCh38
rs530073538 Chr.5: 112169183-112169183 on Build GRCh38
rs114470084 Chr.5: 112168914-112168914 on Build GRCh38
rs180931108 Chr.5: 112168747-112168747 on Build GRCh38
rs767645587 Chr.5: 112168816-112168816 on Build GRCh38
rs766163075 Chr.5: 112168783-112168783 on Build GRCh38
rs544419945 Chr.5: 112168790-112168790 on Build GRCh38
rs17266567 Chr.5: 112169085-112169085 on Build GRCh38
rs373074246 Chr.5: 112168856-112168856 on Build GRCh38
rs985297740 Chr.5: 112169076-112169076 on Build GRCh38
rs745633414 Chr.5: 112168977-112168977 on Build GRCh38
rs748384550 Chr.5: 112169059-112169059 on Build GRCh38
rs996834305 Chr.5: 112169189-112169189 on Build GRCh38
rs1019593421 Chr.5: 112168732-112168732 on Build GRCh38
rs368470046 Chr.5: 112169086-112169086 on Build GRCh38
rs756460326 Chr.5: 112168765-112168765 on Build GRCh38
rs1010712614 Chr.5: 112169158-112169158 on Build GRCh38
rs189409879 Chr.5: 112168746-112168746 on Build GRCh38
rs779756431 Chr.5: 112169053-112169053 on Build GRCh38
rs879217234 Chr.5: 112168748-112168748 on Build GRCh38
rs777398758 Chr.5: 112169034-112169034 on Build GRCh38
rs754709341 Chr.5: 112168771-112168771 on Build GRCh38
rs143781799 Chr.5: 112168828-112168828 on Build GRCh38
rs771037674 Chr.5: 112168971-112168971 on Build GRCh38

View Full Product Details