Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157345251-157345806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYFIP2
Entrez Gene ID
26999
Gene Name
cytoplasmic FMR1 interacting protein 2
Gene Aliases
PIR121
UniGene
Hs.519702
Gene Chromosome Location
Chr.5: 157266080-157395598 on Build GRCh38
Transcripts
NM_001037333.2 NM_001291721.1 NM_001291722.1 NM_014376.3 XM_011534516.2 XM_017009341.1 XM_017009342.1 NM_001001343.3

Gene Symbol
FNDC9
Entrez Gene ID
408263
Gene Name
fibronectin type III domain containing 9
Gene Aliases
C5orf40
UniGene
Hs.437066
Gene Chromosome Location
Chr.5: 157341599-157345721 on Build GRCh38
Transcripts
NM_001037333.2 NM_001291721.1 NM_001291722.1 NM_014376.3 XM_011534516.2 XM_017009341.1 XM_017009342.1 NM_001001343.3

SNP Detail

SNP ID Chromosome Location
rs969871068 Chr.5: 157345443-157345443 on Build GRCh38
rs559873509 Chr.5: 157345700-157345700 on Build GRCh38
rs745631490 Chr.5: 157345474-157345474 on Build GRCh38
rs953185040 Chr.5: 157345470-157345470 on Build GRCh38
rs576716311 Chr.5: 157345431-157345431 on Build GRCh38
rs911467663 Chr.5: 157345710-157345710 on Build GRCh38
rs574190069 Chr.5: 157345532-157345532 on Build GRCh38
rs960208799 Chr.5: 157345407-157345407 on Build GRCh38
rs1032871045 Chr.5: 157345649-157345650 on Build GRCh38
rs997329935 Chr.5: 157345444-157345444 on Build GRCh38
rs1043667443 Chr.5: 157345365-157345365 on Build GRCh38
rs960092111 Chr.5: 157345603-157345603 on Build GRCh38
rs541583328 Chr.5: 157345749-157345749 on Build GRCh38
rs535255914 Chr.5: 157345289-157345289 on Build GRCh38
rs373093918 Chr.5: 157345779-157345779 on Build GRCh38
rs139381072 Chr.5: 157345316-157345316 on Build GRCh38
rs1016554759 Chr.5: 157345489-157345489 on Build GRCh38
rs1030786631 Chr.5: 157345404-157345404 on Build GRCh38
rs999974521 Chr.5: 157345642-157345642 on Build GRCh38
rs1035711651 Chr.5: 157345773-157345773 on Build GRCh38
rs530766492 Chr.5: 157345750-157345750 on Build GRCh38
rs543850220 Chr.5: 157345624-157345624 on Build GRCh38
rs991547059 Chr.5: 157345608-157345608 on Build GRCh38
rs769871749 Chr.5: 157345579-157345579 on Build GRCh38
rs903522092 Chr.5: 157345391-157345391 on Build GRCh38
rs988866748 Chr.5: 157345472-157345472 on Build GRCh38
rs542822551 Chr.5: 157345677-157345677 on Build GRCh38
rs947677061 Chr.5: 157345364-157345364 on Build GRCh38
rs994567127 Chr.5: 157345354-157345354 on Build GRCh38
rs113461082 Chr.5: 157345445-157345445 on Build GRCh38
rs938864693 Chr.5: 157345274-157345274 on Build GRCh38
rs1038581521 Chr.5: 157345550-157345550 on Build GRCh38
rs545088564 Chr.5: 157345742-157345742 on Build GRCh38
rs557327627 Chr.5: 157345530-157345530 on Build GRCh38
rs1050014501 Chr.5: 157345480-157345480 on Build GRCh38
rs768345980 Chr.5: 157345703-157345703 on Build GRCh38
rs184068269 Chr.5: 157345375-157345375 on Build GRCh38
rs557822459 Chr.5: 157345376-157345376 on Build GRCh38
rs1050100399 Chr.5: 157345716-157345716 on Build GRCh38
rs562129143 Chr.5: 157345678-157345678 on Build GRCh38
rs1014090815 Chr.5: 157345612-157345612 on Build GRCh38
rs994286819 Chr.5: 157345556-157345556 on Build GRCh38
rs138483046 Chr.5: 157345729-157345729 on Build GRCh38
rs947503325 Chr.5: 157345637-157345637 on Build GRCh38
rs911354854 Chr.5: 157345519-157345519 on Build GRCh38
rs972465081 Chr.5: 157345566-157345566 on Build GRCh38
rs143340246 Chr.5: 157345363-157345363 on Build GRCh38
rs373356863 Chr.5: 157345475-157345475 on Build GRCh38
rs1038466429 Chr.5: 157345302-157345302 on Build GRCh38
rs941505970 Chr.5: 157345525-157345525 on Build GRCh38
rs1022722663 Chr.5: 157345437-157345437 on Build GRCh38
rs962107029 Chr.5: 157345555-157345555 on Build GRCh38
rs1055959849 Chr.5: 157345297-157345297 on Build GRCh38
rs974219926 Chr.5: 157345741-157345741 on Build GRCh38
rs1005606070 Chr.5: 157345285-157345285 on Build GRCh38
rs1043354887 Chr.5: 157345659-157345659 on Build GRCh38
rs955960068 Chr.5: 157345662-157345662 on Build GRCh38
rs188269777 Chr.5: 157345754-157345754 on Build GRCh38
rs762950837 Chr.5: 157345699-157345699 on Build GRCh38
rs1056559741 Chr.5: 157345753-157345753 on Build GRCh38
rs771131176 Chr.5: 157345398-157345398 on Build GRCh38
rs925698370 Chr.5: 157345459-157345459 on Build GRCh38

View Full Product Details