Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44172982-44173547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN11
Entrez Gene ID
11131
Gene Name
calpain 11
Gene Aliases
calpain11
UniGene
Hs.225953
Gene Chromosome Location
Chr.6: 44158811-44184402 on Build GRCh38
Transcripts
NM_007058.3 XM_006714984.1 XM_006714985.1 XM_006714987.1 XM_011514274.1 XM_011514275.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4153430 c.662+6T>G p.? Chr.6: 44173079-44173079 on Build GRCh38
150098 c.662+8C>G p.? Chr.6: 44173081-44173081 on Build GRCh38
7359416 c.619C>T p.R207C Chr.6: 44173030-44173030 on Build GRCh38
5698910 c.740A>T p.Q247L Chr.6: 44173295-44173295 on Build GRCh38
3627776 c.819T>C p.G273G Chr.6: 44173374-44173374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs933872671 Chr.6: 44173183-44173183 on Build GRCh38
rs767123649 Chr.6: 44173054-44173054 on Build GRCh38
rs1056021429 Chr.6: 44173327-44173327 on Build GRCh38
rs780323994 Chr.6: 44173412-44173412 on Build GRCh38
rs763499312 Chr.6: 44173366-44173366 on Build GRCh38
rs202150569 Chr.6: 44173264-44173264 on Build GRCh38
rs553982188 Chr.6: 44173390-44173390 on Build GRCh38
rs374999083 Chr.6: 44173013-44173013 on Build GRCh38
rs745954774 Chr.6: 44173169-44173169 on Build GRCh38
rs769594113 Chr.6: 44173020-44173020 on Build GRCh38
rs979545700 Chr.6: 44173118-44173118 on Build GRCh38
rs767682793 Chr.6: 44173298-44173298 on Build GRCh38
rs750672835 Chr.6: 44173392-44173392 on Build GRCh38
rs976095333 Chr.6: 44173477-44173477 on Build GRCh38
rs767861802 Chr.6: 44173391-44173391 on Build GRCh38
rs765871453 Chr.6: 44173358-44173358 on Build GRCh38
rs765794525 Chr.6: 44173245-44173245 on Build GRCh38
rs370572396 Chr.6: 44173382-44173382 on Build GRCh38
rs756359148 Chr.6: 44173397-44173397 on Build GRCh38
rs775445812 Chr.6: 44173026-44173026 on Build GRCh38
rs1009525648 Chr.6: 44173308-44173308 on Build GRCh38
rs779026378 Chr.6: 44173421-44173421 on Build GRCh38
rs750325025 Chr.6: 44173371-44173371 on Build GRCh38
rs371227294 Chr.6: 44173200-44173200 on Build GRCh38
rs774285384 Chr.6: 44173222-44173222 on Build GRCh38
rs751274577 Chr.6: 44173038-44173038 on Build GRCh38
rs1015013538 Chr.6: 44173439-44173439 on Build GRCh38
rs755061307 Chr.6: 44173418-44173418 on Build GRCh38
rs1038491080 Chr.6: 44173227-44173227 on Build GRCh38
rs768635315 Chr.6: 44173219-44173219 on Build GRCh38
rs757589679 Chr.6: 44173290-44173290 on Build GRCh38
rs918133682 Chr.6: 44173161-44173161 on Build GRCh38
rs764480583 Chr.6: 44173287-44173287 on Build GRCh38
rs776087132 Chr.6: 44173362-44173362 on Build GRCh38
rs781298511 Chr.6: 44173295-44173295 on Build GRCh38
rs992355441 Chr.6: 44173159-44173159 on Build GRCh38
rs747297049 Chr.6: 44173087-44173087 on Build GRCh38
rs574617476 Chr.6: 44173371-44173371 on Build GRCh38
rs758902018 Chr.6: 44173265-44173265 on Build GRCh38
rs929553658 Chr.6: 44173523-44173523 on Build GRCh38
rs760134227 Chr.6: 44173262-44173262 on Build GRCh38
rs985261959 Chr.6: 44173046-44173046 on Build GRCh38
rs771862820 Chr.6: 44173339-44173339 on Build GRCh38
rs538226958 Chr.6: 44173338-44173338 on Build GRCh38
rs780172016 Chr.6: 44173317-44173317 on Build GRCh38
rs968005493 Chr.6: 44173357-44173357 on Build GRCh38
rs768686523 Chr.6: 44173321-44173321 on Build GRCh38
rs929587941 Chr.6: 44173169-44173169 on Build GRCh38
rs767056484 Chr.6: 44173248-44173248 on Build GRCh38
rs962088350 Chr.6: 44173468-44173468 on Build GRCh38
rs373586466 Chr.6: 44173251-44173251 on Build GRCh38
rs375545266 Chr.6: 44173288-44173288 on Build GRCh38
rs6938938 Chr.6: 44173351-44173351 on Build GRCh38
rs1043448987 Chr.6: 44173313-44173313 on Build GRCh38
rs761601477 Chr.6: 44173224-44173224 on Build GRCh38
rs552423781 Chr.6: 44173135-44173135 on Build GRCh38
rs749873430 Chr.6: 44173060-44173060 on Build GRCh38
rs749360047 Chr.6: 44173413-44173413 on Build GRCh38
rs941041482 Chr.6: 44173226-44173226 on Build GRCh38
rs748051424 Chr.6: 44173425-44173425 on Build GRCh38
rs3813500 Chr.6: 44173081-44173081 on Build GRCh38
rs758767166 Chr.6: 44173082-44173082 on Build GRCh38
rs372617735 Chr.6: 44173031-44173031 on Build GRCh38
rs1023828116 Chr.6: 44173404-44173404 on Build GRCh38
rs1029970071 Chr.6: 44173027-44173027 on Build GRCh38
rs938136070 Chr.6: 44173142-44173142 on Build GRCh38
rs376868200 Chr.6: 44173240-44173240 on Build GRCh38
rs572284322 Chr.6: 44173459-44173459 on Build GRCh38
rs546227632 Chr.6: 44173481-44173481 on Build GRCh38
rs200279946 Chr.6: 44173030-44173030 on Build GRCh38
rs771109804 Chr.6: 44173092-44173092 on Build GRCh38
rs764605029 Chr.6: 44173370-44173370 on Build GRCh38
rs769874215 Chr.6: 44173177-44173177 on Build GRCh38
rs746031431 Chr.6: 44173314-44173314 on Build GRCh38
rs777627112 Chr.6: 44173447-44173447 on Build GRCh38
rs959495883 Chr.6: 44173386-44173386 on Build GRCh38
rs779311925 Chr.6: 44173065-44173065 on Build GRCh38
rs749183714 Chr.6: 44173212-44173212 on Build GRCh38
rs565761856 Chr.6: 44173350-44173350 on Build GRCh38
rs566636586 Chr.6: 44173084-44173084 on Build GRCh38
rs534113991 Chr.6: 44173097-44173097 on Build GRCh38
rs776567421 Chr.6: 44173009-44173009 on Build GRCh38
rs761548409 Chr.6: 44173051-44173051 on Build GRCh38
rs886811693 Chr.6: 44173206-44173206 on Build GRCh38
rs373882901 Chr.6: 44173215-44173215 on Build GRCh38
rs756157496 Chr.6: 44173315-44173315 on Build GRCh38
rs752187272 Chr.6: 44173334-44173334 on Build GRCh38
rs755559441 Chr.6: 44173064-44173064 on Build GRCh38

View Full Product Details