Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 136244622-136245187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTFR2
Entrez Gene ID
113115
Gene Name
mitochondrial fission regulator 2
Gene Aliases
DUFD1, FAM54A
UniGene
Hs.121536
Gene Chromosome Location
Chr.6: 136231030-136253659 on Build GRCh38
Transcripts
NM_001099286.2 NM_001318738.1 NM_138419.4 XM_011535410.2 XM_011535412.2 XM_011535413.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1582666 c.152G>A p.R51Q Chr.6: 136244781-136244781 on Build GRCh38
396170 c.118A>G p.I40V Chr.6: 136244815-136244815 on Build GRCh38
296167 c.116G>A p.R39H Chr.6: 136244817-136244817 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781308861 Chr.6: 136244775-136244775 on Build GRCh38
rs1023032701 Chr.6: 136244956-136244964 on Build GRCh38
rs999094234 Chr.6: 136244949-136244949 on Build GRCh38
rs556099538 Chr.6: 136244667-136244667 on Build GRCh38
rs541755696 Chr.6: 136244850-136244850 on Build GRCh38
rs767170621 Chr.6: 136244822-136244822 on Build GRCh38
rs752346198 Chr.6: 136244833-136244833 on Build GRCh38
rs946784771 Chr.6: 136244987-136244987 on Build GRCh38
rs981958268 Chr.6: 136245112-136245112 on Build GRCh38
rs1043735793 Chr.6: 136245029-136245029 on Build GRCh38
rs1047590320 Chr.6: 136245095-136245095 on Build GRCh38
rs988261838 Chr.6: 136244973-136244973 on Build GRCh38
rs60351480 Chr.6: 136244946-136244954 on Build GRCh38
rs1022650245 Chr.6: 136245158-136245158 on Build GRCh38
rs149359911 Chr.6: 136244806-136244806 on Build GRCh38
rs927592148 Chr.6: 136244860-136244860 on Build GRCh38
rs760132374 Chr.6: 136244842-136244842 on Build GRCh38
rs778014506 Chr.6: 136244913-136244913 on Build GRCh38
rs748348654 Chr.6: 136244895-136244895 on Build GRCh38
rs779372694 Chr.6: 136244723-136244723 on Build GRCh38
rs927917556 Chr.6: 136245135-136245135 on Build GRCh38
rs780407583 Chr.6: 136244769-136244769 on Build GRCh38
rs887548313 Chr.6: 136245132-136245132 on Build GRCh38
rs781604137 Chr.6: 136244687-136244687 on Build GRCh38
rs751375608 Chr.6: 136244804-136244804 on Build GRCh38
rs116661610 Chr.6: 136245090-136245090 on Build GRCh38
rs996885000 Chr.6: 136244991-136244991 on Build GRCh38
rs748237317 Chr.6: 136244776-136244776 on Build GRCh38
rs781424348 Chr.6: 136244889-136244889 on Build GRCh38
rs773242328 Chr.6: 136244794-136244794 on Build GRCh38
rs755346945 Chr.6: 136244886-136244886 on Build GRCh38
rs780488363 Chr.6: 136244858-136244858 on Build GRCh38
rs770794375 Chr.6: 136244813-136244813 on Build GRCh38
rs878950062 Chr.6: 136244964-136244964 on Build GRCh38
rs965390242 Chr.6: 136244710-136244710 on Build GRCh38
rs779056120 Chr.6: 136244963-136244963 on Build GRCh38
rs192921741 Chr.6: 136244643-136244643 on Build GRCh38
rs200368436 Chr.6: 136244817-136244817 on Build GRCh38
rs548108727 Chr.6: 136244973-136244973 on Build GRCh38
rs149212594 Chr.6: 136244911-136244911 on Build GRCh38
rs770988675 Chr.6: 136244915-136244915 on Build GRCh38
rs772803551 Chr.6: 136244739-136244739 on Build GRCh38
rs200860543 Chr.6: 136244782-136244782 on Build GRCh38
rs937683832 Chr.6: 136244875-136244875 on Build GRCh38
rs542729038 Chr.6: 136245108-136245108 on Build GRCh38
rs981410123 Chr.6: 136244967-136244967 on Build GRCh38
rs879606314 Chr.6: 136244955-136244958 on Build GRCh38
rs143698159 Chr.6: 136244818-136244818 on Build GRCh38
rs1028411145 Chr.6: 136245004-136245004 on Build GRCh38
rs914027716 Chr.6: 136244982-136244982 on Build GRCh38
rs184503104 Chr.6: 136244923-136244923 on Build GRCh38
rs370498597 Chr.6: 136244771-136244771 on Build GRCh38
rs118181363 Chr.6: 136244751-136244751 on Build GRCh38
rs373810665 Chr.6: 136245065-136245065 on Build GRCh38
rs763595214 Chr.6: 136244847-136244847 on Build GRCh38
rs774989921 Chr.6: 136244916-136244916 on Build GRCh38
rs892475358 Chr.6: 136244699-136244699 on Build GRCh38
rs187340365 Chr.6: 136245041-136245041 on Build GRCh38
rs928158537 Chr.6: 136244972-136244973 on Build GRCh38
rs747728989 Chr.6: 136244761-136244761 on Build GRCh38
rs377742665 Chr.6: 136244914-136244914 on Build GRCh38
rs952853317 Chr.6: 136245007-136245007 on Build GRCh38
rs751920207 Chr.6: 136244878-136244878 on Build GRCh38
rs1043257994 Chr.6: 136244929-136244929 on Build GRCh38
rs757347228 Chr.6: 136244941-136244941 on Build GRCh38
rs202244416 Chr.6: 136244781-136244781 on Build GRCh38
rs778661731 Chr.6: 136244722-136244723 on Build GRCh38
rs559528448 Chr.6: 136245140-136245140 on Build GRCh38
rs1040655091 Chr.6: 136245072-136245072 on Build GRCh38
rs757979960 Chr.6: 136244884-136244887 on Build GRCh38
rs796794866 Chr.6: 136244955-136244963 on Build GRCh38
rs993363618 Chr.6: 136245092-136245092 on Build GRCh38
rs768914702 Chr.6: 136244772-136244772 on Build GRCh38
rs903362382 Chr.6: 136244943-136244943 on Build GRCh38
rs745628260 Chr.6: 136244857-136244857 on Build GRCh38

View Full Product Details