Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 22944187-22944782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM126A
Entrez Gene ID
84668
Gene Name
family with sequence similarity 126 member A
Gene Aliases
DRCTNNB1A, HCC, HLD5, HYCC1
UniGene
Hs.85603
Gene Chromosome Location
Chr.7: 22895848-23014195 on Build GRCh38
Transcripts
NM_032581.3 XM_005249894.3 XM_005249895.2 XM_011515589.2 XM_011515590.1

SNP Detail

SNP ID Chromosome Location
rs533607208 Chr.7: 22944216-22944216 on Build GRCh38
rs932136565 Chr.7: 22944608-22944608 on Build GRCh38
rs944723434 Chr.7: 22944380-22944380 on Build GRCh38
rs539038545 Chr.7: 22944365-22944365 on Build GRCh38
rs75758928 Chr.7: 22944220-22944220 on Build GRCh38
rs572504055 Chr.7: 22944422-22944422 on Build GRCh38
rs1003276201 Chr.7: 22944313-22944313 on Build GRCh38
rs946255288 Chr.7: 22944646-22944646 on Build GRCh38
rs926652139 Chr.7: 22944573-22944573 on Build GRCh38
rs922046757 Chr.7: 22944582-22944582 on Build GRCh38
rs914755562 Chr.7: 22944642-22944642 on Build GRCh38
rs1027448483 Chr.7: 22944452-22944452 on Build GRCh38
rs560427520 Chr.7: 22944511-22944511 on Build GRCh38
rs1011959750 Chr.7: 22944732-22944732 on Build GRCh38
rs115040380 Chr.7: 22944731-22944731 on Build GRCh38
rs541532700 Chr.7: 22944457-22944457 on Build GRCh38
rs760590320 Chr.7: 22944533-22944533 on Build GRCh38
rs562819927 Chr.7: 22944653-22944653 on Build GRCh38
rs35399557 Chr.7: 22944281-22944282 on Build GRCh38
rs1040786419 Chr.7: 22944393-22944393 on Build GRCh38
rs1013839547 Chr.7: 22944688-22944688 on Build GRCh38
rs774463731 Chr.7: 22944231-22944231 on Build GRCh38
rs886062205 Chr.7: 22944334-22944334 on Build GRCh38
rs979919381 Chr.7: 22944297-22944297 on Build GRCh38
rs957432079 Chr.7: 22944325-22944325 on Build GRCh38
rs1042024215 Chr.7: 22944293-22944293 on Build GRCh38
rs573861679 Chr.7: 22944530-22944530 on Build GRCh38
rs893351750 Chr.7: 22944242-22944242 on Build GRCh38
rs750462825 Chr.7: 22944290-22944290 on Build GRCh38
rs990281165 Chr.7: 22944639-22944639 on Build GRCh38
rs902193274 Chr.7: 22944312-22944312 on Build GRCh38
rs902163307 Chr.7: 22944690-22944690 on Build GRCh38
rs1012074112 Chr.7: 22944449-22944449 on Build GRCh38
rs925794098 Chr.7: 22944323-22944323 on Build GRCh38
rs1027417419 Chr.7: 22944743-22944743 on Build GRCh38
rs4599713 Chr.7: 22944235-22944235 on Build GRCh38
rs970384963 Chr.7: 22944289-22944289 on Build GRCh38
rs373595826 Chr.7: 22944279-22944279 on Build GRCh38
rs558637821 Chr.7: 22944421-22944421 on Build GRCh38
rs376876909 Chr.7: 22944348-22944348 on Build GRCh38
rs1014903375 Chr.7: 22944512-22944512 on Build GRCh38
rs939049169 Chr.7: 22944697-22944697 on Build GRCh38
rs988919575 Chr.7: 22944346-22944346 on Build GRCh38
rs189033320 Chr.7: 22944717-22944717 on Build GRCh38
rs983786708 Chr.7: 22944482-22944482 on Build GRCh38
rs1040878358 Chr.7: 22944212-22944212 on Build GRCh38
rs886062206 Chr.7: 22944582-22944582 on Build GRCh38
rs969973542 Chr.7: 22944734-22944734 on Build GRCh38
rs548120755 Chr.7: 22944234-22944237 on Build GRCh38
rs773073817 Chr.7: 22944417-22944417 on Build GRCh38
rs878944502 Chr.7: 22944335-22944335 on Build GRCh38
rs1583511 Chr.7: 22944271-22944271 on Build GRCh38
rs185370892 Chr.7: 22944581-22944581 on Build GRCh38
rs756232690 Chr.7: 22944374-22944374 on Build GRCh38
rs556418634 Chr.7: 22944281-22944281 on Build GRCh38
rs576633163 Chr.7: 22944341-22944341 on Build GRCh38

View Full Product Details