Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 36153957-36154556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEPD1
Entrez Gene ID
80820
Gene Name
endonuclease/exonuclease/phosphatase family domain containing 1
Gene Aliases
HSPC107
UniGene
Hs.487994
Gene Chromosome Location
Chr.7: 36153149-36301543 on Build GRCh38
Transcripts
NM_030636.2 XM_017012657.1 XM_017012658.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5872655 c.181G>A p.A61T Chr.7: 36154505-36154505 on Build GRCh38
6651826 c.177C>T p.A59A Chr.7: 36154501-36154501 on Build GRCh38
1089365 c.76G>A p.A26T Chr.7: 36154400-36154400 on Build GRCh38
3638413 c.209A>G p.Y70C Chr.7: 36154533-36154533 on Build GRCh38
3832719 c.214G>A p.G72S Chr.7: 36154538-36154538 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923282719 Chr.7: 36154036-36154036 on Build GRCh38
rs750527875 Chr.7: 36154508-36154508 on Build GRCh38
rs765472461 Chr.7: 36154286-36154286 on Build GRCh38
rs888364512 Chr.7: 36154161-36154161 on Build GRCh38
rs965140953 Chr.7: 36153987-36153987 on Build GRCh38
rs754346389 Chr.7: 36154279-36154279 on Build GRCh38
rs780104209 Chr.7: 36154516-36154516 on Build GRCh38
rs765781241 Chr.7: 36154447-36154447 on Build GRCh38
rs148564104 Chr.7: 36154124-36154124 on Build GRCh38
rs1039583006 Chr.7: 36154456-36154456 on Build GRCh38
rs566376048 Chr.7: 36154346-36154346 on Build GRCh38
rs377640807 Chr.7: 36154294-36154294 on Build GRCh38
rs142899131 Chr.7: 36154209-36154209 on Build GRCh38
rs755936310 Chr.7: 36154512-36154512 on Build GRCh38
rs747815515 Chr.7: 36154502-36154502 on Build GRCh38
rs560702787 Chr.7: 36154169-36154169 on Build GRCh38
rs914818048 Chr.7: 36154071-36154071 on Build GRCh38
rs757530322 Chr.7: 36154493-36154493 on Build GRCh38
rs921068792 Chr.7: 36154270-36154270 on Build GRCh38
rs776268630 Chr.7: 36154501-36154501 on Build GRCh38
rs918334592 Chr.7: 36154413-36154413 on Build GRCh38
rs929898417 Chr.7: 36154000-36154000 on Build GRCh38
rs759905183 Chr.7: 36154446-36154446 on Build GRCh38
rs1023113307 Chr.7: 36154222-36154222 on Build GRCh38
rs947125224 Chr.7: 36154084-36154084 on Build GRCh38
rs77967583 Chr.7: 36154109-36154109 on Build GRCh38
rs950664126 Chr.7: 36154249-36154249 on Build GRCh38
rs774200779 Chr.7: 36154395-36154395 on Build GRCh38
rs193120466 Chr.7: 36154178-36154178 on Build GRCh38
rs568340198 Chr.7: 36154261-36154261 on Build GRCh38
rs749074774 Chr.7: 36154522-36154522 on Build GRCh38
rs117198737 Chr.7: 36154228-36154228 on Build GRCh38
rs766892541 Chr.7: 36154276-36154276 on Build GRCh38
rs976162101 Chr.7: 36154013-36154013 on Build GRCh38
rs757377337 Chr.7: 36154311-36154311 on Build GRCh38
rs147463504 Chr.7: 36154481-36154481 on Build GRCh38
rs531619826 Chr.7: 36154224-36154224 on Build GRCh38
rs868548278 Chr.7: 36154227-36154227 on Build GRCh38
rs975681940 Chr.7: 36154265-36154265 on Build GRCh38
rs947595450 Chr.7: 36154074-36154074 on Build GRCh38
rs768417088 Chr.7: 36154383-36154383 on Build GRCh38
rs560835948 Chr.7: 36154103-36154103 on Build GRCh38
rs143269948 Chr.7: 36154419-36154419 on Build GRCh38
rs1044938706 Chr.7: 36154086-36154086 on Build GRCh38
rs772804886 Chr.7: 36154435-36154435 on Build GRCh38
rs199681831 Chr.7: 36154303-36154303 on Build GRCh38
rs899617236 Chr.7: 36154077-36154077 on Build GRCh38
rs952386192 Chr.7: 36154212-36154212 on Build GRCh38
rs758551292 Chr.7: 36154295-36154295 on Build GRCh38
rs989027967 Chr.7: 36154070-36154070 on Build GRCh38
rs927473298 Chr.7: 36154336-36154336 on Build GRCh38
rs983066896 Chr.7: 36154001-36154001 on Build GRCh38
rs993799887 Chr.7: 36154134-36154134 on Build GRCh38
rs371723136 Chr.7: 36154382-36154382 on Build GRCh38
rs912509709 Chr.7: 36154257-36154257 on Build GRCh38
rs759855586 Chr.7: 36154201-36154201 on Build GRCh38
rs778039221 Chr.7: 36154296-36154296 on Build GRCh38
rs1021855036 Chr.7: 36154264-36154264 on Build GRCh38
rs185539735 Chr.7: 36154191-36154191 on Build GRCh38
rs764278451 Chr.7: 36154464-36154464 on Build GRCh38
rs1056861254 Chr.7: 36154110-36154110 on Build GRCh38
rs756097652 Chr.7: 36154350-36154350 on Build GRCh38
rs866579069 Chr.7: 36154251-36154251 on Build GRCh38
rs763368922 Chr.7: 36154461-36154461 on Build GRCh38
rs781152945 Chr.7: 36154315-36154315 on Build GRCh38
rs531409772 Chr.7: 36154100-36154100 on Build GRCh38
rs1006750492 Chr.7: 36154174-36154174 on Build GRCh38
rs1048074739 Chr.7: 36154429-36154429 on Build GRCh38
rs745639259 Chr.7: 36154137-36154137 on Build GRCh38
rs747635934 Chr.7: 36154404-36154404 on Build GRCh38
rs754866005 Chr.7: 36154531-36154531 on Build GRCh38
rs955998353 Chr.7: 36154066-36154066 on Build GRCh38
rs745753282 Chr.7: 36154341-36154341 on Build GRCh38
rs367993321 Chr.7: 36154361-36154361 on Build GRCh38
rs779654385 Chr.7: 36154363-36154363 on Build GRCh38
rs919232867 Chr.7: 36153992-36153992 on Build GRCh38
rs896078357 Chr.7: 36154389-36154389 on Build GRCh38
rs201785924 Chr.7: 36154454-36154454 on Build GRCh38
rs200307132 Chr.7: 36154285-36154285 on Build GRCh38

View Full Product Details