Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 103020830-103021386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1C1
Entrez Gene ID
528
Gene Name
ATPase H+ transporting V1 subunit C1
Gene Aliases
ATP6C, ATP6D, VATC, Vma5
UniGene
Hs.86905
Gene Chromosome Location
Chr.8: 103021020-103073057 on Build GRCh38
Transcripts
NM_001695.4

SNP Detail

SNP ID Chromosome Location
rs1031970687 Chr.8: 103021085-103021085 on Build GRCh38
rs753550827 Chr.8: 103021119-103021119 on Build GRCh38
rs1004991234 Chr.8: 103021029-103021029 on Build GRCh38
rs931710454 Chr.8: 103021042-103021042 on Build GRCh38
rs553439661 Chr.8: 103021344-103021344 on Build GRCh38
rs931223413 Chr.8: 103021167-103021167 on Build GRCh38
rs890636423 Chr.8: 103020869-103020869 on Build GRCh38
rs750717891 Chr.8: 103020998-103020998 on Build GRCh38
rs1036470483 Chr.8: 103021228-103021228 on Build GRCh38
rs894491519 Chr.8: 103021089-103021089 on Build GRCh38
rs542597724 Chr.8: 103021264-103021264 on Build GRCh38
rs981089949 Chr.8: 103021154-103021154 on Build GRCh38
rs1015098059 Chr.8: 103020903-103020903 on Build GRCh38
rs751768589 Chr.8: 103021065-103021065 on Build GRCh38
rs530186283 Chr.8: 103021056-103021056 on Build GRCh38
rs949297305 Chr.8: 103020978-103020978 on Build GRCh38
rs762165368 Chr.8: 103020943-103020943 on Build GRCh38
rs539926851 Chr.8: 103021316-103021316 on Build GRCh38
rs575505061 Chr.8: 103021250-103021250 on Build GRCh38
rs917729305 Chr.8: 103021147-103021147 on Build GRCh38
rs754663265 Chr.8: 103021121-103021121 on Build GRCh38
rs1033532941 Chr.8: 103021349-103021349 on Build GRCh38
rs927277797 Chr.8: 103020992-103020992 on Build GRCh38
rs889326792 Chr.8: 103021272-103021272 on Build GRCh38
rs907784517 Chr.8: 103020989-103020989 on Build GRCh38
rs771701994 Chr.8: 103020908-103020908 on Build GRCh38
rs1051467076 Chr.8: 103021051-103021051 on Build GRCh38
rs887825612 Chr.8: 103021028-103021028 on Build GRCh38
rs868685088 Chr.8: 103021207-103021207 on Build GRCh38
rs970891303 Chr.8: 103021327-103021327 on Build GRCh38
rs550960928 Chr.8: 103020950-103020950 on Build GRCh38
rs567107387 Chr.8: 103021073-103021073 on Build GRCh38
rs755660606 Chr.8: 103021020-103021020 on Build GRCh38
rs539986381 Chr.8: 103020907-103020907 on Build GRCh38
rs925065970 Chr.8: 103020955-103020955 on Build GRCh38
rs564657848 Chr.8: 103021322-103021322 on Build GRCh38
rs2454042 Chr.8: 103021277-103021277 on Build GRCh38
rs909227379 Chr.8: 103021201-103021201 on Build GRCh38
rs921566992 Chr.8: 103021185-103021185 on Build GRCh38
rs1034377584 Chr.8: 103020928-103020928 on Build GRCh38
rs534281783 Chr.8: 103021123-103021123 on Build GRCh38
rs78783640 Chr.8: 103020875-103020875 on Build GRCh38
rs548771178 Chr.8: 103021058-103021058 on Build GRCh38
rs1000451785 Chr.8: 103021084-103021084 on Build GRCh38
rs569558025 Chr.8: 103020980-103020980 on Build GRCh38
rs557566935 Chr.8: 103020941-103020941 on Build GRCh38
rs1005022364 Chr.8: 103020882-103020882 on Build GRCh38
rs1009729743 Chr.8: 103021297-103021297 on Build GRCh38
rs904810881 Chr.8: 103020891-103020891 on Build GRCh38
rs963307998 Chr.8: 103021157-103021157 on Build GRCh38
rs953071148 Chr.8: 103020948-103020948 on Build GRCh38
rs997726481 Chr.8: 103021260-103021260 on Build GRCh38
rs190278495 Chr.8: 103021139-103021139 on Build GRCh38
rs973081302 Chr.8: 103020993-103020993 on Build GRCh38
rs937360045 Chr.8: 103020995-103020995 on Build GRCh38
rs2454041 Chr.8: 103021284-103021284 on Build GRCh38
rs973313183 Chr.8: 103021168-103021168 on Build GRCh38
rs1014316366 Chr.8: 103021109-103021109 on Build GRCh38
rs868702869 Chr.8: 103021035-103021035 on Build GRCh38
rs564518733 Chr.8: 103020934-103020934 on Build GRCh38
rs956463542 Chr.8: 103020956-103020956 on Build GRCh38
rs1049069517 Chr.8: 103021181-103021181 on Build GRCh38
rs1001090305 Chr.8: 103021276-103021276 on Build GRCh38
rs966673954 Chr.8: 103020915-103020915 on Build GRCh38
rs765660444 Chr.8: 103020991-103020991 on Build GRCh38
rs538252279 Chr.8: 103021156-103021156 on Build GRCh38
rs763275657 Chr.8: 103021039-103021039 on Build GRCh38
rs988341085 Chr.8: 103020967-103020967 on Build GRCh38
rs570843340 Chr.8: 103021151-103021151 on Build GRCh38
rs531955098 Chr.8: 103021343-103021343 on Build GRCh38
rs1024761108 Chr.8: 103021313-103021313 on Build GRCh38
rs970627701 Chr.8: 103021149-103021149 on Build GRCh38
rs755119477 Chr.8: 103021102-103021102 on Build GRCh38
rs780862948 Chr.8: 103020999-103021000 on Build GRCh38
rs1049099508 Chr.8: 103021010-103021010 on Build GRCh38
rs953038524 Chr.8: 103021080-103021080 on Build GRCh38
rs1055777695 Chr.8: 103021074-103021074 on Build GRCh38
rs917775800 Chr.8: 103020972-103020972 on Build GRCh38
rs1051038827 Chr.8: 103021266-103021266 on Build GRCh38
rs1024787390 Chr.8: 103020949-103020949 on Build GRCh38
rs902004892 Chr.8: 103021251-103021251 on Build GRCh38
rs556947449 Chr.8: 103021223-103021223 on Build GRCh38

View Full Product Details