Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 231336110-231336654 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC8
Entrez Gene ID
149371
Gene Name
exocyst complex component 8
Gene Aliases
EXO84, Exo84p, SEC84
UniGene
Hs.356198
Gene Chromosome Location
Chr.1: 231332731-231337872 on Build GRCh38
Transcripts
NM_175876.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6125385 c.1174G>C p.E392Q Chr.1: 231336572-231336572 on Build GRCh38
244325 c.1109A>T p.K370I Chr.1: 231336637-231336637 on Build GRCh38
4610173 c.1141G>A p.A381T Chr.1: 231336605-231336605 on Build GRCh38
5958138 c.1446C>T p.D482D Chr.1: 231336300-231336300 on Build GRCh38
280904 c.1571T>C p.V524A Chr.1: 231336175-231336175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778916391 Chr.1: 231336527-231336527 on Build GRCh38
rs146816471 Chr.1: 231336303-231336303 on Build GRCh38
rs374927181 Chr.1: 231336419-231336419 on Build GRCh38
rs772450076 Chr.1: 231336430-231336430 on Build GRCh38
rs897881813 Chr.1: 231336364-231336364 on Build GRCh38
rs770749673 Chr.1: 231336456-231336456 on Build GRCh38
rs759212945 Chr.1: 231336480-231336480 on Build GRCh38
rs781704189 Chr.1: 231336576-231336576 on Build GRCh38
rs938951833 Chr.1: 231336227-231336227 on Build GRCh38
rs567286437 Chr.1: 231336217-231336217 on Build GRCh38
rs753947000 Chr.1: 231336409-231336409 on Build GRCh38
rs777719915 Chr.1: 231336407-231336407 on Build GRCh38
rs767378383 Chr.1: 231336382-231336382 on Build GRCh38
rs779083799 Chr.1: 231336417-231336417 on Build GRCh38
rs769095069 Chr.1: 231336567-231336567 on Build GRCh38
rs139010768 Chr.1: 231336268-231336268 on Build GRCh38
rs774762126 Chr.1: 231336291-231336291 on Build GRCh38
rs140965565 Chr.1: 231336394-231336394 on Build GRCh38
rs756017475 Chr.1: 231336397-231336397 on Build GRCh38
rs542256403 Chr.1: 231336202-231336202 on Build GRCh38
rs371315127 Chr.1: 231336550-231336550 on Build GRCh38
rs1037804650 Chr.1: 231336335-231336335 on Build GRCh38
rs774174436 Chr.1: 231336463-231336463 on Build GRCh38
rs749486063 Chr.1: 231336260-231336260 on Build GRCh38
rs376555975 Chr.1: 231336162-231336162 on Build GRCh38
rs376812212 Chr.1: 231336230-231336230 on Build GRCh38
rs760595810 Chr.1: 231336492-231336492 on Build GRCh38
rs755026621 Chr.1: 231336216-231336216 on Build GRCh38
rs762583523 Chr.1: 231336315-231336315 on Build GRCh38
rs748296740 Chr.1: 231336249-231336249 on Build GRCh38
rs147039275 Chr.1: 231336300-231336300 on Build GRCh38
rs762533589 Chr.1: 231336452-231336452 on Build GRCh38
rs769206630 Chr.1: 231336440-231336440 on Build GRCh38
rs200258755 Chr.1: 231336545-231336545 on Build GRCh38
rs375459948 Chr.1: 231336579-231336579 on Build GRCh38
rs373203929 Chr.1: 231336333-231336333 on Build GRCh38
rs775206321 Chr.1: 231336618-231336618 on Build GRCh38
rs369247477 Chr.1: 231336499-231336499 on Build GRCh38
rs929326623 Chr.1: 231336373-231336373 on Build GRCh38
rs752469576 Chr.1: 231336485-231336485 on Build GRCh38
rs759372445 Chr.1: 231336376-231336376 on Build GRCh38
rs772658865 Chr.1: 231336445-231336445 on Build GRCh38
rs750657660 Chr.1: 231336544-231336544 on Build GRCh38
rs756175779 Chr.1: 231336250-231336250 on Build GRCh38
rs202104918 Chr.1: 231336432-231336432 on Build GRCh38
rs770584042 Chr.1: 231336605-231336605 on Build GRCh38
rs751399432 Chr.1: 231336201-231336201 on Build GRCh38
rs764766498 Chr.1: 231336156-231336156 on Build GRCh38
rs753912067 Chr.1: 231336510-231336510 on Build GRCh38
rs745515473 Chr.1: 231336606-231336606 on Build GRCh38
rs148464625 Chr.1: 231336610-231336610 on Build GRCh38
rs564511057 Chr.1: 231336483-231336483 on Build GRCh38
rs760494680 Chr.1: 231336623-231336623 on Build GRCh38
rs769201213 Chr.1: 231336305-231336305 on Build GRCh38
rs1021390523 Chr.1: 231336625-231336625 on Build GRCh38
rs748623243 Chr.1: 231336583-231336583 on Build GRCh38
rs766687589 Chr.1: 231336415-231336415 on Build GRCh38
rs967491162 Chr.1: 231336626-231336626 on Build GRCh38
rs967081276 Chr.1: 231336144-231336144 on Build GRCh38
rs371887243 Chr.1: 231336288-231336288 on Build GRCh38
rs772609145 Chr.1: 231336297-231336297 on Build GRCh38
rs1024087352 Chr.1: 231336207-231336207 on Build GRCh38
rs141091711 Chr.1: 231336142-231336142 on Build GRCh38
rs776174754 Chr.1: 231336132-231336132 on Build GRCh38
rs373050702 Chr.1: 231336525-231336525 on Build GRCh38
rs928904202 Chr.1: 231336187-231336187 on Build GRCh38
rs766285156 Chr.1: 231336193-231336193 on Build GRCh38
rs1011793693 Chr.1: 231336603-231336603 on Build GRCh38
rs772715056 Chr.1: 231336311-231336311 on Build GRCh38
rs909036523 Chr.1: 231336371-231336371 on Build GRCh38
rs747389794 Chr.1: 231336558-231336558 on Build GRCh38
rs368929509 Chr.1: 231336296-231336296 on Build GRCh38
rs758010951 Chr.1: 231336183-231336183 on Build GRCh38
rs747475988 Chr.1: 231336437-231336437 on Build GRCh38
rs374958998 Chr.1: 231336252-231336252 on Build GRCh38
rs751450788 Chr.1: 231336334-231336334 on Build GRCh38
rs757532802 Chr.1: 231336410-231336410 on Build GRCh38

View Full Product Details