Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 247532377-247532853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCSAML
Entrez Gene ID
148823
Gene Name
germinal center associated signaling and motility like
Gene Aliases
C1orf150
UniGene
Hs.732113
Gene Chromosome Location
Chr.1: 247507058-247577690 on Build GRCh38
Transcripts
NM_001281834.1 NM_001281835.1 NM_001281836.1 NM_001281837.1 NM_001281838.1 NM_001281853.1 XM_017000399.1 NM_198074.4

Gene Symbol
OR2C3
Entrez Gene ID
81472
Gene Name
olfactory receptor family 2 subfamily C member 3
Gene Aliases
OR2C4, OR2C5P, OST742
UniGene
Hs.730218
Gene Chromosome Location
Chr.1: 247530132-247533839 on Build GRCh38
Transcripts
NM_001281834.1 NM_001281835.1 NM_001281836.1 NM_001281837.1 NM_001281838.1 NM_001281853.1 XM_017000399.1 NM_198074.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2232116 c.99delG p.S34fs*39 Chr.1: 247532413-247532413 on Build GRCh38
2232118 c.49G>T p.G17C Chr.1: 247532463-247532463 on Build GRCh38
232962 c.116C>T p.S39L Chr.1: 247532396-247532396 on Build GRCh38
1260121 c.76A>C p.T26P Chr.1: 247532436-247532436 on Build GRCh38
6861374 c.117G>T p.S39S Chr.1: 247532395-247532395 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs6697675 Chr.1: 247532656-247532656 on Build GRCh38
rs11582603 Chr.1: 247532820-247532820 on Build GRCh38
rs77580707 Chr.1: 247532574-247532574 on Build GRCh38
rs761876182 Chr.1: 247532530-247532530 on Build GRCh38
rs74657119 Chr.1: 247532612-247532612 on Build GRCh38
rs1040666184 Chr.1: 247532671-247532671 on Build GRCh38
rs963226511 Chr.1: 247532496-247532496 on Build GRCh38
rs6697472 Chr.1: 247532454-247532454 on Build GRCh38
rs879011369 Chr.1: 247532529-247532529 on Build GRCh38
rs201316551 Chr.1: 247532450-247532450 on Build GRCh38
rs754490526 Chr.1: 247532555-247532555 on Build GRCh38
rs534871422 Chr.1: 247532413-247532413 on Build GRCh38
rs747770729 Chr.1: 247532461-247532461 on Build GRCh38
rs770086261 Chr.1: 247532468-247532468 on Build GRCh38
rs780030916 Chr.1: 247532536-247532536 on Build GRCh38
rs767916298 Chr.1: 247532431-247532431 on Build GRCh38
rs1034257145 Chr.1: 247532827-247532827 on Build GRCh38
rs778698708 Chr.1: 247532439-247532439 on Build GRCh38
rs974336562 Chr.1: 247532516-247532516 on Build GRCh38
rs777178663 Chr.1: 247532520-247532520 on Build GRCh38
rs971757347 Chr.1: 247532444-247532444 on Build GRCh38
rs1018802180 Chr.1: 247532814-247532814 on Build GRCh38
rs750281288 Chr.1: 247532513-247532513 on Build GRCh38
rs1009983634 Chr.1: 247532780-247532780 on Build GRCh38
rs758364765 Chr.1: 247532715-247532715 on Build GRCh38
rs149639992 Chr.1: 247532427-247532427 on Build GRCh38
rs774996618 Chr.1: 247532488-247532488 on Build GRCh38
rs201859497 Chr.1: 247532790-247532790 on Build GRCh38
rs386641879 Chr.1: 247532454-247532455 on Build GRCh38
rs759745560 Chr.1: 247532493-247532493 on Build GRCh38
rs1052341089 Chr.1: 247532670-247532670 on Build GRCh38
rs1056144695 Chr.1: 247532589-247532589 on Build GRCh38
rs760906894 Chr.1: 247532502-247532502 on Build GRCh38
rs1033160677 Chr.1: 247532726-247532726 on Build GRCh38
rs145781714 Chr.1: 247532423-247532423 on Build GRCh38
rs757013130 Chr.1: 247532437-247532437 on Build GRCh38
rs752951002 Chr.1: 247532436-247532436 on Build GRCh38
rs761063792 Chr.1: 247532407-247532407 on Build GRCh38
rs763208283 Chr.1: 247532480-247532480 on Build GRCh38
rs764964001 Chr.1: 247532506-247532506 on Build GRCh38
rs765580448 Chr.1: 247532405-247532405 on Build GRCh38
rs61746303 Chr.1: 247532455-247532455 on Build GRCh38
rs773598629 Chr.1: 247532418-247532418 on Build GRCh38
rs10802521 Chr.1: 247532745-247532745 on Build GRCh38
rs138226636 Chr.1: 247532396-247532396 on Build GRCh38
rs773580993 Chr.1: 247532474-247532474 on Build GRCh38
rs1010214393 Chr.1: 247532467-247532467 on Build GRCh38
rs762144655 Chr.1: 247532402-247532402 on Build GRCh38
rs1029611734 Chr.1: 247532706-247532706 on Build GRCh38
rs767036972 Chr.1: 247532485-247532485 on Build GRCh38
rs761858556 Chr.1: 247532543-247532543 on Build GRCh38
rs76297624 Chr.1: 247532424-247532424 on Build GRCh38
rs144796478 Chr.1: 247532686-247532686 on Build GRCh38
rs118147664 Chr.1: 247532635-247532635 on Build GRCh38
rs758305368 Chr.1: 247532449-247532449 on Build GRCh38
rs149420965 Chr.1: 247532667-247532667 on Build GRCh38
rs879393989 Chr.1: 247532782-247532782 on Build GRCh38
rs900728292 Chr.1: 247532676-247532676 on Build GRCh38
rs1010160485 Chr.1: 247532697-247532697 on Build GRCh38
rs917638792 Chr.1: 247532602-247532602 on Build GRCh38
rs140172211 Chr.1: 247532662-247532662 on Build GRCh38
rs751442863 Chr.1: 247532549-247532549 on Build GRCh38
rs1006909088 Chr.1: 247532813-247532813 on Build GRCh38
rs767644272 Chr.1: 247532495-247532495 on Build GRCh38
rs746559906 Chr.1: 247532451-247532451 on Build GRCh38
rs996376237 Chr.1: 247532683-247532683 on Build GRCh38
rs918845196 Chr.1: 247532558-247532558 on Build GRCh38
rs985545269 Chr.1: 247532492-247532492 on Build GRCh38

View Full Product Details