Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 62813931-62814530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGR2
Entrez Gene ID
1959
Gene Name
early growth response 2
Gene Aliases
AT591, CMT1D, CMT4E, KROX20
UniGene
Hs.1395
Gene Chromosome Location
Chr.10: 62811996-62819167 on Build GRCh38
Transcripts
NM_000399.4 NM_001136177.2 NM_001136178.1 NM_001136179.2 NM_001321037.1 XM_011539427.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
684738 c.266C>T p.T89I Chr.10: 62814372-62814372 on Build GRCh38
3415165 c.322T>C p.Y108H Chr.10: 62814316-62814316 on Build GRCh38
1348562 c.559G>A p.A187T Chr.10: 62814079-62814079 on Build GRCh38
5726955 c.440G>A p.G147E Chr.10: 62814198-62814198 on Build GRCh38
1505934 c.616C>T p.P206S Chr.10: 62814022-62814022 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759939640 Chr.10: 62814222-62814222 on Build GRCh38
rs749981503 Chr.10: 62813957-62813957 on Build GRCh38
rs757301326 Chr.10: 62814178-62814178 on Build GRCh38
rs879254263 Chr.10: 62814298-62814298 on Build GRCh38
rs936268533 Chr.10: 62814025-62814025 on Build GRCh38
rs756375039 Chr.10: 62814137-62814137 on Build GRCh38
rs753161591 Chr.10: 62814133-62814133 on Build GRCh38
rs749421476 Chr.10: 62814013-62814013 on Build GRCh38
rs766849667 Chr.10: 62814102-62814102 on Build GRCh38
rs146631014 Chr.10: 62814446-62814446 on Build GRCh38
rs372491511 Chr.10: 62814078-62814078 on Build GRCh38
rs224083 Chr.10: 62814011-62814011 on Build GRCh38
rs768344815 Chr.10: 62814042-62814042 on Build GRCh38
rs746954913 Chr.10: 62814189-62814189 on Build GRCh38
rs769581130 Chr.10: 62814376-62814376 on Build GRCh38
rs747877927 Chr.10: 62814371-62814371 on Build GRCh38
rs886047095 Chr.10: 62814097-62814097 on Build GRCh38
rs1042629449 Chr.10: 62814436-62814436 on Build GRCh38
rs777521477 Chr.10: 62814487-62814487 on Build GRCh38
rs755853466 Chr.10: 62814478-62814478 on Build GRCh38
rs552315452 Chr.10: 62814213-62814213 on Build GRCh38
rs1057518238 Chr.10: 62814174-62814174 on Build GRCh38
rs768683610 Chr.10: 62814195-62814195 on Build GRCh38
rs139147487 Chr.10: 62813994-62813994 on Build GRCh38
rs540478439 Chr.10: 62814159-62814159 on Build GRCh38
rs752661994 Chr.10: 62814474-62814474 on Build GRCh38
rs147604283 Chr.10: 62814196-62814196 on Build GRCh38
rs755310380 Chr.10: 62813974-62813974 on Build GRCh38
rs940017681 Chr.10: 62814185-62814185 on Build GRCh38
rs764587581 Chr.10: 62814431-62814431 on Build GRCh38
rs768180179 Chr.10: 62814233-62814233 on Build GRCh38
rs563448629 Chr.10: 62814128-62814128 on Build GRCh38
rs750552347 Chr.10: 62814304-62814304 on Build GRCh38
rs750792592 Chr.10: 62814041-62814042 on Build GRCh38
rs781162110 Chr.10: 62814347-62814347 on Build GRCh38
rs997880732 Chr.10: 62814193-62814193 on Build GRCh38
rs756856193 Chr.10: 62814501-62814501 on Build GRCh38
rs764745834 Chr.10: 62813956-62813956 on Build GRCh38
rs753956133 Chr.10: 62814169-62814169 on Build GRCh38
rs1062545 Chr.10: 62814338-62814338 on Build GRCh38
rs778210403 Chr.10: 62814163-62814163 on Build GRCh38
rs756443238 Chr.10: 62814118-62814119 on Build GRCh38
rs144217451 Chr.10: 62814392-62814392 on Build GRCh38
rs144739493 Chr.10: 62814077-62814077 on Build GRCh38
rs749148233 Chr.10: 62814495-62814495 on Build GRCh38
rs1022830303 Chr.10: 62814008-62814008 on Build GRCh38
rs1035411608 Chr.10: 62814448-62814448 on Build GRCh38
rs530614586 Chr.10: 62813973-62813973 on Build GRCh38
rs143793213 Chr.10: 62814464-62814464 on Build GRCh38
rs778832811 Chr.10: 62814270-62814270 on Build GRCh38
rs932803146 Chr.10: 62814258-62814258 on Build GRCh38
rs1055082588 Chr.10: 62814030-62814030 on Build GRCh38
rs772044885 Chr.10: 62814026-62814026 on Build GRCh38
rs762505978 Chr.10: 62814099-62814099 on Build GRCh38
rs770378560 Chr.10: 62814218-62814218 on Build GRCh38
rs761429712 Chr.10: 62814089-62814089 on Build GRCh38
rs889580406 Chr.10: 62814209-62814209 on Build GRCh38
rs769421014 Chr.10: 62814212-62814212 on Build GRCh38
rs794727248 Chr.10: 62813959-62813959 on Build GRCh38
rs554815622 Chr.10: 62814409-62814409 on Build GRCh38
rs774872844 Chr.10: 62814219-62814219 on Build GRCh38
rs961388691 Chr.10: 62814252-62814253 on Build GRCh38
rs758869303 Chr.10: 62813968-62813968 on Build GRCh38
rs759904196 Chr.10: 62814109-62814109 on Build GRCh38
rs749799463 Chr.10: 62814427-62814429 on Build GRCh38
rs200913249 Chr.10: 62814098-62814098 on Build GRCh38
rs764356542 Chr.10: 62814242-62814242 on Build GRCh38
rs748000931 Chr.10: 62814208-62814208 on Build GRCh38
rs201068734 Chr.10: 62814416-62814416 on Build GRCh38
rs761182423 Chr.10: 62814240-62814240 on Build GRCh38
rs370084216 Chr.10: 62814104-62814104 on Build GRCh38
rs776212792 Chr.10: 62814074-62814074 on Build GRCh38
rs566397981 Chr.10: 62814377-62814377 on Build GRCh38
rs1021225769 Chr.10: 62813954-62813954 on Build GRCh38
rs766860417 Chr.10: 62813969-62813969 on Build GRCh38
rs780288079 Chr.10: 62814251-62814256 on Build GRCh38
rs1050343149 Chr.10: 62814353-62814353 on Build GRCh38
rs745451966 Chr.10: 62814415-62814415 on Build GRCh38
rs757323624 Chr.10: 62814268-62814268 on Build GRCh38
rs202183386 Chr.10: 62814181-62814181 on Build GRCh38
rs184558884 Chr.10: 62814282-62814282 on Build GRCh38
rs776156156 Chr.10: 62814428-62814428 on Build GRCh38
rs775350294 Chr.10: 62814029-62814029 on Build GRCh38
rs200488421 Chr.10: 62814360-62814360 on Build GRCh38
rs746763888 Chr.10: 62814035-62814035 on Build GRCh38
rs546268868 Chr.10: 62814307-62814307 on Build GRCh38
rs781408172 Chr.10: 62813993-62813993 on Build GRCh38
rs994194161 Chr.10: 62814254-62814254 on Build GRCh38
rs753108446 Chr.10: 62814235-62814235 on Build GRCh38
rs199906676 Chr.10: 62814256-62814256 on Build GRCh38
rs761905737 Chr.10: 62814433-62814433 on Build GRCh38
rs776869045 Chr.10: 62814432-62814432 on Build GRCh38
rs140201739 Chr.10: 62814454-62814454 on Build GRCh38
rs561006361 Chr.10: 62814076-62814076 on Build GRCh38
rs1017236982 Chr.10: 62814228-62814228 on Build GRCh38
rs773704923 Chr.10: 62814413-62814413 on Build GRCh38
rs769473870 Chr.10: 62814090-62814090 on Build GRCh38
rs201770282 Chr.10: 62814440-62814440 on Build GRCh38

View Full Product Details