Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 114400117-114400707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf71
Entrez Gene ID
54494
Gene Name
chromosome 11 open reading frame 71
Gene Aliases
URLC7
UniGene
Hs.715083
Gene Chromosome Location
Chr.11: 114391448-114400550 on Build GRCh38
Transcripts
NM_001271562.1 NM_019021.3 NM_001286045.1 NM_001286046.1 NM_001286047.1 NM_001286048.1 NM_016090.3

Gene Symbol
RBM7
Entrez Gene ID
10179
Gene Name
RNA binding motif protein 7
Gene Aliases
-
UniGene
Hs.7527
Gene Chromosome Location
Chr.11: 114400529-114410610 on Build GRCh38
Transcripts
NM_001271562.1 NM_019021.3 NM_001286045.1 NM_001286046.1 NM_001286047.1 NM_001286048.1 NM_016090.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3868471 c.137C>T p.P46L Chr.11: 114400195-114400195 on Build GRCh38
686284 c.6G>T p.G2G Chr.11: 114400677-114400677 on Build GRCh38
6444243 c.75C>A p.D25E Chr.11: 114400257-114400257 on Build GRCh38
3368221 c.28C>T p.R10C Chr.11: 114400699-114400699 on Build GRCh38
6552687 c.71G>A p.G24D Chr.11: 114400261-114400261 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566705091 Chr.11: 114400394-114400394 on Build GRCh38
rs750036277 Chr.11: 114400136-114400136 on Build GRCh38
rs578049841 Chr.11: 114400531-114400531 on Build GRCh38
rs958602147 Chr.11: 114400588-114400588 on Build GRCh38
rs9882 Chr.11: 114400243-114400243 on Build GRCh38
rs781557921 Chr.11: 114400256-114400256 on Build GRCh38
rs767142823 Chr.11: 114400631-114400631 on Build GRCh38
rs770896413 Chr.11: 114400671-114400671 on Build GRCh38
rs900623166 Chr.11: 114400426-114400426 on Build GRCh38
rs892728384 Chr.11: 114400680-114400680 on Build GRCh38
rs971600535 Chr.11: 114400399-114400399 on Build GRCh38
rs1001143431 Chr.11: 114400507-114400507 on Build GRCh38
rs375198468 Chr.11: 114400217-114400217 on Build GRCh38
rs115808383 Chr.11: 114400275-114400275 on Build GRCh38
rs200130893 Chr.11: 114400672-114400672 on Build GRCh38
rs747322947 Chr.11: 114400162-114400162 on Build GRCh38
rs779634437 Chr.11: 114400649-114400649 on Build GRCh38
rs763918045 Chr.11: 114400526-114400526 on Build GRCh38
rs749838579 Chr.11: 114400354-114400354 on Build GRCh38
rs1049513324 Chr.11: 114400538-114400538 on Build GRCh38
rs944179112 Chr.11: 114400492-114400492 on Build GRCh38
rs955945622 Chr.11: 114400316-114400316 on Build GRCh38
rs186294171 Chr.11: 114400546-114400546 on Build GRCh38
rs1032779003 Chr.11: 114400358-114400358 on Build GRCh38
rs1032047079 Chr.11: 114400457-114400457 on Build GRCh38
rs377553860 Chr.11: 114400303-114400303 on Build GRCh38
rs570461368 Chr.11: 114400355-114400355 on Build GRCh38
rs1014366269 Chr.11: 114400469-114400469 on Build GRCh38
rs368320968 Chr.11: 114400685-114400685 on Build GRCh38
rs528284748 Chr.11: 114400562-114400562 on Build GRCh38
rs188164114 Chr.11: 114400321-114400321 on Build GRCh38
rs921994914 Chr.11: 114400524-114400524 on Build GRCh38
rs774329256 Chr.11: 114400675-114400675 on Build GRCh38
rs374018167 Chr.11: 114400632-114400632 on Build GRCh38
rs747934533 Chr.11: 114400623-114400623 on Build GRCh38
rs772422711 Chr.11: 114400367-114400367 on Build GRCh38
rs944742083 Chr.11: 114400392-114400392 on Build GRCh38
rs761769384 Chr.11: 114400306-114400306 on Build GRCh38
rs886493884 Chr.11: 114400197-114400197 on Build GRCh38
rs192687124 Chr.11: 114400215-114400215 on Build GRCh38
rs550501384 Chr.11: 114400335-114400335 on Build GRCh38
rs151045096 Chr.11: 114400532-114400532 on Build GRCh38
rs181995153 Chr.11: 114400325-114400325 on Build GRCh38
rs759112663 Chr.11: 114400629-114400629 on Build GRCh38
rs754636336 Chr.11: 114400327-114400327 on Build GRCh38
rs951083780 Chr.11: 114400614-114400614 on Build GRCh38
rs369811614 Chr.11: 114400389-114400389 on Build GRCh38
rs552773786 Chr.11: 114400627-114400627 on Build GRCh38
rs540576834 Chr.11: 114400424-114400424 on Build GRCh38
rs891453793 Chr.11: 114400461-114400461 on Build GRCh38
rs779553438 Chr.11: 114400242-114400242 on Build GRCh38
rs765747087 Chr.11: 114400628-114400628 on Build GRCh38
rs762340956 Chr.11: 114400310-114400310 on Build GRCh38
rs1038036891 Chr.11: 114400298-114400298 on Build GRCh38
rs766998280 Chr.11: 114400317-114400317 on Build GRCh38
rs900119686 Chr.11: 114400347-114400347 on Build GRCh38
rs757789966 Chr.11: 114400642-114400642 on Build GRCh38
rs769815897 Chr.11: 114400201-114400201 on Build GRCh38
rs769670822 Chr.11: 114400625-114400625 on Build GRCh38
rs544285320 Chr.11: 114400496-114400496 on Build GRCh38
rs973691779 Chr.11: 114400449-114400449 on Build GRCh38
rs774402019 Chr.11: 114400287-114400287 on Build GRCh38
rs769488021 Chr.11: 114400644-114400644 on Build GRCh38
rs778408255 Chr.11: 114400272-114400272 on Build GRCh38
rs775080859 Chr.11: 114400687-114400687 on Build GRCh38
rs769289287 Chr.11: 114400652-114400652 on Build GRCh38
rs561777897 Chr.11: 114400195-114400195 on Build GRCh38
rs606671 Chr.11: 114400485-114400485 on Build GRCh38
rs765162349 Chr.11: 114400171-114400171 on Build GRCh38
rs1051885 Chr.11: 114400138-114400138 on Build GRCh38
rs764017168 Chr.11: 114400220-114400220 on Build GRCh38
rs779823587 Chr.11: 114400156-114400156 on Build GRCh38
rs369732707 Chr.11: 114400683-114400683 on Build GRCh38
rs773071626 Chr.11: 114400663-114400663 on Build GRCh38
rs112068263 Chr.11: 114400239-114400239 on Build GRCh38
rs892480422 Chr.11: 114400511-114400512 on Build GRCh38
rs371796136 Chr.11: 114400366-114400366 on Build GRCh38
rs1032419912 Chr.11: 114400172-114400172 on Build GRCh38
rs778123239 Chr.11: 114400495-114400495 on Build GRCh38
rs898242148 Chr.11: 114400302-114400302 on Build GRCh38
rs369588538 Chr.11: 114400288-114400288 on Build GRCh38
rs769428168 Chr.11: 114400662-114400662 on Build GRCh38
rs199945919 Chr.11: 114400307-114400307 on Build GRCh38
rs749900235 Chr.11: 114400234-114400234 on Build GRCh38
rs976511385 Chr.11: 114400518-114400518 on Build GRCh38
rs764740213 Chr.11: 114400231-114400231 on Build GRCh38
rs891730566 Chr.11: 114400594-114400594 on Build GRCh38
rs550322677 Chr.11: 114400597-114400597 on Build GRCh38
rs555631313 Chr.11: 114400437-114400437 on Build GRCh38
rs753267277 Chr.11: 114400471-114400471 on Build GRCh38
rs779150791 Chr.11: 114400364-114400364 on Build GRCh38
rs1047740983 Chr.11: 114400155-114400155 on Build GRCh38
rs772872779 Chr.11: 114400676-114400677 on Build GRCh38
rs980290847 Chr.11: 114400345-114400345 on Build GRCh38
rs773177096 Chr.11: 114400205-114400205 on Build GRCh38
rs768517083 Chr.11: 114400290-114400290 on Build GRCh38
rs752392310 Chr.11: 114400634-114400634 on Build GRCh38
rs758601777 Chr.11: 114400656-114400656 on Build GRCh38
rs748595867 Chr.11: 114400261-114400261 on Build GRCh38
rs745932550 Chr.11: 114400655-114400655 on Build GRCh38
rs969814596 Chr.11: 114400477-114400477 on Build GRCh38
rs371890478 Chr.11: 114400142-114400142 on Build GRCh38
rs1000122162 Chr.11: 114400453-114400453 on Build GRCh38
rs932466533 Chr.11: 114400435-114400435 on Build GRCh38
rs770288194 Chr.11: 114400268-114400268 on Build GRCh38
rs530791313 Chr.11: 114400595-114400595 on Build GRCh38
rs920824298 Chr.11: 114400362-114400362 on Build GRCh38
rs764547187 Chr.11: 114400636-114400636 on Build GRCh38
rs981250820 Chr.11: 114400400-114400400 on Build GRCh38
rs17629751 Chr.11: 114400500-114400500 on Build GRCh38
rs927207963 Chr.11: 114400420-114400420 on Build GRCh38
rs979435910 Chr.11: 114400165-114400165 on Build GRCh38
rs867490030 Chr.11: 114400493-114400493 on Build GRCh38
rs1045439533 Chr.11: 114400599-114400599 on Build GRCh38
rs754244257 Chr.11: 114400637-114400637 on Build GRCh38
rs957044303 Chr.11: 114400213-114400213 on Build GRCh38
rs1039880436 Chr.11: 114400635-114400635 on Build GRCh38
rs200550352 Chr.11: 114400331-114400331 on Build GRCh38
rs542189278 Chr.11: 114400564-114400564 on Build GRCh38
rs759658229 Chr.11: 114400409-114400409 on Build GRCh38
rs747289395 Chr.11: 114400661-114400661 on Build GRCh38
rs781642078 Chr.11: 114400189-114400189 on Build GRCh38
rs373835784 Chr.11: 114400244-114400244 on Build GRCh38
rs553174314 Chr.11: 114400390-114400390 on Build GRCh38
rs374455783 Chr.11: 114400294-114400294 on Build GRCh38
rs760519922 Chr.11: 114400219-114400219 on Build GRCh38
rs773253208 Chr.11: 114400309-114400309 on Build GRCh38
rs775916604 Chr.11: 114400381-114400381 on Build GRCh38
rs780393406 Chr.11: 114400658-114400658 on Build GRCh38
rs370321807 Chr.11: 114400382-114400382 on Build GRCh38
rs769057261 Chr.11: 114400188-114400188 on Build GRCh38
rs757994016 Chr.11: 114400145-114400145 on Build GRCh38
rs1027831992 Chr.11: 114400491-114400491 on Build GRCh38
rs892436760 Chr.11: 114400530-114400530 on Build GRCh38
rs749112066 Chr.11: 114400669-114400669 on Build GRCh38
rs564422718 Chr.11: 114400223-114400223 on Build GRCh38
rs561836324 Chr.11: 114400576-114400576 on Build GRCh38
rs900064352 Chr.11: 114400506-114400506 on Build GRCh38
rs368055793 Chr.11: 114400334-114400334 on Build GRCh38
rs904416802 Chr.11: 114400446-114400446 on Build GRCh38
rs544732282 Chr.11: 114400221-114400221 on Build GRCh38
rs754055816 Chr.11: 114400346-114400346 on Build GRCh38

View Full Product Details