Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111297109-111297694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLCA1
Entrez Gene ID
399948
Gene Name
colorectal cancer associated 1
Gene Aliases
C11orf92, CASC12, LOH11CR1F
UniGene
Hs.729225
Gene Chromosome Location
Chr.11: 111293389-111305048 on Build GRCh38
Transcripts
NM_001302644.1 NM_001302645.1 NM_001302646.1 NM_001302647.1 NM_001302648.1 NM_207429.3

SNP Detail

SNP ID Chromosome Location
rs765801134 Chr.11: 111297301-111297301 on Build GRCh38
rs1041612856 Chr.11: 111297466-111297466 on Build GRCh38
rs550418981 Chr.11: 111297459-111297459 on Build GRCh38
rs1043576641 Chr.11: 111297351-111297351 on Build GRCh38
rs946541776 Chr.11: 111297350-111297350 on Build GRCh38
rs773599195 Chr.11: 111297511-111297511 on Build GRCh38
rs770490133 Chr.11: 111297491-111297491 on Build GRCh38
rs571620327 Chr.11: 111297478-111297478 on Build GRCh38
rs898278238 Chr.11: 111297440-111297440 on Build GRCh38
rs956196099 Chr.11: 111297581-111297581 on Build GRCh38
rs1021573106 Chr.11: 111297609-111297609 on Build GRCh38
rs867908770 Chr.11: 111297670-111297670 on Build GRCh38
rs79667458 Chr.11: 111297562-111297562 on Build GRCh38
rs970772690 Chr.11: 111297357-111297357 on Build GRCh38
rs928604566 Chr.11: 111297244-111297244 on Build GRCh38
rs921065707 Chr.11: 111297667-111297667 on Build GRCh38
rs765922606 Chr.11: 111297160-111297160 on Build GRCh38
rs963161957 Chr.11: 111297142-111297142 on Build GRCh38
rs981662624 Chr.11: 111297542-111297542 on Build GRCh38
rs558452368 Chr.11: 111297219-111297219 on Build GRCh38
rs531889057 Chr.11: 111297439-111297439 on Build GRCh38
rs994391604 Chr.11: 111297306-111297306 on Build GRCh38
rs943769490 Chr.11: 111297464-111297464 on Build GRCh38
rs753226655 Chr.11: 111297303-111297303 on Build GRCh38
rs989151442 Chr.11: 111297605-111297605 on Build GRCh38
rs145981314 Chr.11: 111297611-111297611 on Build GRCh38
rs932595973 Chr.11: 111297386-111297386 on Build GRCh38
rs1003882813 Chr.11: 111297411-111297411 on Build GRCh38
rs190398957 Chr.11: 111297335-111297335 on Build GRCh38
rs766777381 Chr.11: 111297300-111297301 on Build GRCh38
rs955868008 Chr.11: 111297575-111297575 on Build GRCh38
rs1041228679 Chr.11: 111297225-111297225 on Build GRCh38
rs1003541375 Chr.11: 111297557-111297557 on Build GRCh38
rs1050979053 Chr.11: 111297392-111297392 on Build GRCh38
rs185243863 Chr.11: 111297236-111297236 on Build GRCh38
rs985113054 Chr.11: 111297444-111297444 on Build GRCh38
rs981273864 Chr.11: 111297371-111297371 on Build GRCh38
rs947504744 Chr.11: 111297638-111297638 on Build GRCh38
rs930234654 Chr.11: 111297481-111297481 on Build GRCh38
rs758889380 Chr.11: 111297635-111297635 on Build GRCh38
rs529861406 Chr.11: 111297339-111297339 on Build GRCh38
rs543737476 Chr.11: 111297190-111297190 on Build GRCh38
rs532525472 Chr.11: 111297509-111297509 on Build GRCh38
rs974277405 Chr.11: 111297156-111297156 on Build GRCh38
rs1023583461 Chr.11: 111297532-111297532 on Build GRCh38
rs142329453 Chr.11: 111297183-111297183 on Build GRCh38
rs1035954580 Chr.11: 111297550-111297550 on Build GRCh38
rs11213822 Chr.11: 111297299-111297299 on Build GRCh38
rs1027408983 Chr.11: 111297309-111297309 on Build GRCh38
rs899748301 Chr.11: 111297360-111297360 on Build GRCh38
rs1012231169 Chr.11: 111297512-111297512 on Build GRCh38
rs118177873 Chr.11: 111297346-111297346 on Build GRCh38
rs914635682 Chr.11: 111297615-111297615 on Build GRCh38
rs563330711 Chr.11: 111297397-111297397 on Build GRCh38
rs914664760 Chr.11: 111297308-111297308 on Build GRCh38
rs979217983 Chr.11: 111297642-111297642 on Build GRCh38
rs981421963 Chr.11: 111297203-111297203 on Build GRCh38
rs1035938543 Chr.11: 111297560-111297560 on Build GRCh38

View Full Product Details