Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 128461341-128461925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETS1
Entrez Gene ID
2113
Gene Name
ETS proto-oncogene 1, transcription factor
Gene Aliases
ETS-1, EWSR2, c-ets-1, p54
UniGene
Hs.369438
Gene Chromosome Location
Chr.11: 128458761-128587593 on Build GRCh38
Transcripts
NM_001143820.1 NM_001162422.1 NM_005238.3 XM_011542649.2 XM_011542650.1 XM_011542651.1 XM_011542652.1 XM_017017314.1 XM_017017315.1 XM_017017316.1 XM_017017317.1

SNP Detail

SNP ID Chromosome Location
rs922965633 Chr.11: 128461864-128461864 on Build GRCh38
rs115805775 Chr.11: 128461393-128461393 on Build GRCh38
rs960969641 Chr.11: 128461489-128461489 on Build GRCh38
rs867154240 Chr.11: 128461517-128461517 on Build GRCh38
rs1017473091 Chr.11: 128461757-128461758 on Build GRCh38
rs918995641 Chr.11: 128461542-128461542 on Build GRCh38
rs749102251 Chr.11: 128461704-128461704 on Build GRCh38
rs969966392 Chr.11: 128461741-128461741 on Build GRCh38
rs565010143 Chr.11: 128461625-128461625 on Build GRCh38
rs973031223 Chr.11: 128461793-128461793 on Build GRCh38
rs999086643 Chr.11: 128461460-128461460 on Build GRCh38
rs932998252 Chr.11: 128461869-128461869 on Build GRCh38
rs756953565 Chr.11: 128461724-128461724 on Build GRCh38
rs559216616 Chr.11: 128461509-128461510 on Build GRCh38
rs773465789 Chr.11: 128461396-128461396 on Build GRCh38
rs1016988075 Chr.11: 128461653-128461653 on Build GRCh38
rs78505561 Chr.11: 128461555-128461555 on Build GRCh38
rs928797848 Chr.11: 128461573-128461573 on Build GRCh38
rs762261499 Chr.11: 128461379-128461379 on Build GRCh38
rs763171901 Chr.11: 128461469-128461469 on Build GRCh38
rs1030850154 Chr.11: 128461647-128461648 on Build GRCh38
rs112417808 Chr.11: 128461843-128461843 on Build GRCh38
rs910440710 Chr.11: 128461890-128461890 on Build GRCh38
rs1023852584 Chr.11: 128461740-128461740 on Build GRCh38
rs1006013736 Chr.11: 128461649-128461649 on Build GRCh38
rs888986385 Chr.11: 128461639-128461639 on Build GRCh38
rs544394233 Chr.11: 128461532-128461532 on Build GRCh38
rs551766745 Chr.11: 128461621-128461621 on Build GRCh38
rs1003151911 Chr.11: 128461371-128461371 on Build GRCh38
rs934995349 Chr.11: 128461586-128461586 on Build GRCh38
rs76139003 Chr.11: 128461758-128461758 on Build GRCh38
rs74338971 Chr.11: 128461535-128461535 on Build GRCh38
rs962694273 Chr.11: 128461404-128461404 on Build GRCh38
rs933261196 Chr.11: 128461624-128461624 on Build GRCh38
rs1025871813 Chr.11: 128461470-128461470 on Build GRCh38
rs966961091 Chr.11: 128461402-128461402 on Build GRCh38
rs192439666 Chr.11: 128461859-128461859 on Build GRCh38
rs910477721 Chr.11: 128461530-128461530 on Build GRCh38
rs529463514 Chr.11: 128461762-128461762 on Build GRCh38
rs992411193 Chr.11: 128461499-128461499 on Build GRCh38
rs370028742 Chr.11: 128461807-128461807 on Build GRCh38
rs927708752 Chr.11: 128461889-128461890 on Build GRCh38
rs997376650 Chr.11: 128461383-128461383 on Build GRCh38
rs550720487 Chr.11: 128461747-128461747 on Build GRCh38
rs141331615 Chr.11: 128461594-128461594 on Build GRCh38
rs1043432215 Chr.11: 128461897-128461897 on Build GRCh38
rs1050773940 Chr.11: 128461886-128461886 on Build GRCh38
rs745652336 Chr.11: 128461729-128461729 on Build GRCh38
rs75220088 Chr.11: 128461544-128461544 on Build GRCh38
rs960733943 Chr.11: 128461717-128461717 on Build GRCh38
rs991694824 Chr.11: 128461895-128461895 on Build GRCh38
rs74489553 Chr.11: 128461688-128461688 on Build GRCh38
rs533564714 Chr.11: 128461808-128461808 on Build GRCh38
rs967526863 Chr.11: 128461410-128461410 on Build GRCh38
rs992862131 Chr.11: 128461728-128461728 on Build GRCh38
rs938791901 Chr.11: 128461890-128461890 on Build GRCh38
rs559675527 Chr.11: 128461585-128461585 on Build GRCh38
rs1016971626 Chr.11: 128461397-128461397 on Build GRCh38
rs765592515 Chr.11: 128461384-128461384 on Build GRCh38
rs898563136 Chr.11: 128461687-128461687 on Build GRCh38
rs941877482 Chr.11: 128461896-128461896 on Build GRCh38

View Full Product Details