Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32203681-32204229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC28B
Entrez Gene ID
79140
Gene Name
coiled-coil domain containing 28B
Gene Aliases
-
UniGene
Hs.534482
Gene Chromosome Location
Chr.1: 32200386-32205390 on Build GRCh38
Transcripts
NM_001301011.1 NM_024296.4 XM_011542113.2 XM_017002306.1 XM_017002307.1 XM_017002308.1 XM_017002309.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
426088 c.211G>T p.G71C Chr.1: 32203925-32203925 on Build GRCh38
4533940 c.202G>A p.G68S Chr.1: 32203916-32203916 on Build GRCh38
6622034 c.315C>T p.G105G Chr.1: 32204029-32204029 on Build GRCh38
6622035 c.317G>A p.R106Q Chr.1: 32204031-32204031 on Build GRCh38
4543428 c.337G>A p.E113K Chr.1: 32204191-32204191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925987936 Chr.1: 32203786-32203789 on Build GRCh38
rs952595550 Chr.1: 32204161-32204161 on Build GRCh38
rs143254196 Chr.1: 32203843-32203843 on Build GRCh38
rs758268177 Chr.1: 32203960-32203960 on Build GRCh38
rs562912411 Chr.1: 32204188-32204188 on Build GRCh38
rs771285741 Chr.1: 32203834-32203834 on Build GRCh38
rs763825042 Chr.1: 32203857-32203857 on Build GRCh38
rs770590932 Chr.1: 32204196-32204196 on Build GRCh38
rs759498949 Chr.1: 32204070-32204070 on Build GRCh38
rs753894609 Chr.1: 32204141-32204141 on Build GRCh38
rs202075564 Chr.1: 32203933-32203933 on Build GRCh38
rs777850558 Chr.1: 32203990-32203990 on Build GRCh38
rs751519072 Chr.1: 32203966-32203966 on Build GRCh38
rs771234393 Chr.1: 32203904-32203904 on Build GRCh38
rs895573256 Chr.1: 32203899-32203899 on Build GRCh38
rs978778977 Chr.1: 32203783-32203783 on Build GRCh38
rs753612588 Chr.1: 32203860-32203860 on Build GRCh38
rs756127479 Chr.1: 32203976-32203976 on Build GRCh38
rs368003386 Chr.1: 32203838-32203838 on Build GRCh38
rs988787948 Chr.1: 32204132-32204132 on Build GRCh38
rs908238425 Chr.1: 32203864-32203864 on Build GRCh38
rs769261061 Chr.1: 32204184-32204184 on Build GRCh38
rs764812524 Chr.1: 32204175-32204175 on Build GRCh38
rs762687710 Chr.1: 32204193-32204193 on Build GRCh38
rs199919893 Chr.1: 32203872-32203872 on Build GRCh38
rs767430968 Chr.1: 32204209-32204209 on Build GRCh38
rs556568897 Chr.1: 32203880-32203880 on Build GRCh38
rs746285494 Chr.1: 32203911-32203911 on Build GRCh38
rs775797765 Chr.1: 32204013-32204013 on Build GRCh38
rs763295978 Chr.1: 32203833-32203833 on Build GRCh38
rs767016937 Chr.1: 32203841-32203841 on Build GRCh38
rs1000797794 Chr.1: 32203708-32203708 on Build GRCh38
rs965493384 Chr.1: 32204131-32204131 on Build GRCh38
rs774731331 Chr.1: 32203905-32203905 on Build GRCh38
rs747436955 Chr.1: 32204178-32204178 on Build GRCh38
rs41263993 Chr.1: 32204044-32204044 on Build GRCh38
rs772436511 Chr.1: 32203924-32203924 on Build GRCh38
rs777877169 Chr.1: 32204084-32204107 on Build GRCh38
rs752877740 Chr.1: 32203757-32203757 on Build GRCh38
rs766218771 Chr.1: 32203964-32203964 on Build GRCh38
rs778998750 Chr.1: 32204153-32204153 on Build GRCh38
rs754198114 Chr.1: 32203813-32203813 on Build GRCh38
rs913800131 Chr.1: 32204172-32204172 on Build GRCh38
rs769306908 Chr.1: 32203782-32203785 on Build GRCh38
rs757047485 Chr.1: 32203861-32203861 on Build GRCh38
rs199647520 Chr.1: 32204190-32204190 on Build GRCh38
rs772559410 Chr.1: 32204011-32204011 on Build GRCh38
rs776248241 Chr.1: 32204030-32204030 on Build GRCh38
rs140759385 Chr.1: 32203828-32203828 on Build GRCh38
rs757387251 Chr.1: 32204151-32204151 on Build GRCh38
rs778638794 Chr.1: 32203898-32203898 on Build GRCh38
rs756155701 Chr.1: 32204081-32204082 on Build GRCh38
rs776367978 Chr.1: 32203936-32203936 on Build GRCh38
rs371830706 Chr.1: 32203935-32203935 on Build GRCh38
rs529593244 Chr.1: 32203801-32203801 on Build GRCh38
rs751403021 Chr.1: 32204067-32204067 on Build GRCh38
rs6697312 Chr.1: 32203823-32203823 on Build GRCh38
rs1032318020 Chr.1: 32203714-32203714 on Build GRCh38
rs200163249 Chr.1: 32203995-32203995 on Build GRCh38
rs559984271 Chr.1: 32203867-32203867 on Build GRCh38
rs748162864 Chr.1: 32203878-32203878 on Build GRCh38
rs761454975 Chr.1: 32204031-32204031 on Build GRCh38
rs561982752 Chr.1: 32204023-32204023 on Build GRCh38
rs574977655 Chr.1: 32203881-32203881 on Build GRCh38
rs988207779 Chr.1: 32203761-32203761 on Build GRCh38
rs202021004 Chr.1: 32204071-32204071 on Build GRCh38
rs934683610 Chr.1: 32203796-32203796 on Build GRCh38
rs1020083515 Chr.1: 32204116-32204116 on Build GRCh38
rs142367160 Chr.1: 32204207-32204207 on Build GRCh38
rs562940903 Chr.1: 32203740-32203740 on Build GRCh38
rs979217136 Chr.1: 32204055-32204055 on Build GRCh38
rs534133056 Chr.1: 32203840-32203840 on Build GRCh38
rs745773314 Chr.1: 32204028-32204028 on Build GRCh38
rs202020131 Chr.1: 32203939-32203939 on Build GRCh38
rs191894571 Chr.1: 32203856-32203856 on Build GRCh38
rs766209681 Chr.1: 32204062-32204062 on Build GRCh38
rs375463372 Chr.1: 32203938-32203938 on Build GRCh38
rs758635766 Chr.1: 32204169-32204169 on Build GRCh38
rs183850860 Chr.1: 32204137-32204137 on Build GRCh38
rs750344185 Chr.1: 32203868-32203868 on Build GRCh38
rs1001100101 Chr.1: 32203994-32203994 on Build GRCh38
rs774162573 Chr.1: 32204198-32204198 on Build GRCh38
rs943984774 Chr.1: 32204179-32204179 on Build GRCh38
rs201881362 Chr.1: 32203943-32203943 on Build GRCh38
rs749497454 Chr.1: 32204170-32204171 on Build GRCh38
rs758235327 Chr.1: 32203871-32203871 on Build GRCh38
rs747652526 Chr.1: 32204186-32204186 on Build GRCh38
rs762587976 Chr.1: 32204046-32204046 on Build GRCh38
rs750666303 Chr.1: 32204156-32204156 on Build GRCh38
rs760103282 Chr.1: 32203932-32203932 on Build GRCh38
rs145792550 Chr.1: 32203925-32203925 on Build GRCh38
rs749470747 Chr.1: 32203991-32203991 on Build GRCh38
rs373803490 Chr.1: 32203831-32203831 on Build GRCh38
rs370036102 Chr.1: 32204185-32204185 on Build GRCh38
rs772979728 Chr.1: 32204045-32204045 on Build GRCh38
rs377238220 Chr.1: 32204183-32204183 on Build GRCh38

View Full Product Details