Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 68474740-68475071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEPDC1
Entrez Gene ID
55635
Gene Name
DEP domain containing 1
Gene Aliases
DEP.8, DEPDC1-V2, DEPDC1A, SDP35
UniGene
Hs.445098
Gene Chromosome Location
Chr.1: 68474152-68497221 on Build GRCh38
Transcripts
NM_001114120.2 NM_017779.5

SNP Detail

SNP ID Chromosome Location
rs534526782 Chr.1: 68475034-68475034 on Build GRCh38
rs748835138 Chr.1: 68475024-68475024 on Build GRCh38
rs116436345 Chr.1: 68474913-68474913 on Build GRCh38
rs776766168 Chr.1: 68474862-68474862 on Build GRCh38
rs971140480 Chr.1: 68474902-68474902 on Build GRCh38
rs958644589 Chr.1: 68474879-68474879 on Build GRCh38
rs1053989136 Chr.1: 68474941-68474941 on Build GRCh38
rs528841643 Chr.1: 68474893-68474893 on Build GRCh38
rs552031895 Chr.1: 68475006-68475006 on Build GRCh38
rs890524001 Chr.1: 68474828-68474828 on Build GRCh38
rs1027520580 Chr.1: 68474871-68474871 on Build GRCh38
rs56150903 Chr.1: 68474909-68474909 on Build GRCh38
rs571722774 Chr.1: 68475015-68475015 on Build GRCh38
rs890689561 Chr.1: 68474987-68474987 on Build GRCh38
rs754444622 Chr.1: 68474865-68474868 on Build GRCh38
rs942086648 Chr.1: 68474906-68474906 on Build GRCh38
rs1017736566 Chr.1: 68475037-68475037 on Build GRCh38
rs968427794 Chr.1: 68474857-68474857 on Build GRCh38
rs12757862 Chr.1: 68474802-68474802 on Build GRCh38
rs894099086 Chr.1: 68474798-68474798 on Build GRCh38
rs531419092 Chr.1: 68475002-68475002 on Build GRCh38
rs915481825 Chr.1: 68474956-68474956 on Build GRCh38
rs1007706436 Chr.1: 68474840-68474840 on Build GRCh38
rs558830553 Chr.1: 68474831-68474831 on Build GRCh38
rs114500445 Chr.1: 68474959-68474959 on Build GRCh38
rs1050882979 Chr.1: 68474807-68474807 on Build GRCh38
rs533285544 Chr.1: 68474806-68474806 on Build GRCh38
rs540997097 Chr.1: 68474905-68474905 on Build GRCh38
rs1009410500 Chr.1: 68474842-68474842 on Build GRCh38
rs867834912 Chr.1: 68474868-68474868 on Build GRCh38
rs1044181084 Chr.1: 68474858-68474858 on Build GRCh38
rs1030572670 Chr.1: 68474801-68474801 on Build GRCh38
rs12737443 Chr.1: 68474800-68474800 on Build GRCh38
rs75750431 Chr.1: 68474803-68474803 on Build GRCh38
rs17097400 Chr.1: 68474883-68474883 on Build GRCh38
rs74083708 Chr.1: 68474887-68474887 on Build GRCh38
rs1017789887 Chr.1: 68474847-68474848 on Build GRCh38

View Full Product Details