Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 62237943-62238542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANK4
Entrez Gene ID
163782
Gene Name
KN motif and ankyrin repeat domains 4
Gene Aliases
ANKRD38, dJ1078M7.1
UniGene
Hs.283398
Gene Chromosome Location
Chr.1: 62236165-62319414 on Build GRCh38
Transcripts
NM_001320269.1 NM_181712.4 XM_011540843.2 XM_011540845.2 XM_017000483.1 XM_017000484.1 XM_017000485.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7372913 c.2924C>A p.T975N Chr.1: 62238341-62238341 on Build GRCh38
2238134 c.2978T>G p.L993R Chr.1: 62238287-62238287 on Build GRCh38
5462520 c.2890C>T p.R964C Chr.1: 62238375-62238375 on Build GRCh38
3419369 c.2908G>A p.A970T Chr.1: 62238357-62238357 on Build GRCh38
5614027 c.2965C>A p.Q989K Chr.1: 62238300-62238300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145877699 Chr.1: 62238335-62238335 on Build GRCh38
rs778192341 Chr.1: 62238306-62238306 on Build GRCh38
rs972828370 Chr.1: 62238463-62238464 on Build GRCh38
rs988987730 Chr.1: 62238174-62238174 on Build GRCh38
rs1029861707 Chr.1: 62238344-62238344 on Build GRCh38
rs973672675 Chr.1: 62238382-62238382 on Build GRCh38
rs779338939 Chr.1: 62238161-62238161 on Build GRCh38
rs34116366 Chr.1: 62238214-62238214 on Build GRCh38
rs747654760 Chr.1: 62238427-62238427 on Build GRCh38
rs1014081062 Chr.1: 62238291-62238291 on Build GRCh38
rs144097182 Chr.1: 62238280-62238280 on Build GRCh38
rs200811588 Chr.1: 62238375-62238375 on Build GRCh38
rs953897991 Chr.1: 62238442-62238442 on Build GRCh38
rs985367537 Chr.1: 62238451-62238451 on Build GRCh38
rs532344987 Chr.1: 62238219-62238219 on Build GRCh38
rs778753535 Chr.1: 62238420-62238420 on Build GRCh38
rs76827207 Chr.1: 62238509-62238509 on Build GRCh38
rs920842632 Chr.1: 62238424-62238424 on Build GRCh38
rs758867090 Chr.1: 62238484-62238484 on Build GRCh38
rs769836890 Chr.1: 62238235-62238235 on Build GRCh38
rs377403827 Chr.1: 62238249-62238249 on Build GRCh38
rs553096133 Chr.1: 62238433-62238433 on Build GRCh38
rs922360865 Chr.1: 62238182-62238182 on Build GRCh38
rs950146384 Chr.1: 62238483-62238483 on Build GRCh38
rs151151056 Chr.1: 62238307-62238307 on Build GRCh38
rs545908915 Chr.1: 62238074-62238074 on Build GRCh38
rs999969496 Chr.1: 62238495-62238495 on Build GRCh38
rs891705134 Chr.1: 62238508-62238508 on Build GRCh38
rs768235868 Chr.1: 62238248-62238248 on Build GRCh38
rs781040356 Chr.1: 62238229-62238229 on Build GRCh38
rs541342625 Chr.1: 62238220-62238220 on Build GRCh38
rs557407389 Chr.1: 62238521-62238521 on Build GRCh38
rs189629688 Chr.1: 62238183-62238183 on Build GRCh38
rs766738410 Chr.1: 62238372-62238372 on Build GRCh38
rs185617490 Chr.1: 62238015-62238015 on Build GRCh38
rs41289438 Chr.1: 62238462-62238462 on Build GRCh38
rs1048156391 Chr.1: 62238494-62238494 on Build GRCh38
rs751853926 Chr.1: 62238401-62238401 on Build GRCh38
rs978611390 Chr.1: 62238089-62238089 on Build GRCh38
rs141702470 Chr.1: 62238357-62238357 on Build GRCh38
rs1037926400 Chr.1: 62238244-62238244 on Build GRCh38
rs369274343 Chr.1: 62238033-62238033 on Build GRCh38
rs954995330 Chr.1: 62238522-62238522 on Build GRCh38
rs748768364 Chr.1: 62238246-62238246 on Build GRCh38
rs1029336504 Chr.1: 62238514-62238514 on Build GRCh38
rs1037229589 Chr.1: 62237967-62237967 on Build GRCh38
rs17123220 Chr.1: 62238014-62238014 on Build GRCh38
rs1026372904 Chr.1: 62237984-62237984 on Build GRCh38
rs780002485 Chr.1: 62238411-62238411 on Build GRCh38
rs569683686 Chr.1: 62238355-62238355 on Build GRCh38
rs752783751 Chr.1: 62238304-62238304 on Build GRCh38
rs753644289 Chr.1: 62238415-62238415 on Build GRCh38
rs557192350 Chr.1: 62238028-62238028 on Build GRCh38
rs765361445 Chr.1: 62238299-62238299 on Build GRCh38
rs759036725 Chr.1: 62238259-62238259 on Build GRCh38
rs993999974 Chr.1: 62237972-62237972 on Build GRCh38
rs780091111 Chr.1: 62238333-62238333 on Build GRCh38
rs773897694 Chr.1: 62238338-62238338 on Build GRCh38
rs994080404 Chr.1: 62237986-62237986 on Build GRCh38
rs116331058 Chr.1: 62238358-62238358 on Build GRCh38
rs761068420 Chr.1: 62238268-62238268 on Build GRCh38
rs866212071 Chr.1: 62238340-62238340 on Build GRCh38
rs34591898 Chr.1: 62238305-62238305 on Build GRCh38
rs1013104657 Chr.1: 62238346-62238346 on Build GRCh38
rs548622982 Chr.1: 62237969-62237969 on Build GRCh38
rs888050276 Chr.1: 62238011-62238011 on Build GRCh38
rs773542476 Chr.1: 62238257-62238257 on Build GRCh38
rs548272273 Chr.1: 62238314-62238314 on Build GRCh38
rs764374922 Chr.1: 62238393-62238393 on Build GRCh38
rs570609607 Chr.1: 62238369-62238369 on Build GRCh38
rs555909206 Chr.1: 62238248-62238249 on Build GRCh38
rs758452262 Chr.1: 62238379-62238379 on Build GRCh38
rs754950633 Chr.1: 62238289-62238289 on Build GRCh38
rs899130040 Chr.1: 62237970-62237970 on Build GRCh38
rs905723675 Chr.1: 62238486-62238486 on Build GRCh38
rs1027823267 Chr.1: 62238009-62238009 on Build GRCh38
rs943607815 Chr.1: 62238226-62238226 on Build GRCh38
rs746032364 Chr.1: 62238256-62238256 on Build GRCh38
rs754361735 Chr.1: 62238286-62238286 on Build GRCh38
rs755916041 Chr.1: 62238404-62238404 on Build GRCh38
rs373930045 Chr.1: 62238429-62238429 on Build GRCh38
rs546890587 Chr.1: 62238417-62238417 on Build GRCh38
rs1044585280 Chr.1: 62238511-62238511 on Build GRCh38
rs1033373444 Chr.1: 62238500-62238500 on Build GRCh38
rs749237761 Chr.1: 62237987-62237987 on Build GRCh38
rs745717698 Chr.1: 62238233-62238233 on Build GRCh38
rs987631997 Chr.1: 62238218-62238218 on Build GRCh38
rs1032648095 Chr.1: 62238131-62238131 on Build GRCh38
rs151055866 Chr.1: 62238374-62238374 on Build GRCh38

View Full Product Details